Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96808688-96809196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM4
Entrez Gene ID
26504
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
ACDP4
UniGene
Hs.175043
Gene Chromosome Location
Chr.2: 96760902-96811891 on Build GRCh38
Transcripts
NM_020184.3 XM_005263914.3 XM_005263915.3 XM_017003799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4096437 c.553T>G p.F185V Chr.2: 96808704-96808704 on Build GRCh38
4134148 c.2093T>G p.F698C Chr.2: 96808705-96808705 on Build GRCh38
6996406 c.2105C>T p.A702V Chr.2: 96808717-96808717 on Build GRCh38
4096436 c.2092T>G p.F698V Chr.2: 96808704-96808704 on Build GRCh38
4640469 c.2101C>T p.R701W Chr.2: 96808713-96808713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867658390 Chr.2: 96809051-96809051 on Build GRCh38
rs999212743 Chr.2: 96808736-96808736 on Build GRCh38
rs766235218 Chr.2: 96809003-96809003 on Build GRCh38
rs200517269 Chr.2: 96808747-96808747 on Build GRCh38
rs748576674 Chr.2: 96809122-96809122 on Build GRCh38
rs372182086 Chr.2: 96808713-96808713 on Build GRCh38
rs571280008 Chr.2: 96809138-96809138 on Build GRCh38
rs994148263 Chr.2: 96809102-96809102 on Build GRCh38
rs774796340 Chr.2: 96808731-96808731 on Build GRCh38
rs988296856 Chr.2: 96808880-96808880 on Build GRCh38
rs891188839 Chr.2: 96809117-96809117 on Build GRCh38
rs917882673 Chr.2: 96809003-96809003 on Build GRCh38
rs950496948 Chr.2: 96809120-96809120 on Build GRCh38
rs905140812 Chr.2: 96809079-96809079 on Build GRCh38
rs796761028 Chr.2: 96809002-96809003 on Build GRCh38
rs760782158 Chr.2: 96808762-96808762 on Build GRCh38
rs776720198 Chr.2: 96808765-96808765 on Build GRCh38
rs748791059 Chr.2: 96808837-96808837 on Build GRCh38
rs769590753 Chr.2: 96808722-96808722 on Build GRCh38
rs547057101 Chr.2: 96809127-96809127 on Build GRCh38
rs199613079 Chr.2: 96808789-96808789 on Build GRCh38
rs367620019 Chr.2: 96808778-96808778 on Build GRCh38
rs764069917 Chr.2: 96808786-96808786 on Build GRCh38
rs909487111 Chr.2: 96808934-96808934 on Build GRCh38
rs757562113 Chr.2: 96808763-96808764 on Build GRCh38
rs917638678 Chr.2: 96808892-96808892 on Build GRCh38
rs1032559270 Chr.2: 96809162-96809162 on Build GRCh38
rs552796110 Chr.2: 96809150-96809150 on Build GRCh38
rs114718472 Chr.2: 96808840-96808840 on Build GRCh38
rs569368351 Chr.2: 96809165-96809165 on Build GRCh38
rs756325462 Chr.2: 96808721-96808721 on Build GRCh38
rs191111547 Chr.2: 96808957-96808957 on Build GRCh38
rs866429658 Chr.2: 96808883-96808883 on Build GRCh38
rs758188369 Chr.2: 96808784-96808784 on Build GRCh38
rs977304321 Chr.2: 96808953-96808953 on Build GRCh38
rs545287155 Chr.2: 96808954-96808954 on Build GRCh38
rs558898866 Chr.2: 96809174-96809174 on Build GRCh38
rs766406912 Chr.2: 96808764-96808764 on Build GRCh38
rs74930746 Chr.2: 96809143-96809143 on Build GRCh38
rs530201570 Chr.2: 96809088-96809088 on Build GRCh38
rs749059747 Chr.2: 96808776-96808776 on Build GRCh38
rs538407064 Chr.2: 96809166-96809166 on Build GRCh38
rs941939142 Chr.2: 96809080-96809080 on Build GRCh38
rs369472918 Chr.2: 96808999-96808999 on Build GRCh38
rs1008424784 Chr.2: 96809132-96809132 on Build GRCh38
rs544399752 Chr.2: 96809016-96809016 on Build GRCh38
rs554845203 Chr.2: 96809167-96809167 on Build GRCh38
rs141935727 Chr.2: 96809064-96809064 on Build GRCh38
rs919456123 Chr.2: 96808864-96808864 on Build GRCh38
rs949232745 Chr.2: 96808935-96808935 on Build GRCh38
rs572844826 Chr.2: 96808866-96808866 on Build GRCh38
rs866803728 Chr.2: 96809033-96809033 on Build GRCh38
rs181963918 Chr.2: 96809142-96809142 on Build GRCh38
rs762272088 Chr.2: 96808739-96808739 on Build GRCh38
rs898486949 Chr.2: 96809099-96809099 on Build GRCh38
rs1044852267 Chr.2: 96809069-96809069 on Build GRCh38
rs759699161 Chr.2: 96808766-96808766 on Build GRCh38
rs1042466973 Chr.2: 96809042-96809042 on Build GRCh38
rs773645844 Chr.2: 96808758-96808758 on Build GRCh38
rs757251824 Chr.2: 96808793-96808793 on Build GRCh38
rs965676387 Chr.2: 96809089-96809089 on Build GRCh38
rs1007393250 Chr.2: 96809087-96809087 on Build GRCh38
rs985777525 Chr.2: 96808774-96808774 on Build GRCh38
rs903011513 Chr.2: 96808712-96808712 on Build GRCh38
rs754035021 Chr.2: 96808761-96808762 on Build GRCh38
rs1026163048 Chr.2: 96809112-96809112 on Build GRCh38

View Full Product Details