Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10449979-10450475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APITD1-CORT
Entrez Gene ID
100526739
Gene Name
APITD1-CORT readthrough
Gene Aliases
-
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10452003 on Build GRCh38
Transcripts
NM_001243768.1 NM_001270517.1 NM_198544.3 NM_199006.2 NR_037187.1 NM_001302.4

Gene Symbol
CORT
Entrez Gene ID
1325
Gene Name
cortistatin
Gene Aliases
CST-14, CST-17, CST-29
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10449719-10452003 on Build GRCh38
Transcripts
NM_001243768.1 NM_001270517.1 NM_198544.3 NM_199006.2 NR_037187.1 NM_001302.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6281223 c.195C>T p.A65A Chr.1: 10450268-10450268 on Build GRCh38
4020067 c.3G>A p.M1I Chr.1: 10450076-10450076 on Build GRCh38
6496493 c.198G>A p.T66T Chr.1: 10450271-10450271 on Build GRCh38
4020068 c.164C>A p.P55H Chr.1: 10450237-10450237 on Build GRCh38
4626715 c.234C>T p.T78T Chr.1: 10450307-10450307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761036242 Chr.1: 10450278-10450278 on Build GRCh38
rs749301910 Chr.1: 10450180-10450180 on Build GRCh38
rs746163861 Chr.1: 10450169-10450169 on Build GRCh38
rs775559314 Chr.1: 10450172-10450172 on Build GRCh38
rs978204376 Chr.1: 10450403-10450403 on Build GRCh38
rs554414328 Chr.1: 10450124-10450124 on Build GRCh38
rs776039862 Chr.1: 10450084-10450084 on Build GRCh38
rs777360908 Chr.1: 10450041-10450047 on Build GRCh38
rs560545465 Chr.1: 10450398-10450398 on Build GRCh38
rs1053963881 Chr.1: 10450436-10450436 on Build GRCh38
rs772545408 Chr.1: 10450276-10450276 on Build GRCh38
rs763434278 Chr.1: 10450085-10450085 on Build GRCh38
rs142020907 Chr.1: 10450154-10450154 on Build GRCh38
rs772464701 Chr.1: 10450336-10450336 on Build GRCh38
rs767743207 Chr.1: 10450198-10450198 on Build GRCh38
rs760933460 Chr.1: 10450142-10450142 on Build GRCh38
rs780645425 Chr.1: 10450039-10450039 on Build GRCh38
rs370903835 Chr.1: 10450319-10450319 on Build GRCh38
rs996242087 Chr.1: 10450393-10450396 on Build GRCh38
rs764523716 Chr.1: 10450088-10450088 on Build GRCh38
rs767225736 Chr.1: 10450115-10450115 on Build GRCh38
rs765753601 Chr.1: 10450428-10450428 on Build GRCh38
rs1053681176 Chr.1: 10450209-10450209 on Build GRCh38
rs1004825849 Chr.1: 10450424-10450424 on Build GRCh38
rs780805484 Chr.1: 10450364-10450365 on Build GRCh38
rs752733385 Chr.1: 10450306-10450306 on Build GRCh38
rs751345586 Chr.1: 10450311-10450311 on Build GRCh38
rs778847098 Chr.1: 10450330-10450330 on Build GRCh38
rs758276462 Chr.1: 10450243-10450243 on Build GRCh38
rs774863235 Chr.1: 10450367-10450367 on Build GRCh38
rs778952757 Chr.1: 10450138-10450138 on Build GRCh38
rs374605608 Chr.1: 10450263-10450263 on Build GRCh38
rs749984934 Chr.1: 10450041-10450041 on Build GRCh38
rs764022669 Chr.1: 10450308-10450308 on Build GRCh38
rs747977320 Chr.1: 10450145-10450145 on Build GRCh38
rs763344643 Chr.1: 10450027-10450027 on Build GRCh38
rs759156889 Chr.1: 10450289-10450289 on Build GRCh38
rs201903837 Chr.1: 10450060-10450060 on Build GRCh38
rs1042560750 Chr.1: 10450170-10450170 on Build GRCh38
rs762046604 Chr.1: 10450111-10450111 on Build GRCh38
rs766073481 Chr.1: 10450228-10450228 on Build GRCh38
rs900252730 Chr.1: 10450392-10450393 on Build GRCh38
rs35962039 Chr.1: 10450442-10450443 on Build GRCh38
rs775638493 Chr.1: 10450117-10450117 on Build GRCh38
rs776492345 Chr.1: 10450356-10450356 on Build GRCh38
rs995431063 Chr.1: 10450298-10450298 on Build GRCh38
rs189899917 Chr.1: 10450017-10450017 on Build GRCh38
rs769970397 Chr.1: 10450165-10450165 on Build GRCh38
rs754487841 Chr.1: 10450236-10450236 on Build GRCh38
rs375806455 Chr.1: 10450238-10450238 on Build GRCh38
rs780307279 Chr.1: 10450314-10450314 on Build GRCh38
rs762291116 Chr.1: 10450194-10450194 on Build GRCh38
rs774882640 Chr.1: 10450270-10450270 on Build GRCh38
rs536349877 Chr.1: 10450073-10450073 on Build GRCh38
rs539189765 Chr.1: 10450442-10450442 on Build GRCh38
rs747883035 Chr.1: 10450067-10450067 on Build GRCh38
rs369706158 Chr.1: 10450031-10450031 on Build GRCh38
rs142755741 Chr.1: 10450239-10450239 on Build GRCh38
rs755117596 Chr.1: 10450323-10450323 on Build GRCh38
rs766149209 Chr.1: 10450281-10450281 on Build GRCh38
rs368651066 Chr.1: 10450287-10450287 on Build GRCh38
rs771389549 Chr.1: 10450349-10450349 on Build GRCh38
rs377214659 Chr.1: 10450123-10450123 on Build GRCh38
rs937977668 Chr.1: 10450318-10450318 on Build GRCh38
rs768641510 Chr.1: 10450057-10450057 on Build GRCh38
rs556873478 Chr.1: 10450291-10450291 on Build GRCh38
rs756999760 Chr.1: 10450312-10450312 on Build GRCh38
rs372771717 Chr.1: 10450038-10450038 on Build GRCh38
rs759710175 Chr.1: 10450076-10450076 on Build GRCh38
rs749206441 Chr.1: 10450050-10450050 on Build GRCh38
rs565298894 Chr.1: 10450244-10450244 on Build GRCh38
rs375245773 Chr.1: 10450339-10450339 on Build GRCh38
rs746069598 Chr.1: 10450164-10450164 on Build GRCh38
rs753428581 Chr.1: 10450234-10450234 on Build GRCh38
rs769185888 Chr.1: 10450184-10450184 on Build GRCh38
rs769510142 Chr.1: 10450363-10450363 on Build GRCh38
rs751929665 Chr.1: 10450109-10450109 on Build GRCh38
rs182159138 Chr.1: 10450277-10450277 on Build GRCh38
rs755533538 Chr.1: 10450046-10450046 on Build GRCh38
rs757577961 Chr.1: 10450409-10450409 on Build GRCh38
rs772985649 Chr.1: 10450204-10450204 on Build GRCh38
rs574274425 Chr.1: 10450271-10450271 on Build GRCh38
rs367976064 Chr.1: 10450340-10450340 on Build GRCh38
rs769727511 Chr.1: 10450078-10450078 on Build GRCh38
rs752590489 Chr.1: 10450127-10450128 on Build GRCh38
rs949672945 Chr.1: 10450343-10450343 on Build GRCh38
rs748202780 Chr.1: 10450332-10450332 on Build GRCh38
rs200789768 Chr.1: 10450030-10450030 on Build GRCh38
rs368245686 Chr.1: 10450264-10450264 on Build GRCh38
rs753886185 Chr.1: 10450131-10450131 on Build GRCh38
rs772591880 Chr.1: 10450395-10450395 on Build GRCh38
rs758193275 Chr.1: 10450129-10450131 on Build GRCh38
rs745394293 Chr.1: 10450370-10450371 on Build GRCh38
rs758278055 Chr.1: 10450307-10450307 on Build GRCh38
rs1014866459 Chr.1: 10450426-10450426 on Build GRCh38
rs370073315 Chr.1: 10450193-10450193 on Build GRCh38
rs759246091 Chr.1: 10450361-10450361 on Build GRCh38
rs760232306 Chr.1: 10450217-10450217 on Build GRCh38
rs756292279 Chr.1: 10450260-10450260 on Build GRCh38
rs779426632 Chr.1: 10450129-10450129 on Build GRCh38
rs780089892 Chr.1: 10450049-10450049 on Build GRCh38
rs771214827 Chr.1: 10450069-10450069 on Build GRCh38
rs780211807 Chr.1: 10450262-10450262 on Build GRCh38
rs763863000 Chr.1: 10450182-10450184 on Build GRCh38
rs754981461 Chr.1: 10450136-10450136 on Build GRCh38
rs185532953 Chr.1: 10450006-10450006 on Build GRCh38
rs753179420 Chr.1: 10450025-10450025 on Build GRCh38
rs898511847 Chr.1: 10450074-10450074 on Build GRCh38
rs776079055 Chr.1: 10450359-10450360 on Build GRCh38
rs764412437 Chr.1: 10450159-10450159 on Build GRCh38
rs751279638 Chr.1: 10450244-10450245 on Build GRCh38
rs961308755 Chr.1: 10450047-10450047 on Build GRCh38

View Full Product Details