Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89577427-89577934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4623458 c.335G>A p.G112D Chr.16: 89577699-89577699 on Build GRCh38
5529140 c.328C>T p.L110L Chr.16: 89577692-89577692 on Build GRCh38
5519420 c.264G>A p.R88R Chr.16: 89577628-89577628 on Build GRCh38
4063620 c.270C>T p.R90R Chr.16: 89577634-89577634 on Build GRCh38
7410753 c.291T>A p.H97Q Chr.16: 89577655-89577655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961869672 Chr.16: 89577751-89577751 on Build GRCh38
rs140209477 Chr.16: 89577674-89577674 on Build GRCh38
rs377027196 Chr.16: 89577741-89577741 on Build GRCh38
rs761301695 Chr.16: 89577614-89577614 on Build GRCh38
rs780450140 Chr.16: 89577742-89577742 on Build GRCh38
rs776597450 Chr.16: 89577844-89577844 on Build GRCh38
rs150570411 Chr.16: 89577875-89577875 on Build GRCh38
rs200538988 Chr.16: 89577510-89577510 on Build GRCh38
rs578025330 Chr.16: 89577755-89577755 on Build GRCh38
rs780555581 Chr.16: 89577491-89577491 on Build GRCh38
rs766823094 Chr.16: 89577492-89577492 on Build GRCh38
rs545130174 Chr.16: 89577762-89577762 on Build GRCh38
rs967949466 Chr.16: 89577791-89577791 on Build GRCh38
rs777870492 Chr.16: 89577703-89577703 on Build GRCh38
rs536529003 Chr.16: 89577656-89577656 on Build GRCh38
rs553386533 Chr.16: 89577712-89577712 on Build GRCh38
rs771912937 Chr.16: 89577599-89577600 on Build GRCh38
rs752650209 Chr.16: 89577698-89577698 on Build GRCh38
rs765991871 Chr.16: 89577682-89577682 on Build GRCh38
rs752534377 Chr.16: 89577564-89577564 on Build GRCh38
rs746145877 Chr.16: 89577733-89577733 on Build GRCh38
rs963712394 Chr.16: 89577852-89577852 on Build GRCh38
rs151180960 Chr.16: 89577665-89577665 on Build GRCh38
rs1048773183 Chr.16: 89577908-89577908 on Build GRCh38
rs779183997 Chr.16: 89577505-89577505 on Build GRCh38
rs755955923 Chr.16: 89577585-89577585 on Build GRCh38
rs763486441 Chr.16: 89577529-89577529 on Build GRCh38
rs749587094 Chr.16: 89577597-89577597 on Build GRCh38
rs762466550 Chr.16: 89577756-89577756 on Build GRCh38
rs976382369 Chr.16: 89577866-89577866 on Build GRCh38
rs866038042 Chr.16: 89577485-89577485 on Build GRCh38
rs778036738 Chr.16: 89577653-89577653 on Build GRCh38
rs561432152 Chr.16: 89577801-89577801 on Build GRCh38
rs868758891 Chr.16: 89577470-89577470 on Build GRCh38
rs751184121 Chr.16: 89577685-89577685 on Build GRCh38
rs889570902 Chr.16: 89577521-89577521 on Build GRCh38
rs985420874 Chr.16: 89577647-89577647 on Build GRCh38
rs1011952746 Chr.16: 89577729-89577729 on Build GRCh38
rs764799215 Chr.16: 89577620-89577620 on Build GRCh38
rs760643293 Chr.16: 89577745-89577747 on Build GRCh38
rs750049144 Chr.16: 89577622-89577622 on Build GRCh38
rs779185992 Chr.16: 89577732-89577732 on Build GRCh38
rs961499702 Chr.16: 89577608-89577608 on Build GRCh38
rs373905421 Chr.16: 89577680-89577680 on Build GRCh38
rs528637744 Chr.16: 89577826-89577826 on Build GRCh38
rs753911675 Chr.16: 89577709-89577709 on Build GRCh38
rs746189263 Chr.16: 89577659-89577659 on Build GRCh38
rs954127253 Chr.16: 89577765-89577765 on Build GRCh38
rs455527 Chr.16: 89577593-89577593 on Build GRCh38
rs983753231 Chr.16: 89577631-89577631 on Build GRCh38
rs201422940 Chr.16: 89577499-89577499 on Build GRCh38
rs542878705 Chr.16: 89577796-89577796 on Build GRCh38
rs532282879 Chr.16: 89577632-89577632 on Build GRCh38
rs754129211 Chr.16: 89577613-89577613 on Build GRCh38
rs779031450 Chr.16: 89577658-89577658 on Build GRCh38
rs11644942 Chr.16: 89577672-89577672 on Build GRCh38
rs916372047 Chr.16: 89577857-89577857 on Build GRCh38
rs143407646 Chr.16: 89577771-89577771 on Build GRCh38
rs34344861 Chr.16: 89577550-89577550 on Build GRCh38
rs952666423 Chr.16: 89577841-89577841 on Build GRCh38
rs541488981 Chr.16: 89577462-89577462 on Build GRCh38
rs745754885 Chr.16: 89577849-89577851 on Build GRCh38
rs35701751 Chr.16: 89577589-89577589 on Build GRCh38
rs983972527 Chr.16: 89577843-89577843 on Build GRCh38
rs200668535 Chr.16: 89577651-89577651 on Build GRCh38
rs375866241 Chr.16: 89577771-89577772 on Build GRCh38
rs756166378 Chr.16: 89577699-89577699 on Build GRCh38
rs759360639 Chr.16: 89577686-89577686 on Build GRCh38
rs866303878 Chr.16: 89577551-89577551 on Build GRCh38
rs1010948341 Chr.16: 89577569-89577569 on Build GRCh38
rs559921302 Chr.16: 89577652-89577652 on Build GRCh38
rs142649340 Chr.16: 89577463-89577463 on Build GRCh38
rs2271914 Chr.16: 89577770-89577770 on Build GRCh38
rs577930324 Chr.16: 89577489-89577489 on Build GRCh38
rs561118662 Chr.16: 89577579-89577579 on Build GRCh38
rs769112203 Chr.16: 89577754-89577754 on Build GRCh38
rs781348698 Chr.16: 89577646-89577646 on Build GRCh38
rs1049657143 Chr.16: 89577520-89577520 on Build GRCh38
rs575255442 Chr.16: 89577782-89577782 on Build GRCh38
rs747397399 Chr.16: 89577745-89577745 on Build GRCh38
rs962430013 Chr.16: 89577776-89577776 on Build GRCh38
rs960327189 Chr.16: 89577449-89577449 on Build GRCh38
rs547662887 Chr.16: 89577654-89577654 on Build GRCh38
rs753293523 Chr.16: 89577536-89577536 on Build GRCh38
rs773840851 Chr.16: 89577453-89577453 on Build GRCh38
rs571255703 Chr.16: 89577602-89577602 on Build GRCh38
rs373791273 Chr.16: 89577572-89577572 on Build GRCh38
rs923863065 Chr.16: 89577818-89577818 on Build GRCh38
rs746899856 Chr.16: 89577577-89577577 on Build GRCh38
rs939209775 Chr.16: 89577840-89577840 on Build GRCh38
rs932685020 Chr.16: 89577643-89577643 on Build GRCh38
rs972964752 Chr.16: 89577611-89577611 on Build GRCh38
rs922418091 Chr.16: 89577895-89577895 on Build GRCh38
rs932603560 Chr.16: 89577518-89577518 on Build GRCh38
rs775398112 Chr.16: 89577693-89577694 on Build GRCh38
rs1056292264 Chr.16: 89577846-89577846 on Build GRCh38
rs374056543 Chr.16: 89577777-89577777 on Build GRCh38
rs369107618 Chr.16: 89577561-89577561 on Build GRCh38
rs748171497 Chr.16: 89577583-89577583 on Build GRCh38
rs767486690 Chr.16: 89577695-89577695 on Build GRCh38
rs978388134 Chr.16: 89577802-89577802 on Build GRCh38
rs770403197 Chr.16: 89577761-89577761 on Build GRCh38
rs113410325 Chr.16: 89577664-89577664 on Build GRCh38
rs888710217 Chr.16: 89577740-89577740 on Build GRCh38
rs946587922 Chr.16: 89577539-89577539 on Build GRCh38
rs774825737 Chr.16: 89577598-89577598 on Build GRCh38
rs757547950 Chr.16: 89577872-89577872 on Build GRCh38
rs369719899 Chr.16: 89577578-89577578 on Build GRCh38
rs199749974 Chr.16: 89577557-89577557 on Build GRCh38
rs1025858402 Chr.16: 89577781-89577781 on Build GRCh38
rs947752450 Chr.16: 89577862-89577862 on Build GRCh38
rs572985580 Chr.16: 89577452-89577452 on Build GRCh38
rs772660295 Chr.16: 89577675-89577675 on Build GRCh38
rs758119244 Chr.16: 89577628-89577628 on Build GRCh38
rs764815949 Chr.16: 89577560-89577560 on Build GRCh38
rs758126808 Chr.16: 89577565-89577565 on Build GRCh38

View Full Product Details