Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89578457-89578990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063621 c.385G>A p.V129M Chr.16: 89578884-89578884 on Build GRCh38
4841032 c.384C>T p.V128V Chr.16: 89578883-89578883 on Build GRCh38
5626698 c.398C>G p.S133C Chr.16: 89578897-89578897 on Build GRCh38
7211673 c.421G>A p.G141R Chr.16: 89578920-89578920 on Build GRCh38
4674397 c.445T>C p.C149R Chr.16: 89578944-89578944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373648106 Chr.16: 89578951-89578951 on Build GRCh38
rs374128987 Chr.16: 89578906-89578908 on Build GRCh38
rs35811163 Chr.16: 89578777-89578778 on Build GRCh38
rs1001851079 Chr.16: 89578674-89578674 on Build GRCh38
rs778904372 Chr.16: 89578818-89578818 on Build GRCh38
rs375992589 Chr.16: 89578777-89578779 on Build GRCh38
rs78176582 Chr.16: 89578913-89578913 on Build GRCh38
rs765246853 Chr.16: 89578914-89578914 on Build GRCh38
rs78990556 Chr.16: 89578960-89578960 on Build GRCh38
rs761700586 Chr.16: 89578957-89578957 on Build GRCh38
rs151106955 Chr.16: 89578699-89578699 on Build GRCh38
rs762677044 Chr.16: 89578485-89578485 on Build GRCh38
rs536386166 Chr.16: 89578539-89578539 on Build GRCh38
rs945303270 Chr.16: 89578853-89578853 on Build GRCh38
rs559221439 Chr.16: 89578647-89578647 on Build GRCh38
rs146919277 Chr.16: 89578859-89578859 on Build GRCh38
rs1050944538 Chr.16: 89578621-89578621 on Build GRCh38
rs149812493 Chr.16: 89578952-89578952 on Build GRCh38
rs760485243 Chr.16: 89578899-89578899 on Build GRCh38
rs1009451698 Chr.16: 89578631-89578631 on Build GRCh38
rs960435321 Chr.16: 89578690-89578690 on Build GRCh38
rs745389352 Chr.16: 89578884-89578884 on Build GRCh38
rs960885604 Chr.16: 89578562-89578562 on Build GRCh38
rs933852959 Chr.16: 89578617-89578617 on Build GRCh38
rs373180490 Chr.16: 89578827-89578827 on Build GRCh38
rs910183335 Chr.16: 89578702-89578703 on Build GRCh38
rs935548261 Chr.16: 89578795-89578795 on Build GRCh38
rs62068670 Chr.16: 89578525-89578525 on Build GRCh38
rs925451451 Chr.16: 89578536-89578536 on Build GRCh38
rs879094818 Chr.16: 89578760-89578762 on Build GRCh38
rs869249157 Chr.16: 89578774-89578778 on Build GRCh38
rs974708225 Chr.16: 89578801-89578801 on Build GRCh38
rs753308974 Chr.16: 89578831-89578832 on Build GRCh38
rs950959844 Chr.16: 89578576-89578576 on Build GRCh38
rs909189621 Chr.16: 89578586-89578586 on Build GRCh38
rs1043686901 Chr.16: 89578855-89578855 on Build GRCh38
rs933380879 Chr.16: 89578839-89578839 on Build GRCh38
rs1025703586 Chr.16: 89578785-89578785 on Build GRCh38
rs954611255 Chr.16: 89578686-89578686 on Build GRCh38
rs145683595 Chr.16: 89578675-89578675 on Build GRCh38
rs112572691 Chr.16: 89578589-89578589 on Build GRCh38
rs748667922 Chr.16: 89578862-89578862 on Build GRCh38
rs925514672 Chr.16: 89578786-89578786 on Build GRCh38
rs762404341 Chr.16: 89578761-89578763 on Build GRCh38
rs1045050976 Chr.16: 89578614-89578614 on Build GRCh38
rs374165970 Chr.16: 89578868-89578868 on Build GRCh38
rs923345647 Chr.16: 89578591-89578591 on Build GRCh38
rs919543975 Chr.16: 89578572-89578572 on Build GRCh38
rs148102686 Chr.16: 89578966-89578966 on Build GRCh38
rs146913973 Chr.16: 89578928-89578928 on Build GRCh38
rs368397763 Chr.16: 89578692-89578692 on Build GRCh38
rs554651248 Chr.16: 89578542-89578542 on Build GRCh38
rs369250260 Chr.16: 89578807-89578807 on Build GRCh38
rs763008448 Chr.16: 89578967-89578967 on Build GRCh38
rs987045624 Chr.16: 89578846-89578846 on Build GRCh38
rs183824116 Chr.16: 89578626-89578626 on Build GRCh38
rs367779313 Chr.16: 89578530-89578530 on Build GRCh38
rs796116400 Chr.16: 89578779-89578779 on Build GRCh38
rs369025222 Chr.16: 89578852-89578852 on Build GRCh38
rs985206148 Chr.16: 89578579-89578579 on Build GRCh38
rs992324487 Chr.16: 89578570-89578570 on Build GRCh38
rs766533546 Chr.16: 89578918-89578918 on Build GRCh38
rs985038512 Chr.16: 89578776-89578776 on Build GRCh38
rs755329950 Chr.16: 89578921-89578921 on Build GRCh38
rs773782633 Chr.16: 89578877-89578877 on Build GRCh38
rs575637298 Chr.16: 89578661-89578661 on Build GRCh38
rs77799181 Chr.16: 89578898-89578898 on Build GRCh38
rs374006079 Chr.16: 89578917-89578917 on Build GRCh38
rs1037651104 Chr.16: 89578925-89578925 on Build GRCh38
rs541935172 Chr.16: 89578790-89578790 on Build GRCh38
rs781714858 Chr.16: 89578860-89578860 on Build GRCh38
rs577475085 Chr.16: 89578657-89578657 on Build GRCh38
rs377581262 Chr.16: 89578845-89578845 on Build GRCh38
rs377398671 Chr.16: 89578902-89578902 on Build GRCh38
rs746701832 Chr.16: 89578950-89578950 on Build GRCh38
rs1043880379 Chr.16: 89578528-89578528 on Build GRCh38
rs1043424806 Chr.16: 89578642-89578642 on Build GRCh38
rs780286654 Chr.16: 89578835-89578835 on Build GRCh38
rs751839737 Chr.16: 89578920-89578920 on Build GRCh38
rs985751551 Chr.16: 89578507-89578507 on Build GRCh38
rs763143503 Chr.16: 89578760-89578763 on Build GRCh38
rs943475626 Chr.16: 89578587-89578587 on Build GRCh38
rs747309371 Chr.16: 89578837-89578837 on Build GRCh38
rs757356211 Chr.16: 89578814-89578814 on Build GRCh38
rs561379517 Chr.16: 89578807-89578808 on Build GRCh38
rs954428848 Chr.16: 89578491-89578491 on Build GRCh38
rs368156323 Chr.16: 89578778-89578779 on Build GRCh38
rs750640063 Chr.16: 89578823-89578823 on Build GRCh38
rs534022060 Chr.16: 89578759-89578760 on Build GRCh38
rs1030114246 Chr.16: 89578677-89578677 on Build GRCh38
rs964274930 Chr.16: 89578727-89578727 on Build GRCh38
rs961925692 Chr.16: 89578650-89578650 on Build GRCh38
rs1026451694 Chr.16: 89578735-89578735 on Build GRCh38
rs753780727 Chr.16: 89578808-89578808 on Build GRCh38
rs778371454 Chr.16: 89578935-89578935 on Build GRCh38
rs765136360 Chr.16: 89578762-89578763 on Build GRCh38
rs527294284 Chr.16: 89578883-89578883 on Build GRCh38
rs141048497 Chr.16: 89578929-89578929 on Build GRCh38
rs1021922352 Chr.16: 89578482-89578482 on Build GRCh38
rs775280836 Chr.16: 89578949-89578949 on Build GRCh38
rs903757169 Chr.16: 89578870-89578870 on Build GRCh38
rs9944362 Chr.16: 89578937-89578937 on Build GRCh38
rs776602589 Chr.16: 89578907-89578907 on Build GRCh38
rs1052561041 Chr.16: 89578585-89578585 on Build GRCh38
rs771806328 Chr.16: 89578889-89578889 on Build GRCh38
rs550614069 Chr.16: 89578519-89578519 on Build GRCh38
rs951048630 Chr.16: 89578737-89578737 on Build GRCh38
rs903495340 Chr.16: 89578655-89578655 on Build GRCh38
rs761465540 Chr.16: 89578873-89578873 on Build GRCh38
rs549402469 Chr.16: 89578760-89578761 on Build GRCh38
rs979678792 Chr.16: 89578767-89578767 on Build GRCh38
rs370192350 Chr.16: 89578940-89578940 on Build GRCh38

View Full Product Details