Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18606344-18606838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLF1
Entrez Gene ID
9244
Gene Name
cytokine receptor like factor 1
Gene Aliases
CISS, CISS1, CLF, CLF-1, NR6, zcytor5
UniGene
Hs.114948
Gene Chromosome Location
Chr.19: 18593225-18606850 on Build GRCh38
Transcripts
NM_004750.4 XM_011528423.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5446001 c.64_66delCTG p.L26delL Chr.19: 18606591-18606593 on Build GRCh38
5030631 c.65_67delTGC p.L26delL Chr.19: 18606590-18606592 on Build GRCh38
1611838 c.115+3G>T p.? Chr.19: 18606539-18606539 on Build GRCh38
1611839 c.75_77delGCT p.L26delL Chr.19: 18606580-18606582 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1001570616 Chr.19: 18606591-18606591 on Build GRCh38
rs946321821 Chr.19: 18606754-18606754 on Build GRCh38
rs936129468 Chr.19: 18606735-18606735 on Build GRCh38
rs962001828 Chr.19: 18606669-18606669 on Build GRCh38
rs113093604 Chr.19: 18606454-18606454 on Build GRCh38
rs931760531 Chr.19: 18606472-18606472 on Build GRCh38
rs1054639315 Chr.19: 18606751-18606751 on Build GRCh38
rs34503316 Chr.19: 18606580-18606582 on Build GRCh38
rs1001518292 Chr.19: 18606804-18606804 on Build GRCh38
rs985871392 Chr.19: 18606718-18606718 on Build GRCh38
rs984664488 Chr.19: 18606455-18606456 on Build GRCh38
rs565524847 Chr.19: 18606510-18606510 on Build GRCh38
rs951869440 Chr.19: 18606632-18606632 on Build GRCh38
rs887881897 Chr.19: 18606822-18606822 on Build GRCh38
rs907192369 Chr.19: 18606770-18606770 on Build GRCh38
rs1024785410 Chr.19: 18606395-18606395 on Build GRCh38
rs922968172 Chr.19: 18606703-18606703 on Build GRCh38
rs975792257 Chr.19: 18606469-18606469 on Build GRCh38
rs925045973 Chr.19: 18606732-18606732 on Build GRCh38
rs907826821 Chr.19: 18606461-18606461 on Build GRCh38
rs1010622335 Chr.19: 18606528-18606528 on Build GRCh38
rs940332918 Chr.19: 18606462-18606462 on Build GRCh38
rs137853925 Chr.19: 18606582-18606584 on Build GRCh38
rs562764342 Chr.19: 18606404-18606404 on Build GRCh38
rs922985163 Chr.19: 18606471-18606471 on Build GRCh38
rs927935351 Chr.19: 18606711-18606712 on Build GRCh38
rs892006609 Chr.19: 18606753-18606753 on Build GRCh38
rs1034379834 Chr.19: 18606807-18606807 on Build GRCh38
rs1046441847 Chr.19: 18606760-18606760 on Build GRCh38
rs936181821 Chr.19: 18606516-18606516 on Build GRCh38
rs1054693816 Chr.19: 18606526-18606526 on Build GRCh38
rs1034430435 Chr.19: 18606612-18606612 on Build GRCh38
rs1010259934 Chr.19: 18606373-18606373 on Build GRCh38
rs1024317469 Chr.19: 18606569-18606569 on Build GRCh38
rs1015232472 Chr.19: 18606665-18606665 on Build GRCh38
rs137853929 Chr.19: 18606604-18606626 on Build GRCh38
rs976122959 Chr.19: 18606679-18606679 on Build GRCh38
rs979870655 Chr.19: 18606442-18606442 on Build GRCh38
rs971516718 Chr.19: 18606400-18606400 on Build GRCh38
rs188747632 Chr.19: 18606485-18606485 on Build GRCh38
rs1034149002 Chr.19: 18606451-18606451 on Build GRCh38
rs984614003 Chr.19: 18606648-18606648 on Build GRCh38

View Full Product Details