Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33144874-33145376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAP
Entrez Gene ID
10491
Gene Name
cartilage associated protein
Gene Aliases
CASP, LEPREL3, OI7, P3H5
UniGene
Hs.517888
Gene Chromosome Location
Chr.3: 33113958-33147773 on Build GRCh38
Transcripts
NM_006371.4

SNP Detail

SNP ID Chromosome Location
rs147135876 Chr.3: 33145324-33145324 on Build GRCh38
rs746736695 Chr.3: 33144981-33144981 on Build GRCh38
rs541113914 Chr.3: 33145322-33145322 on Build GRCh38
rs1024796563 Chr.3: 33145013-33145013 on Build GRCh38
rs115744599 Chr.3: 33145076-33145076 on Build GRCh38
rs547363446 Chr.3: 33145127-33145127 on Build GRCh38
rs566327751 Chr.3: 33144904-33144904 on Build GRCh38
rs565676142 Chr.3: 33145135-33145135 on Build GRCh38
rs936276962 Chr.3: 33145278-33145278 on Build GRCh38
rs72857502 Chr.3: 33145349-33145349 on Build GRCh38
rs1039187197 Chr.3: 33145251-33145251 on Build GRCh38
rs769953681 Chr.3: 33145134-33145134 on Build GRCh38
rs878916788 Chr.3: 33145030-33145030 on Build GRCh38
rs149468116 Chr.3: 33145141-33145141 on Build GRCh38
rs935288684 Chr.3: 33145111-33145111 on Build GRCh38
rs78745278 Chr.3: 33144922-33144922 on Build GRCh38
rs963292371 Chr.3: 33145100-33145100 on Build GRCh38
rs374956331 Chr.3: 33145125-33145125 on Build GRCh38
rs537364267 Chr.3: 33145248-33145248 on Build GRCh38
rs777641110 Chr.3: 33144971-33144971 on Build GRCh38
rs774780902 Chr.3: 33145340-33145340 on Build GRCh38
rs373882880 Chr.3: 33144954-33144954 on Build GRCh38
rs758223063 Chr.3: 33144960-33144960 on Build GRCh38
rs528845478 Chr.3: 33145122-33145122 on Build GRCh38
rs138063065 Chr.3: 33145055-33145055 on Build GRCh38
rs757110442 Chr.3: 33145031-33145031 on Build GRCh38
rs970588760 Chr.3: 33145035-33145035 on Build GRCh38
rs943603181 Chr.3: 33144912-33144912 on Build GRCh38
rs932174031 Chr.3: 33145001-33145001 on Build GRCh38
rs1051816709 Chr.3: 33145073-33145073 on Build GRCh38
rs573980279 Chr.3: 33145319-33145319 on Build GRCh38
rs764661352 Chr.3: 33145285-33145285 on Build GRCh38
rs569885677 Chr.3: 33145169-33145169 on Build GRCh38
rs1051908410 Chr.3: 33144903-33144903 on Build GRCh38
rs539500198 Chr.3: 33145136-33145136 on Build GRCh38
rs555598507 Chr.3: 33145301-33145301 on Build GRCh38
rs904746576 Chr.3: 33145269-33145269 on Build GRCh38
rs951148580 Chr.3: 33145217-33145217 on Build GRCh38
rs1001998330 Chr.3: 33145329-33145329 on Build GRCh38
rs953501139 Chr.3: 33145325-33145325 on Build GRCh38
rs980676425 Chr.3: 33145067-33145067 on Build GRCh38
rs185729872 Chr.3: 33144992-33144992 on Build GRCh38
rs150800828 Chr.3: 33144972-33144972 on Build GRCh38
rs749823997 Chr.3: 33145231-33145231 on Build GRCh38
rs1052432139 Chr.3: 33145114-33145115 on Build GRCh38
rs557812750 Chr.3: 33145232-33145232 on Build GRCh38
rs886058365 Chr.3: 33145033-33145033 on Build GRCh38
rs922123498 Chr.3: 33144988-33144989 on Build GRCh38
rs1053098667 Chr.3: 33145283-33145283 on Build GRCh38

View Full Product Details