Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142910561-142911101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11B2
Entrez Gene ID
1585
Gene Name
cytochrome P450 family 11 subfamily B member 2
Gene Aliases
ALDOS, CPN2, CYP11B, CYP11BL, CYPXIB2, P-450C18, P450C18, P450aldo
UniGene
Hs.632054
Gene Chromosome Location
Chr.8: 142910559-142917843 on Build GRCh38
Transcripts
NM_000498.3

SNP Detail

SNP ID Chromosome Location
rs1005277521 Chr.8: 142910633-142910633 on Build GRCh38
rs953793443 Chr.8: 142910608-142910608 on Build GRCh38
rs981168733 Chr.8: 142910953-142910953 on Build GRCh38
rs930051563 Chr.8: 142911068-142911068 on Build GRCh38
rs543906752 Chr.8: 142910776-142910776 on Build GRCh38
rs559842795 Chr.8: 142911028-142911028 on Build GRCh38
rs961295031 Chr.8: 142910655-142910655 on Build GRCh38
rs140738939 Chr.8: 142910819-142910819 on Build GRCh38
rs865909117 Chr.8: 142910858-142910858 on Build GRCh38
rs993722389 Chr.8: 142910831-142910831 on Build GRCh38
rs35574522 Chr.8: 142910784-142910785 on Build GRCh38
rs576148290 Chr.8: 142910732-142910734 on Build GRCh38
rs868030508 Chr.8: 142911074-142911074 on Build GRCh38
rs972424267 Chr.8: 142910656-142910656 on Build GRCh38
rs576625541 Chr.8: 142910772-142910772 on Build GRCh38
rs966233223 Chr.8: 142910615-142910615 on Build GRCh38
rs984401179 Chr.8: 142910673-142910673 on Build GRCh38
rs1026168746 Chr.8: 142910854-142910854 on Build GRCh38
rs569831537 Chr.8: 142910849-142910849 on Build GRCh38
rs1029147528 Chr.8: 142910604-142910604 on Build GRCh38
rs761068577 Chr.8: 142910954-142910954 on Build GRCh38
rs927108183 Chr.8: 142910743-142910743 on Build GRCh38
rs570285601 Chr.8: 142910592-142910592 on Build GRCh38
rs988986096 Chr.8: 142910995-142910995 on Build GRCh38
rs530398511 Chr.8: 142911059-142911059 on Build GRCh38
rs574670091 Chr.8: 142910710-142910710 on Build GRCh38
rs969655359 Chr.8: 142910937-142910937 on Build GRCh38
rs886062739 Chr.8: 142910933-142910933 on Build GRCh38
rs3802228 Chr.8: 142910802-142910802 on Build GRCh38
rs61763988 Chr.8: 142910640-142910640 on Build GRCh38
rs911518164 Chr.8: 142911038-142911038 on Build GRCh38
rs952024016 Chr.8: 142910668-142910668 on Build GRCh38
rs944013431 Chr.8: 142911042-142911042 on Build GRCh38
rs985673192 Chr.8: 142911037-142911037 on Build GRCh38
rs61763989 Chr.8: 142910987-142910987 on Build GRCh38
rs1026640378 Chr.8: 142910660-142910660 on Build GRCh38
rs745532151 Chr.8: 142910703-142910703 on Build GRCh38
rs542681319 Chr.8: 142910734-142910734 on Build GRCh38
rs887628134 Chr.8: 142910859-142910859 on Build GRCh38
rs886062740 Chr.8: 142911008-142911008 on Build GRCh38
rs914717258 Chr.8: 142911002-142911002 on Build GRCh38
rs554933515 Chr.8: 142910746-142910746 on Build GRCh38
rs541605165 Chr.8: 142910931-142910931 on Build GRCh38
rs1006065351 Chr.8: 142910921-142910921 on Build GRCh38
rs797014379 Chr.8: 142910694-142910694 on Build GRCh38
rs188784518 Chr.8: 142910688-142910688 on Build GRCh38
rs915551240 Chr.8: 142910678-142910678 on Build GRCh38
rs897273036 Chr.8: 142910830-142910830 on Build GRCh38
rs889759894 Chr.8: 142910812-142910812 on Build GRCh38
rs771771915 Chr.8: 142910720-142910720 on Build GRCh38
rs1041525626 Chr.8: 142910823-142910824 on Build GRCh38
rs1041079238 Chr.8: 142911056-142911056 on Build GRCh38
rs535660076 Chr.8: 142910920-142910920 on Build GRCh38

View Full Product Details