Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203279614-203280115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP20A1
Entrez Gene ID
57404
Gene Name
cytochrome P450 family 20 subfamily A member 1
Gene Aliases
CYP-M
UniGene
Hs.446065
Gene Chromosome Location
Chr.2: 203238441-203305840 on Build GRCh38
Transcripts
NM_177538.2 XM_005246698.3 XM_005246699.3 XM_005246701.3 XM_006712644.3 XM_006712645.3 XM_017004555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015118 c.821C>A p.S274Y Chr.2: 203280084-203280084 on Build GRCh38
4559372 c.802G>A p.E268K Chr.2: 203280065-203280065 on Build GRCh38
6543830 c.813G>T p.M271I Chr.2: 203280076-203280076 on Build GRCh38
3798535 c.845C>A p.A282E Chr.2: 203280108-203280108 on Build GRCh38
6204941 c.827C>A p.A276D Chr.2: 203280090-203280090 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1007613144 Chr.2: 203279980-203279980 on Build GRCh38
rs548598480 Chr.2: 203280022-203280022 on Build GRCh38
rs1017344420 Chr.2: 203279934-203279934 on Build GRCh38
rs934207341 Chr.2: 203279711-203279711 on Build GRCh38
rs905219438 Chr.2: 203279780-203279780 on Build GRCh38
rs1001038104 Chr.2: 203279923-203279923 on Build GRCh38
rs1056336396 Chr.2: 203279837-203279837 on Build GRCh38
rs766331078 Chr.2: 203280069-203280069 on Build GRCh38
rs1019069133 Chr.2: 203279645-203279645 on Build GRCh38
rs1009441889 Chr.2: 203279885-203279885 on Build GRCh38
rs769987827 Chr.2: 203280016-203280016 on Build GRCh38
rs767166150 Chr.2: 203280035-203280035 on Build GRCh38
rs774068295 Chr.2: 203280026-203280026 on Build GRCh38
rs759494065 Chr.2: 203280061-203280061 on Build GRCh38
rs533398515 Chr.2: 203279809-203279809 on Build GRCh38
rs542854636 Chr.2: 203279677-203279677 on Build GRCh38
rs552868627 Chr.2: 203279800-203279800 on Build GRCh38
rs538393487 Chr.2: 203279947-203279947 on Build GRCh38
rs889041578 Chr.2: 203279952-203279953 on Build GRCh38
rs985036246 Chr.2: 203279731-203279731 on Build GRCh38
rs761437767 Chr.2: 203280032-203280032 on Build GRCh38
rs552078871 Chr.2: 203279870-203279870 on Build GRCh38
rs527706604 Chr.2: 203279796-203279796 on Build GRCh38
rs1026470832 Chr.2: 203279714-203279714 on Build GRCh38
rs903652739 Chr.2: 203279911-203279911 on Build GRCh38
rs571170110 Chr.2: 203279889-203279889 on Build GRCh38
rs774656220 Chr.2: 203280012-203280012 on Build GRCh38
rs373713500 Chr.2: 203280071-203280071 on Build GRCh38
rs941447769 Chr.2: 203279719-203279719 on Build GRCh38
rs945451415 Chr.2: 203279802-203279802 on Build GRCh38
rs908668831 Chr.2: 203279718-203279718 on Build GRCh38
rs975683077 Chr.2: 203279681-203279681 on Build GRCh38
rs11690137 Chr.2: 203279737-203279737 on Build GRCh38
rs746266477 Chr.2: 203280013-203280013 on Build GRCh38
rs917968130 Chr.2: 203279772-203279772 on Build GRCh38
rs959599432 Chr.2: 203279747-203279747 on Build GRCh38
rs530176171 Chr.2: 203279717-203279717 on Build GRCh38
rs559809638 Chr.2: 203279763-203279763 on Build GRCh38
rs924445455 Chr.2: 203279887-203279887 on Build GRCh38
rs757323627 Chr.2: 203280074-203280076 on Build GRCh38
rs1040052628 Chr.2: 203280000-203280000 on Build GRCh38
rs760128483 Chr.2: 203280063-203280063 on Build GRCh38
rs772758149 Chr.2: 203280044-203280044 on Build GRCh38
rs553312682 Chr.2: 203279722-203279731 on Build GRCh38
rs965839974 Chr.2: 203279666-203279666 on Build GRCh38
rs767638726 Chr.2: 203280062-203280062 on Build GRCh38
rs560313349 Chr.2: 203279759-203279759 on Build GRCh38
rs117634224 Chr.2: 203279683-203279683 on Build GRCh38
rs567872480 Chr.2: 203279803-203279803 on Build GRCh38
rs896385869 Chr.2: 203279852-203279852 on Build GRCh38

View Full Product Details