Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49487184-49487650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAG1
Entrez Gene ID
1605
Gene Name
dystroglycan 1
Gene Aliases
156DAG, A3a, AGRNR, DAG, MDDGA9, MDDGC7, MDDGC9
UniGene
Hs.76111
Gene Chromosome Location
Chr.3: 49468703-49535618 on Build GRCh38
Transcripts
NM_001165928.3 NM_001177634.2 NM_001177635.2 NM_001177636.2 NM_001177637.2 NM_001177638.2 NM_001177639.2 NM_001177640.2 NM_001177641.2 NM_001177642.2 NM_001177643.2 NM_001177644.2 NM_004393.5

SNP Detail

SNP ID Chromosome Location
rs117264689 Chr.3: 49487428-49487428 on Build GRCh38
rs367892973 Chr.3: 49487523-49487523 on Build GRCh38
rs905012639 Chr.3: 49487311-49487311 on Build GRCh38
rs956117224 Chr.3: 49487326-49487326 on Build GRCh38
rs1020591384 Chr.3: 49487370-49487370 on Build GRCh38
rs975018478 Chr.3: 49487398-49487398 on Build GRCh38
rs144338789 Chr.3: 49487477-49487477 on Build GRCh38
rs199830382 Chr.3: 49487508-49487508 on Build GRCh38
rs966015026 Chr.3: 49487360-49487360 on Build GRCh38
rs905751363 Chr.3: 49487550-49487550 on Build GRCh38
rs1022824083 Chr.3: 49487275-49487275 on Build GRCh38
rs867889655 Chr.3: 49487283-49487283 on Build GRCh38
rs554279847 Chr.3: 49487268-49487268 on Build GRCh38
rs77537969 Chr.3: 49487316-49487316 on Build GRCh38
rs80020442 Chr.3: 49487566-49487566 on Build GRCh38
rs1038645669 Chr.3: 49487304-49487304 on Build GRCh38
rs752199638 Chr.3: 49487222-49487222 on Build GRCh38
rs939757529 Chr.3: 49487517-49487517 on Build GRCh38
rs771483488 Chr.3: 49487444-49487444 on Build GRCh38
rs979194730 Chr.3: 49487423-49487423 on Build GRCh38
rs1036665289 Chr.3: 49487218-49487218 on Build GRCh38
rs552765791 Chr.3: 49487252-49487252 on Build GRCh38
rs1010343166 Chr.3: 49487616-49487616 on Build GRCh38
rs949714142 Chr.3: 49487533-49487533 on Build GRCh38
rs891892100 Chr.3: 49487594-49487594 on Build GRCh38
rs999985731 Chr.3: 49487300-49487300 on Build GRCh38
rs984937527 Chr.3: 49487488-49487488 on Build GRCh38
rs916943928 Chr.3: 49487526-49487526 on Build GRCh38
rs1034961083 Chr.3: 49487317-49487317 on Build GRCh38
rs182065802 Chr.3: 49487250-49487250 on Build GRCh38
rs908543039 Chr.3: 49487541-49487541 on Build GRCh38
rs934008728 Chr.3: 49487259-49487259 on Build GRCh38
rs909630374 Chr.3: 49487504-49487504 on Build GRCh38
rs1018922076 Chr.3: 49487356-49487356 on Build GRCh38
rs922866864 Chr.3: 49487487-49487487 on Build GRCh38
rs1044330425 Chr.3: 49487535-49487535 on Build GRCh38
rs577400159 Chr.3: 49487230-49487230 on Build GRCh38
rs1000446422 Chr.3: 49487557-49487557 on Build GRCh38
rs1047092750 Chr.3: 49487537-49487537 on Build GRCh38
rs939683422 Chr.3: 49487215-49487215 on Build GRCh38
rs185036262 Chr.3: 49487611-49487611 on Build GRCh38
rs964265274 Chr.3: 49487440-49487440 on Build GRCh38
rs6783003 Chr.3: 49487345-49487345 on Build GRCh38
rs967826921 Chr.3: 49487396-49487396 on Build GRCh38

View Full Product Details