Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 412034-412531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR2
Entrez Gene ID
26063
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
Gene Aliases
PDCR, SDR17C1
UniGene
Hs.628831
Gene Chromosome Location
Chr.16: 401858-412487 on Build GRCh38
Transcripts
NM_020664.3

SNP Detail

SNP ID Chromosome Location
rs575202895 Chr.16: 412090-412115 on Build GRCh38
rs539439704 Chr.16: 412194-412194 on Build GRCh38
rs994506231 Chr.16: 412306-412306 on Build GRCh38
rs1050707340 Chr.16: 412250-412250 on Build GRCh38
rs139886474 Chr.16: 412397-412397 on Build GRCh38
rs897608597 Chr.16: 412304-412304 on Build GRCh38
rs912034642 Chr.16: 412279-412279 on Build GRCh38
rs997585353 Chr.16: 412341-412341 on Build GRCh38
rs367933977 Chr.16: 412456-412459 on Build GRCh38
rs748040306 Chr.16: 412127-412127 on Build GRCh38
rs777772363 Chr.16: 412230-412230 on Build GRCh38
rs763896291 Chr.16: 412384-412384 on Build GRCh38
rs1036126287 Chr.16: 412100-412100 on Build GRCh38
rs1020316028 Chr.16: 412364-412364 on Build GRCh38
rs932184137 Chr.16: 412231-412231 on Build GRCh38
rs1045098 Chr.16: 412343-412343 on Build GRCh38
rs944844434 Chr.16: 412497-412497 on Build GRCh38
rs533096280 Chr.16: 412435-412435 on Build GRCh38
rs769668190 Chr.16: 412165-412165 on Build GRCh38
rs1030090024 Chr.16: 412344-412344 on Build GRCh38
rs890631322 Chr.16: 412081-412081 on Build GRCh38
rs1009380633 Chr.16: 412362-412362 on Build GRCh38
rs986763047 Chr.16: 412400-412400 on Build GRCh38
rs1045090 Chr.16: 412315-412315 on Build GRCh38
rs775462770 Chr.16: 412223-412223 on Build GRCh38
rs1002041385 Chr.16: 412138-412138 on Build GRCh38
rs897470133 Chr.16: 412101-412101 on Build GRCh38
rs970786049 Chr.16: 412499-412499 on Build GRCh38
rs142570754 Chr.16: 412285-412285 on Build GRCh38
rs1040489223 Chr.16: 412069-412094 on Build GRCh38
rs575380193 Chr.16: 412176-412176 on Build GRCh38
rs371859523 Chr.16: 412498-412498 on Build GRCh38
rs76469232 Chr.16: 412280-412280 on Build GRCh38
rs546851304 Chr.16: 412108-412108 on Build GRCh38
rs982258664 Chr.16: 412257-412257 on Build GRCh38
rs568486109 Chr.16: 412284-412284 on Build GRCh38
rs967337612 Chr.16: 412199-412199 on Build GRCh38
rs894025676 Chr.16: 412087-412087 on Build GRCh38
rs551069682 Chr.16: 412479-412479 on Build GRCh38
rs557038032 Chr.16: 412168-412168 on Build GRCh38
rs1016078742 Chr.16: 412377-412377 on Build GRCh38
rs762672239 Chr.16: 412328-412329 on Build GRCh38
rs912171721 Chr.16: 412422-412422 on Build GRCh38
rs1008617574 Chr.16: 412096-412096 on Build GRCh38
rs573886893 Chr.16: 412391-412391 on Build GRCh38
rs201853407 Chr.16: 412451-412454 on Build GRCh38
rs1007411585 Chr.16: 412367-412367 on Build GRCh38
rs905385700 Chr.16: 412067-412067 on Build GRCh38
rs960506817 Chr.16: 412342-412342 on Build GRCh38
rs148421637 Chr.16: 412115-412115 on Build GRCh38
rs1027560620 Chr.16: 412120-412120 on Build GRCh38
rs368865292 Chr.16: 412324-412324 on Build GRCh38
rs1035228728 Chr.16: 412139-412139 on Build GRCh38
rs1045101 Chr.16: 412361-412361 on Build GRCh38
rs1049244115 Chr.16: 412308-412308 on Build GRCh38
rs568332503 Chr.16: 412113-412113 on Build GRCh38
rs746060124 Chr.16: 412355-412355 on Build GRCh38
rs967472134 Chr.16: 412385-412385 on Build GRCh38
rs978869108 Chr.16: 412390-412390 on Build GRCh38
rs150656216 Chr.16: 412323-412323 on Build GRCh38
rs1011197585 Chr.16: 412092-412092 on Build GRCh38
rs9930335 Chr.16: 412338-412338 on Build GRCh38
rs992846187 Chr.16: 412173-412173 on Build GRCh38
rs115651634 Chr.16: 412395-412395 on Build GRCh38
rs564027626 Chr.16: 412075-412075 on Build GRCh38
rs1030475407 Chr.16: 412131-412131 on Build GRCh38
rs1026234606 Chr.16: 412490-412490 on Build GRCh38

View Full Product Details