Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65463651-65464087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP8
Entrez Gene ID
54878
Gene Name
dipeptidyl peptidase 8
Gene Aliases
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
UniGene
Hs.458609
Gene Chromosome Location
Chr.15: 65442463-65517713 on Build GRCh38
Transcripts
NM_001320875.1 NM_001320876.1 NM_017743.5 NM_130434.4 NM_197960.3 NM_197961.3 NR_135485.1 NR_135486.1 XM_005254505.1 XM_011521730.1 XM_011521731.1 XM_011521732.2 XM_011521733.1 XM_011521734.1 XM_017022373.1 XM_017022374.1 XM_017022375.1 XM_017022376.1 XM_017022377.1 XM_017022378.1 XM_017022379.1 XM_017022380.1 XM_017022381.1 XM_017022382.1 XM_017022383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1301346 c.1844C>T p.T615I Chr.15: 65463888-65463888 on Build GRCh38
4870888 c.1964A>T p.D655V Chr.15: 65463816-65463816 on Build GRCh38
6574977 c.1969C>T p.Q657* Chr.15: 65463763-65463763 on Build GRCh38
3932136 c.1910C>G p.S637C Chr.15: 65463870-65463870 on Build GRCh38
4599698 c.1978A>G p.K660E Chr.15: 65463802-65463802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040298420 Chr.15: 65463687-65463687 on Build GRCh38
rs923594204 Chr.15: 65463678-65463678 on Build GRCh38
rs967857434 Chr.15: 65463991-65463991 on Build GRCh38
rs369179039 Chr.15: 65463832-65463832 on Build GRCh38
rs774406925 Chr.15: 65463880-65463880 on Build GRCh38
rs1034579975 Chr.15: 65463953-65463953 on Build GRCh38
rs772569533 Chr.15: 65464040-65464040 on Build GRCh38
rs923626569 Chr.15: 65464052-65464052 on Build GRCh38
rs773014230 Chr.15: 65463930-65463930 on Build GRCh38
rs755037917 Chr.15: 65463910-65463910 on Build GRCh38
rs556197235 Chr.15: 65463962-65463962 on Build GRCh38
rs780132902 Chr.15: 65463738-65463738 on Build GRCh38
rs34898704 Chr.15: 65463900-65463901 on Build GRCh38
rs201526280 Chr.15: 65463949-65463949 on Build GRCh38
rs61736633 Chr.15: 65463788-65463788 on Build GRCh38
rs1022473529 Chr.15: 65463984-65463984 on Build GRCh38
rs569084315 Chr.15: 65463696-65463696 on Build GRCh38
rs996525832 Chr.15: 65463801-65463801 on Build GRCh38
rs762648468 Chr.15: 65463794-65463794 on Build GRCh38
rs781318317 Chr.15: 65463911-65463911 on Build GRCh38
rs769346470 Chr.15: 65463974-65463974 on Build GRCh38
rs868735619 Chr.15: 65463918-65463918 on Build GRCh38
rs552270623 Chr.15: 65463675-65463675 on Build GRCh38
rs995699370 Chr.15: 65463710-65463710 on Build GRCh38
rs990297065 Chr.15: 65463978-65463978 on Build GRCh38
rs779485010 Chr.15: 65463732-65463732 on Build GRCh38
rs749489221 Chr.15: 65463680-65463680 on Build GRCh38
rs372897611 Chr.15: 65463933-65463933 on Build GRCh38
rs531488500 Chr.15: 65463721-65463721 on Build GRCh38
rs918025133 Chr.15: 65464007-65464007 on Build GRCh38
rs371554795 Chr.15: 65463814-65463814 on Build GRCh38
rs758978456 Chr.15: 65463815-65463815 on Build GRCh38
rs201962360 Chr.15: 65463889-65463889 on Build GRCh38
rs3825891 Chr.15: 65463734-65463734 on Build GRCh38
rs905482857 Chr.15: 65463689-65463689 on Build GRCh38
rs112046769 Chr.15: 65464003-65464003 on Build GRCh38
rs367934072 Chr.15: 65463964-65463964 on Build GRCh38
rs747052608 Chr.15: 65463900-65463900 on Build GRCh38
rs747089802 Chr.15: 65463742-65463742 on Build GRCh38
rs576231429 Chr.15: 65464000-65464000 on Build GRCh38
rs548080372 Chr.15: 65463726-65463726 on Build GRCh38
rs780143724 Chr.15: 65463891-65463891 on Build GRCh38
rs542453731 Chr.15: 65464016-65464016 on Build GRCh38
rs757876927 Chr.15: 65463854-65463854 on Build GRCh38
rs764732665 Chr.15: 65463720-65463720 on Build GRCh38
rs924471092 Chr.15: 65463951-65463951 on Build GRCh38
rs369131089 Chr.15: 65463776-65463776 on Build GRCh38
rs754389779 Chr.15: 65463851-65463851 on Build GRCh38
rs758594957 Chr.15: 65463735-65463735 on Build GRCh38
rs776564607 Chr.15: 65463782-65463782 on Build GRCh38
rs748048066 Chr.15: 65463925-65463925 on Build GRCh38
rs112569209 Chr.15: 65463728-65463728 on Build GRCh38
rs765973271 Chr.15: 65463805-65463805 on Build GRCh38
rs1004755294 Chr.15: 65463754-65463754 on Build GRCh38
rs142999124 Chr.15: 65463858-65463858 on Build GRCh38
rs887275260 Chr.15: 65463741-65463741 on Build GRCh38
rs772923711 Chr.15: 65463793-65463793 on Build GRCh38

View Full Product Details