Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99698695-99699194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX15
Entrez Gene ID
1355
Gene Name
COX15, cytochrome c oxidase assembly homolog
Gene Aliases
CEMCOX2
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99694793-99732667 on Build GRCh38
Transcripts
NM_001320974.1 XM_006717634.3 XM_017015715.1 NM_020354.3

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99659506-99711245 on Build GRCh38
Transcripts
NM_001320974.1 XM_006717634.3 XM_017015715.1 NM_020354.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950463 c.1246_1254delTACGGCTTC p.Y416_F418delYGF Chr.10: 99698769-99698777 on Build GRCh38
4969689 c.1248C>T p.Y416Y Chr.10: 99698771-99698771 on Build GRCh38
6064875 c.1192G>C p.A398P Chr.10: 99698715-99698715 on Build GRCh38
4639645 c.1289G>A p.R430H Chr.10: 99698812-99698812 on Build GRCh38
913779 c.1301G>A p.R434H Chr.10: 99698824-99698824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764542399 Chr.10: 99698779-99698779 on Build GRCh38
rs769743709 Chr.10: 99698858-99698858 on Build GRCh38
rs763404254 Chr.10: 99698907-99698907 on Build GRCh38
rs962818599 Chr.10: 99698925-99698925 on Build GRCh38
rs758101413 Chr.10: 99698823-99698823 on Build GRCh38
rs374095085 Chr.10: 99698895-99698895 on Build GRCh38
rs778461277 Chr.10: 99698732-99698732 on Build GRCh38
rs201688058 Chr.10: 99698786-99698786 on Build GRCh38
rs879255416 Chr.10: 99698834-99698834 on Build GRCh38
rs532151677 Chr.10: 99699057-99699057 on Build GRCh38
rs754592693 Chr.10: 99698731-99698731 on Build GRCh38
rs557257815 Chr.10: 99698719-99698719 on Build GRCh38
rs567016128 Chr.10: 99699150-99699150 on Build GRCh38
rs770792937 Chr.10: 99698826-99698826 on Build GRCh38
rs149136007 Chr.10: 99698741-99698741 on Build GRCh38
rs184607396 Chr.10: 99699158-99699158 on Build GRCh38
rs183633106 Chr.10: 99698853-99698853 on Build GRCh38
rs374494342 Chr.10: 99698824-99698824 on Build GRCh38
rs752039062 Chr.10: 99698780-99698780 on Build GRCh38
rs763265710 Chr.10: 99698890-99698890 on Build GRCh38
rs907468283 Chr.10: 99699004-99699004 on Build GRCh38
rs193162909 Chr.10: 99698915-99698915 on Build GRCh38
rs563166590 Chr.10: 99699105-99699105 on Build GRCh38
rs752266858 Chr.10: 99698818-99698818 on Build GRCh38
rs368809171 Chr.10: 99698740-99698740 on Build GRCh38
rs746448756 Chr.10: 99698811-99698811 on Build GRCh38
rs781566336 Chr.10: 99698768-99698768 on Build GRCh38
rs936080505 Chr.10: 99698929-99698929 on Build GRCh38
rs975729234 Chr.10: 99698852-99698852 on Build GRCh38
rs894480049 Chr.10: 99698972-99698972 on Build GRCh38
rs778524683 Chr.10: 99698812-99698812 on Build GRCh38
rs141762300 Chr.10: 99698772-99698772 on Build GRCh38
rs61744268 Chr.10: 99698720-99698720 on Build GRCh38
rs775699176 Chr.10: 99698859-99698859 on Build GRCh38
rs924663608 Chr.10: 99698924-99698924 on Build GRCh38
rs979711815 Chr.10: 99699154-99699154 on Build GRCh38
rs548681448 Chr.10: 99699131-99699131 on Build GRCh38
rs764457469 Chr.10: 99699114-99699114 on Build GRCh38
rs762345381 Chr.10: 99698781-99698781 on Build GRCh38
rs1018218788 Chr.10: 99699055-99699055 on Build GRCh38
rs747927998 Chr.10: 99698733-99698733 on Build GRCh38
rs781507848 Chr.10: 99698850-99698850 on Build GRCh38
rs530574230 Chr.10: 99699110-99699110 on Build GRCh38
rs533054127 Chr.10: 99699041-99699041 on Build GRCh38
rs753340088 Chr.10: 99698793-99698793 on Build GRCh38
rs1054420384 Chr.10: 99698944-99698944 on Build GRCh38
rs1046385506 Chr.10: 99699003-99699003 on Build GRCh38
rs759561702 Chr.10: 99698763-99698763 on Build GRCh38
rs746807230 Chr.10: 99698825-99698825 on Build GRCh38
rs763317784 Chr.10: 99698777-99698777 on Build GRCh38
rs61729540 Chr.10: 99698771-99698771 on Build GRCh38
rs188569806 Chr.10: 99698912-99698912 on Build GRCh38
rs948776527 Chr.10: 99698976-99698976 on Build GRCh38
rs775759283 Chr.10: 99698862-99698865 on Build GRCh38
rs776719484 Chr.10: 99698747-99698747 on Build GRCh38
rs781027538 Chr.10: 99698851-99698851 on Build GRCh38
rs753300933 Chr.10: 99698730-99698730 on Build GRCh38
rs757905285 Chr.10: 99698729-99698729 on Build GRCh38
rs986695592 Chr.10: 99699053-99699053 on Build GRCh38
rs555052941 Chr.10: 99698811-99698811 on Build GRCh38
rs146917199 Chr.10: 99698745-99698745 on Build GRCh38
rs770917437 Chr.10: 99698746-99698746 on Build GRCh38
rs375158055 Chr.10: 99698738-99698738 on Build GRCh38
rs752079975 Chr.10: 99698725-99698725 on Build GRCh38

View Full Product Details