Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99691415-99691951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99659506-99711245 on Build GRCh38
Transcripts
NM_020354.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6518185 c.784G>A p.A262T Chr.10: 99691459-99691459 on Build GRCh38
5049819 c.748C>T p.R250W Chr.10: 99691423-99691423 on Build GRCh38
426907 c.772G>T p.D258Y Chr.10: 99691447-99691447 on Build GRCh38
6654342 c.749G>A p.R250Q Chr.10: 99691424-99691424 on Build GRCh38
6381002 c.761T>A p.V254E Chr.10: 99691436-99691436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184757665 Chr.10: 99691779-99691779 on Build GRCh38
rs935935887 Chr.10: 99691903-99691903 on Build GRCh38
rs778999791 Chr.10: 99691455-99691455 on Build GRCh38
rs781604372 Chr.10: 99691926-99691926 on Build GRCh38
rs963733225 Chr.10: 99691717-99691717 on Build GRCh38
rs1013013737 Chr.10: 99691597-99691597 on Build GRCh38
rs1045722254 Chr.10: 99691644-99691644 on Build GRCh38
rs374879820 Chr.10: 99691561-99691561 on Build GRCh38
rs758575613 Chr.10: 99691460-99691460 on Build GRCh38
rs753334722 Chr.10: 99691754-99691754 on Build GRCh38
rs747188963 Chr.10: 99691466-99691466 on Build GRCh38
rs577180618 Chr.10: 99691476-99691476 on Build GRCh38
rs148606856 Chr.10: 99691473-99691473 on Build GRCh38
rs1018204714 Chr.10: 99691709-99691709 on Build GRCh38
rs754854032 Chr.10: 99691528-99691528 on Build GRCh38
rs907199195 Chr.10: 99691653-99691653 on Build GRCh38
rs756066551 Chr.10: 99691707-99691707 on Build GRCh38
rs75877025 Chr.10: 99691745-99691745 on Build GRCh38
rs1026553086 Chr.10: 99691702-99691702 on Build GRCh38
rs754816498 Chr.10: 99691451-99691451 on Build GRCh38
rs560281113 Chr.10: 99691536-99691536 on Build GRCh38
rs369321164 Chr.10: 99691481-99691481 on Build GRCh38
rs747768309 Chr.10: 99691791-99691791 on Build GRCh38
rs565106446 Chr.10: 99691749-99691750 on Build GRCh38
rs188763532 Chr.10: 99691532-99691532 on Build GRCh38
rs1025851530 Chr.10: 99691643-99691643 on Build GRCh38
rs10748788 Chr.10: 99691789-99691789 on Build GRCh38
rs11190245 Chr.10: 99691502-99691502 on Build GRCh38
rs72832512 Chr.10: 99691437-99691437 on Build GRCh38
rs146737561 Chr.10: 99691779-99691780 on Build GRCh38
rs1041614029 Chr.10: 99691478-99691478 on Build GRCh38
rs772848315 Chr.10: 99691705-99691705 on Build GRCh38
rs993529072 Chr.10: 99691665-99691665 on Build GRCh38
rs569560241 Chr.10: 99691927-99691927 on Build GRCh38
rs755753985 Chr.10: 99691519-99691519 on Build GRCh38
rs771682518 Chr.10: 99691504-99691504 on Build GRCh38
rs181580618 Chr.10: 99691628-99691628 on Build GRCh38
rs772845848 Chr.10: 99691511-99691511 on Build GRCh38
rs753511391 Chr.10: 99691525-99691525 on Build GRCh38
rs758453824 Chr.10: 99691554-99691554 on Build GRCh38
rs761629032 Chr.10: 99691496-99691496 on Build GRCh38
rs560764796 Chr.10: 99691610-99691610 on Build GRCh38
rs748202130 Chr.10: 99691456-99691456 on Build GRCh38
rs775997005 Chr.10: 99691488-99691488 on Build GRCh38
rs935273711 Chr.10: 99691806-99691806 on Build GRCh38
rs994279748 Chr.10: 99691642-99691642 on Build GRCh38
rs778053103 Chr.10: 99691463-99691463 on Build GRCh38
rs763499551 Chr.10: 99691489-99691489 on Build GRCh38
rs753724661 Chr.10: 99691443-99691443 on Build GRCh38
rs1018443704 Chr.10: 99691698-99691699 on Build GRCh38
rs76728867 Chr.10: 99691596-99691596 on Build GRCh38
rs770245006 Chr.10: 99691486-99691486 on Build GRCh38
rs11190246 Chr.10: 99691854-99691854 on Build GRCh38
rs977086101 Chr.10: 99691802-99691802 on Build GRCh38

View Full Product Details