Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 11104610-11105106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERVFRD-1
Entrez Gene ID
405754
Gene Name
endogenous retrovirus group FRD member 1
Gene Aliases
ERVFRDE1, GLLL6191, HERV-FRD, HERV-W/FRD, UNQ6191, envFRD
UniGene
Hs.631996
Gene Chromosome Location
Chr.6: 11102489-11111838 on Build GRCh38
Transcripts
NM_207582.2 NM_001135575.1

Gene Symbol
SMIM13
Entrez Gene ID
221710
Gene Name
small integral membrane protein 13
Gene Aliases
C6orf228
UniGene
Hs.656600
Gene Chromosome Location
Chr.6: 11094033-11138738 on Build GRCh38
Transcripts
NM_207582.2 NM_001135575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366259 c.602G>A p.S201N Chr.6: 11104709-11104709 on Build GRCh38
3155910 c.326T>G p.I109S Chr.6: 11104985-11104985 on Build GRCh38
5450171 c.531T>C p.N177N Chr.6: 11104780-11104780 on Build GRCh38
74891 c.221T>G p.I74S Chr.6: 11105090-11105090 on Build GRCh38
180810 c.675G>A p.A225A Chr.6: 11104636-11104636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765650768 Chr.6: 11104953-11104953 on Build GRCh38
rs757819098 Chr.6: 11104766-11104766 on Build GRCh38
rs774035002 Chr.6: 11104784-11104784 on Build GRCh38
rs768146373 Chr.6: 11104783-11104783 on Build GRCh38
rs900271216 Chr.6: 11105003-11105003 on Build GRCh38
rs531962436 Chr.6: 11104902-11104902 on Build GRCh38
rs758368739 Chr.6: 11105016-11105016 on Build GRCh38
rs201709473 Chr.6: 11104757-11104757 on Build GRCh38
rs373531632 Chr.6: 11105001-11105001 on Build GRCh38
rs11753622 Chr.6: 11104880-11104880 on Build GRCh38
rs781681644 Chr.6: 11104873-11104873 on Build GRCh38
rs767472288 Chr.6: 11104688-11104688 on Build GRCh38
rs771992064 Chr.6: 11104794-11104794 on Build GRCh38
rs542375812 Chr.6: 11104952-11104952 on Build GRCh38
rs764429308 Chr.6: 11104843-11104843 on Build GRCh38
rs773979486 Chr.6: 11105032-11105034 on Build GRCh38
rs764409358 Chr.6: 11104675-11104675 on Build GRCh38
rs770816762 Chr.6: 11105079-11105079 on Build GRCh38
rs756641090 Chr.6: 11105036-11105036 on Build GRCh38
rs747563224 Chr.6: 11104637-11104637 on Build GRCh38
rs760765071 Chr.6: 11104698-11104698 on Build GRCh38
rs369615242 Chr.6: 11105000-11105000 on Build GRCh38
rs377429449 Chr.6: 11104633-11104633 on Build GRCh38
rs766557849 Chr.6: 11104703-11104703 on Build GRCh38
rs758079525 Chr.6: 11105045-11105045 on Build GRCh38
rs917003507 Chr.6: 11104738-11104738 on Build GRCh38
rs370411526 Chr.6: 11104910-11104910 on Build GRCh38
rs767814724 Chr.6: 11105004-11105004 on Build GRCh38
rs547443793 Chr.6: 11105061-11105061 on Build GRCh38
rs201156872 Chr.6: 11104658-11104658 on Build GRCh38
rs752931524 Chr.6: 11104831-11104831 on Build GRCh38
rs137974365 Chr.6: 11104704-11104704 on Build GRCh38
rs138512061 Chr.6: 11104936-11104936 on Build GRCh38
rs752264135 Chr.6: 11105041-11105041 on Build GRCh38
rs777093759 Chr.6: 11104668-11104668 on Build GRCh38
rs1011677696 Chr.6: 11104984-11104984 on Build GRCh38
rs201528012 Chr.6: 11104770-11104770 on Build GRCh38
rs777639545 Chr.6: 11104944-11104944 on Build GRCh38
rs760104349 Chr.6: 11104882-11104883 on Build GRCh38
rs140934168 Chr.6: 11104811-11104811 on Build GRCh38
rs778893236 Chr.6: 11104689-11104689 on Build GRCh38
rs765281057 Chr.6: 11104828-11104828 on Build GRCh38
rs769911187 Chr.6: 11104966-11104966 on Build GRCh38
rs778432865 Chr.6: 11105037-11105037 on Build GRCh38
rs74690822 Chr.6: 11105081-11105081 on Build GRCh38
rs773979029 Chr.6: 11104672-11104672 on Build GRCh38
rs757096237 Chr.6: 11104687-11104687 on Build GRCh38
rs188870088 Chr.6: 11104961-11104961 on Build GRCh38
rs893016711 Chr.6: 11104977-11104977 on Build GRCh38
rs559395063 Chr.6: 11104636-11104636 on Build GRCh38
rs763103170 Chr.6: 11104994-11104994 on Build GRCh38
rs375359410 Chr.6: 11104918-11104918 on Build GRCh38
rs773203163 Chr.6: 11104795-11104795 on Build GRCh38
rs765524999 Chr.6: 11104725-11104725 on Build GRCh38
rs776530304 Chr.6: 11104812-11104812 on Build GRCh38
rs775048038 Chr.6: 11104657-11104657 on Build GRCh38
rs561422018 Chr.6: 11104841-11104841 on Build GRCh38
rs752953220 Chr.6: 11104739-11104739 on Build GRCh38
rs771444243 Chr.6: 11104644-11104644 on Build GRCh38
rs748546197 Chr.6: 11104631-11104631 on Build GRCh38
rs949485572 Chr.6: 11104819-11104819 on Build GRCh38
rs141542647 Chr.6: 11105017-11105017 on Build GRCh38
rs748861093 Chr.6: 11104782-11104782 on Build GRCh38
rs200769035 Chr.6: 11104897-11104897 on Build GRCh38
rs759681553 Chr.6: 11104818-11104818 on Build GRCh38
rs543348563 Chr.6: 11104788-11104788 on Build GRCh38
rs372287476 Chr.6: 11104958-11104958 on Build GRCh38
rs144698959 Chr.6: 11104861-11104861 on Build GRCh38
rs968627583 Chr.6: 11104666-11104666 on Build GRCh38
rs759430701 Chr.6: 11104963-11104963 on Build GRCh38
rs754173224 Chr.6: 11104686-11104686 on Build GRCh38
rs750277998 Chr.6: 11104864-11104864 on Build GRCh38
rs758765949 Chr.6: 11104740-11104740 on Build GRCh38
rs1056843483 Chr.6: 11104716-11104716 on Build GRCh38
rs768360608 Chr.6: 11104663-11104663 on Build GRCh38
rs747510057 Chr.6: 11104758-11104758 on Build GRCh38
rs755085420 Chr.6: 11104724-11104724 on Build GRCh38
rs887083915 Chr.6: 11104884-11104884 on Build GRCh38
rs775489257 Chr.6: 11104992-11104992 on Build GRCh38
rs747826273 Chr.6: 11104928-11104928 on Build GRCh38

View Full Product Details