Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241866691-241867097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO1
Entrez Gene ID
9156
Gene Name
exonuclease 1
Gene Aliases
HEX1, hExoI
UniGene
Hs.498248
Gene Chromosome Location
Chr.1: 241847967-241889939 on Build GRCh38
Transcripts
NM_001319224.1 NM_003686.4 NM_006027.4 NM_130398.3 XM_006711840.2 XM_011544321.1 XM_011544322.1 XM_011544323.2 XM_011544324.2 XM_011544325.2 XM_017002793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681989 c.1122T>C p.N374N Chr.1: 241866910-241866910 on Build GRCh38
6062489 c.1155A>T p.S385S Chr.1: 241866943-241866943 on Build GRCh38
7136213 c.1228C>T p.L410F Chr.1: 241867016-241867016 on Build GRCh38
5437602 c.1130C>G p.P377R Chr.1: 241866918-241866918 on Build GRCh38
13945 c.1119G>A p.R373R Chr.1: 241866907-241866907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751275581 Chr.1: 241867001-241867001 on Build GRCh38
rs780398481 Chr.1: 241866880-241866880 on Build GRCh38
rs148510810 Chr.1: 241867053-241867053 on Build GRCh38
rs774688889 Chr.1: 241866955-241866955 on Build GRCh38
rs753120591 Chr.1: 241867066-241867066 on Build GRCh38
rs769046083 Chr.1: 241866814-241866814 on Build GRCh38
rs566255049 Chr.1: 241866719-241866719 on Build GRCh38
rs558408748 Chr.1: 241866746-241866746 on Build GRCh38
rs954452620 Chr.1: 241866949-241866949 on Build GRCh38
rs987962104 Chr.1: 241866857-241866857 on Build GRCh38
rs761759671 Chr.1: 241866977-241866977 on Build GRCh38
rs758060228 Chr.1: 241866870-241866870 on Build GRCh38
rs762348945 Chr.1: 241867049-241867049 on Build GRCh38
rs766149355 Chr.1: 241866837-241866837 on Build GRCh38
rs763985875 Chr.1: 241866972-241866972 on Build GRCh38
rs556027234 Chr.1: 241866833-241866833 on Build GRCh38
rs183040380 Chr.1: 241866835-241866835 on Build GRCh38
rs757992445 Chr.1: 241866990-241866990 on Build GRCh38
rs767938801 Chr.1: 241867060-241867060 on Build GRCh38
rs768283548 Chr.1: 241867032-241867032 on Build GRCh38
rs755284086 Chr.1: 241866752-241866753 on Build GRCh38
rs149029711 Chr.1: 241866834-241866834 on Build GRCh38
rs200612631 Chr.1: 241866931-241866931 on Build GRCh38
rs532992049 Chr.1: 241867007-241867007 on Build GRCh38
rs539525213 Chr.1: 241866819-241866819 on Build GRCh38
rs735943 Chr.1: 241866849-241866849 on Build GRCh38
rs944391727 Chr.1: 241866935-241866935 on Build GRCh38
rs753922756 Chr.1: 241866747-241866748 on Build GRCh38
rs748705205 Chr.1: 241866902-241866902 on Build GRCh38
rs773192633 Chr.1: 241867045-241867045 on Build GRCh38
rs761669697 Chr.1: 241867035-241867035 on Build GRCh38
rs543887227 Chr.1: 241867029-241867029 on Build GRCh38
rs764668398 Chr.1: 241866989-241866989 on Build GRCh38
rs535198503 Chr.1: 241866737-241866737 on Build GRCh38
rs756726442 Chr.1: 241866866-241866866 on Build GRCh38
rs571928768 Chr.1: 241867017-241867017 on Build GRCh38
rs769717898 Chr.1: 241867036-241867036 on Build GRCh38
rs202181576 Chr.1: 241866869-241866869 on Build GRCh38
rs1028737484 Chr.1: 241866915-241866915 on Build GRCh38
rs910179760 Chr.1: 241866896-241866896 on Build GRCh38
rs373381159 Chr.1: 241867033-241867033 on Build GRCh38
rs751133380 Chr.1: 241867059-241867059 on Build GRCh38
rs752993922 Chr.1: 241866958-241866960 on Build GRCh38
rs770043352 Chr.1: 241866824-241866824 on Build GRCh38
rs367681310 Chr.1: 241866976-241866976 on Build GRCh38
rs773451365 Chr.1: 241866930-241866930 on Build GRCh38
rs754871794 Chr.1: 241866882-241866882 on Build GRCh38
rs753819273 Chr.1: 241866851-241866851 on Build GRCh38
rs35794652 Chr.1: 241866868-241866868 on Build GRCh38
rs781529397 Chr.1: 241866901-241866901 on Build GRCh38
rs149349773 Chr.1: 241866905-241866905 on Build GRCh38
rs745556629 Chr.1: 241866785-241866785 on Build GRCh38
rs201387950 Chr.1: 241866806-241866806 on Build GRCh38
rs756618089 Chr.1: 241867071-241867071 on Build GRCh38
rs780304214 Chr.1: 241866794-241866794 on Build GRCh38
rs985916495 Chr.1: 241866873-241866873 on Build GRCh38
rs767811883 Chr.1: 241866868-241866869 on Build GRCh38
rs748619330 Chr.1: 241867012-241867012 on Build GRCh38
rs756704351 Chr.1: 241867021-241867021 on Build GRCh38
rs947363952 Chr.1: 241866788-241866788 on Build GRCh38
rs760597120 Chr.1: 241866848-241866848 on Build GRCh38
rs986935204 Chr.1: 241866956-241866956 on Build GRCh38
rs748226923 Chr.1: 241866822-241866822 on Build GRCh38
rs755429902 Chr.1: 241866886-241866886 on Build GRCh38
rs757055833 Chr.1: 241867032-241867032 on Build GRCh38
rs778967797 Chr.1: 241867030-241867030 on Build GRCh38
rs367906829 Chr.1: 241866787-241866787 on Build GRCh38
rs369864583 Chr.1: 241867004-241867004 on Build GRCh38
rs199674255 Chr.1: 241866953-241866953 on Build GRCh38
rs1005660427 Chr.1: 241866780-241866780 on Build GRCh38
rs777937653 Chr.1: 241867024-241867024 on Build GRCh38
rs369253528 Chr.1: 241866839-241866839 on Build GRCh38
rs760362865 Chr.1: 241866965-241866965 on Build GRCh38

View Full Product Details