Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44095553-44096043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXT2
Entrez Gene ID
2132
Gene Name
exostosin glycosyltransferase 2
Gene Aliases
SOTV, SSMS
UniGene
Hs.368404
Gene Chromosome Location
Chr.11: 44095549-44245430 on Build GRCh38
Transcripts
NM_001178083.1 NM_207122.1 XM_011519951.1

SNP Detail

SNP ID Chromosome Location
rs1013685919 Chr.11: 44096014-44096014 on Build GRCh38
rs980029905 Chr.11: 44095853-44095853 on Build GRCh38
rs572718799 Chr.11: 44095733-44095733 on Build GRCh38
rs994045289 Chr.11: 44095887-44095887 on Build GRCh38
rs1040695368 Chr.11: 44095724-44095724 on Build GRCh38
rs751465262 Chr.11: 44095659-44095659 on Build GRCh38
rs977395242 Chr.11: 44095774-44095774 on Build GRCh38
rs947908856 Chr.11: 44095829-44095829 on Build GRCh38
rs1014579177 Chr.11: 44095799-44095799 on Build GRCh38
rs557739127 Chr.11: 44095788-44095788 on Build GRCh38
rs1027835725 Chr.11: 44095574-44095574 on Build GRCh38
rs946082301 Chr.11: 44095672-44095672 on Build GRCh38
rs531222883 Chr.11: 44095713-44095713 on Build GRCh38
rs12800404 Chr.11: 44095822-44095822 on Build GRCh38
rs960371927 Chr.11: 44095801-44095801 on Build GRCh38
rs12365753 Chr.11: 44095874-44095874 on Build GRCh38
rs12362775 Chr.11: 44095894-44095894 on Build GRCh38
rs3923808 Chr.11: 44095572-44095572 on Build GRCh38
rs887521497 Chr.11: 44095957-44095957 on Build GRCh38
rs904655925 Chr.11: 44095676-44095676 on Build GRCh38
rs925975964 Chr.11: 44095974-44095974 on Build GRCh38
rs115238130 Chr.11: 44095735-44095735 on Build GRCh38
rs991492005 Chr.11: 44095998-44095998 on Build GRCh38
rs549099091 Chr.11: 44095645-44095645 on Build GRCh38
rs906434941 Chr.11: 44095704-44095704 on Build GRCh38
rs894944784 Chr.11: 44095691-44095691 on Build GRCh38
rs796338269 Chr.11: 44095748-44095748 on Build GRCh38
rs916013137 Chr.11: 44096009-44096009 on Build GRCh38
rs922862352 Chr.11: 44095627-44095627 on Build GRCh38
rs58861092 Chr.11: 44095859-44095859 on Build GRCh38
rs1043796695 Chr.11: 44095682-44095682 on Build GRCh38
rs992280595 Chr.11: 44095622-44095622 on Build GRCh38
rs565589672 Chr.11: 44095975-44095975 on Build GRCh38
rs184253329 Chr.11: 44095618-44095618 on Build GRCh38
rs78395259 Chr.11: 44095686-44095686 on Build GRCh38
rs983836420 Chr.11: 44095712-44095712 on Build GRCh38
rs935298494 Chr.11: 44095701-44095701 on Build GRCh38
rs996005866 Chr.11: 44095918-44095918 on Build GRCh38
rs575980497 Chr.11: 44095817-44095817 on Build GRCh38
rs942971705 Chr.11: 44095609-44095609 on Build GRCh38
rs969557659 Chr.11: 44095836-44095836 on Build GRCh38
rs1052528701 Chr.11: 44095888-44095888 on Build GRCh38
rs1014631733 Chr.11: 44095959-44095959 on Build GRCh38
rs952440919 Chr.11: 44095708-44095708 on Build GRCh38
rs551657363 Chr.11: 44096011-44096011 on Build GRCh38
rs373686902 Chr.11: 44095952-44095952 on Build GRCh38
rs1027393651 Chr.11: 44095736-44095736 on Build GRCh38
rs992333007 Chr.11: 44095819-44095819 on Build GRCh38
rs1040262929 Chr.11: 44095904-44095904 on Build GRCh38
rs960985891 Chr.11: 44095612-44095612 on Build GRCh38
rs908117420 Chr.11: 44095610-44095610 on Build GRCh38
rs984325590 Chr.11: 44095594-44095594 on Build GRCh38
rs900446807 Chr.11: 44095912-44095912 on Build GRCh38
rs963821410 Chr.11: 44095949-44095949 on Build GRCh38
rs911338719 Chr.11: 44095600-44095601 on Build GRCh38
rs1008784470 Chr.11: 44095718-44095718 on Build GRCh38
rs983108788 Chr.11: 44095585-44095585 on Build GRCh38
rs950158579 Chr.11: 44096023-44096023 on Build GRCh38
rs998800511 Chr.11: 44095951-44095951 on Build GRCh38
rs951822395 Chr.11: 44095755-44095755 on Build GRCh38
rs900309940 Chr.11: 44095731-44095731 on Build GRCh38
rs1052473184 Chr.11: 44095706-44095706 on Build GRCh38
rs140369819 Chr.11: 44095958-44095958 on Build GRCh38
rs944037934 Chr.11: 44095897-44095897 on Build GRCh38
rs537149054 Chr.11: 44095643-44095643 on Build GRCh38
rs1044601491 Chr.11: 44095997-44095997 on Build GRCh38
rs189113630 Chr.11: 44095673-44095673 on Build GRCh38
rs569630494 Chr.11: 44095576-44095576 on Build GRCh38
rs529057271 Chr.11: 44095860-44095860 on Build GRCh38
rs993868292 Chr.11: 44095705-44095705 on Build GRCh38

View Full Product Details