Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49869736-49870233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM2
Entrez Gene ID
23017
Gene Name
Fas apoptotic inhibitory molecule 2
Gene Aliases
LFG, LFG2, NGP35, NMP35, TMBIM2
UniGene
Hs.567424
Gene Chromosome Location
Chr.12: 49866896-49904275 on Build GRCh38
Transcripts
NM_012306.3 XM_005268730.3 XM_017019040.1

SNP Detail

SNP ID Chromosome Location
rs964200368 Chr.12: 49869878-49869878 on Build GRCh38
rs893759620 Chr.12: 49869980-49869980 on Build GRCh38
rs575669812 Chr.12: 49869804-49869804 on Build GRCh38
rs910973606 Chr.12: 49869791-49869791 on Build GRCh38
rs930221727 Chr.12: 49869848-49869848 on Build GRCh38
rs531577656 Chr.12: 49869882-49869882 on Build GRCh38
rs941639463 Chr.12: 49870100-49870100 on Build GRCh38
rs936465004 Chr.12: 49870006-49870006 on Build GRCh38
rs560309798 Chr.12: 49870093-49870093 on Build GRCh38
rs1018364220 Chr.12: 49869877-49869877 on Build GRCh38
rs1020275376 Chr.12: 49869833-49869833 on Build GRCh38
rs963802537 Chr.12: 49870009-49870009 on Build GRCh38
rs982446808 Chr.12: 49870074-49870074 on Build GRCh38
rs555019236 Chr.12: 49869945-49869945 on Build GRCh38
rs376264571 Chr.12: 49870003-49870003 on Build GRCh38
rs574821448 Chr.12: 49870033-49870033 on Build GRCh38
rs1027526689 Chr.12: 49870182-49870182 on Build GRCh38
rs866714234 Chr.12: 49869868-49869868 on Build GRCh38
rs976928007 Chr.12: 49870017-49870017 on Build GRCh38
rs995621889 Chr.12: 49870171-49870171 on Build GRCh38
rs1054002533 Chr.12: 49870023-49870023 on Build GRCh38
rs1044747217 Chr.12: 49869982-49869982 on Build GRCh38
rs917548113 Chr.12: 49869966-49869966 on Build GRCh38
rs1046233922 Chr.12: 49870195-49870195 on Build GRCh38
rs762802127 Chr.12: 49869964-49869964 on Build GRCh38
rs890412691 Chr.12: 49870102-49870102 on Build GRCh38
rs892319474 Chr.12: 49870045-49870045 on Build GRCh38
rs1008229987 Chr.12: 49870137-49870137 on Build GRCh38
rs577157226 Chr.12: 49870101-49870101 on Build GRCh38
rs540849571 Chr.12: 49870053-49870053 on Build GRCh38
rs763978713 Chr.12: 49869985-49869985 on Build GRCh38
rs910132598 Chr.12: 49870075-49870076 on Build GRCh38
rs763520401 Chr.12: 49869921-49869921 on Build GRCh38
rs930195448 Chr.12: 49869939-49869939 on Build GRCh38
rs1018299646 Chr.12: 49870146-49870146 on Build GRCh38
rs537956783 Chr.12: 49869826-49869826 on Build GRCh38
rs976433555 Chr.12: 49869800-49869800 on Build GRCh38
rs894541541 Chr.12: 49869906-49869907 on Build GRCh38
rs1009450308 Chr.12: 49870046-49870046 on Build GRCh38
rs1029292407 Chr.12: 49870041-49870041 on Build GRCh38
rs964446664 Chr.12: 49870153-49870153 on Build GRCh38
rs949046264 Chr.12: 49869979-49869979 on Build GRCh38
rs1051690452 Chr.12: 49870078-49870078 on Build GRCh38
rs752502964 Chr.12: 49869940-49869940 on Build GRCh38
rs989193765 Chr.12: 49869756-49869756 on Build GRCh38
rs974634289 Chr.12: 49869895-49869895 on Build GRCh38
rs904920590 Chr.12: 49869983-49869983 on Build GRCh38
rs71464978 Chr.12: 49869818-49869818 on Build GRCh38
rs986849290 Chr.12: 49869875-49869875 on Build GRCh38
rs955361597 Chr.12: 49869846-49869846 on Build GRCh38
rs1045702766 Chr.12: 49869927-49869927 on Build GRCh38
rs892366198 Chr.12: 49869797-49869797 on Build GRCh38
rs950138781 Chr.12: 49870175-49870175 on Build GRCh38

View Full Product Details