Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 93718296-93718832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM92A1
Entrez Gene ID
137392
Gene Name
family with sequence similarity 92 member A1
Gene Aliases
-
UniGene
Hs.125038
Gene Chromosome Location
Chr.8: 93700252-93732041 on Build GRCh38
Transcripts
NM_001283034.1 NM_145269.4 NR_104267.1 NR_104268.1 XM_005250787.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644201 c.639T>G p.D213E Chr.8: 93718770-93718770 on Build GRCh38
5358610 c.544-10delT p.? Chr.8: 93718665-93718665 on Build GRCh38
1206483 c.571G>A p.E191K Chr.8: 93718702-93718702 on Build GRCh38
1635909 c.593C>T p.A198V Chr.8: 93718724-93718724 on Build GRCh38
160977 c.608C>A p.T203N Chr.8: 93718739-93718739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780541574 Chr.8: 93718664-93718665 on Build GRCh38
rs867090153 Chr.8: 93718718-93718718 on Build GRCh38
rs1008464011 Chr.8: 93718433-93718437 on Build GRCh38
rs201261789 Chr.8: 93718737-93718737 on Build GRCh38
rs557237603 Chr.8: 93718747-93718747 on Build GRCh38
rs962902378 Chr.8: 93718349-93718349 on Build GRCh38
rs569523992 Chr.8: 93718416-93718416 on Build GRCh38
rs539958551 Chr.8: 93718665-93718665 on Build GRCh38
rs947248825 Chr.8: 93718555-93718555 on Build GRCh38
rs534808639 Chr.8: 93718599-93718599 on Build GRCh38
rs774717777 Chr.8: 93718700-93718700 on Build GRCh38
rs765355183 Chr.8: 93718547-93718547 on Build GRCh38
rs898669249 Chr.8: 93718764-93718764 on Build GRCh38
rs1041362581 Chr.8: 93718452-93718452 on Build GRCh38
rs750518677 Chr.8: 93718716-93718716 on Build GRCh38
rs771711587 Chr.8: 93718800-93718803 on Build GRCh38
rs770065976 Chr.8: 93718647-93718651 on Build GRCh38
rs188187666 Chr.8: 93718432-93718432 on Build GRCh38
rs994355711 Chr.8: 93718493-93718493 on Build GRCh38
rs778224531 Chr.8: 93718656-93718656 on Build GRCh38
rs1051892824 Chr.8: 93718678-93718678 on Build GRCh38
rs771474777 Chr.8: 93718694-93718694 on Build GRCh38
rs761445666 Chr.8: 93718755-93718755 on Build GRCh38
rs147773881 Chr.8: 93718408-93718408 on Build GRCh38
rs914329580 Chr.8: 93718338-93718338 on Build GRCh38
rs774049779 Chr.8: 93718739-93718740 on Build GRCh38
rs200324656 Chr.8: 93718664-93718664 on Build GRCh38
rs768264071 Chr.8: 93718672-93718673 on Build GRCh38
rs952846802 Chr.8: 93718549-93718549 on Build GRCh38
rs368183102 Chr.8: 93718803-93718803 on Build GRCh38
rs1035765506 Chr.8: 93718439-93718439 on Build GRCh38
rs1027446632 Chr.8: 93718521-93718521 on Build GRCh38
rs754435008 Chr.8: 93718393-93718395 on Build GRCh38
rs754674520 Chr.8: 93718805-93718805 on Build GRCh38
rs759670572 Chr.8: 93718701-93718701 on Build GRCh38
rs772117314 Chr.8: 93718705-93718705 on Build GRCh38
rs914417150 Chr.8: 93718539-93718539 on Build GRCh38
rs374300628 Chr.8: 93718717-93718717 on Build GRCh38
rs995720847 Chr.8: 93718403-93718403 on Build GRCh38
rs544038159 Chr.8: 93718388-93718392 on Build GRCh38
rs537313242 Chr.8: 93718732-93718732 on Build GRCh38
rs947143235 Chr.8: 93718738-93718738 on Build GRCh38
rs577151758 Chr.8: 93718770-93718770 on Build GRCh38
rs546206142 Chr.8: 93718795-93718795 on Build GRCh38
rs1039811345 Chr.8: 93718725-93718725 on Build GRCh38
rs554764017 Chr.8: 93718619-93718619 on Build GRCh38
rs951409060 Chr.8: 93718412-93718412 on Build GRCh38
rs565796478 Chr.8: 93718556-93718556 on Build GRCh38
rs551972713 Chr.8: 93718458-93718458 on Build GRCh38
rs761444535 Chr.8: 93718798-93718801 on Build GRCh38
rs995670401 Chr.8: 93718777-93718777 on Build GRCh38
rs758026049 Chr.8: 93718780-93718780 on Build GRCh38
rs1012633854 Chr.8: 93718610-93718610 on Build GRCh38
rs959066362 Chr.8: 93718453-93718453 on Build GRCh38
rs955833002 Chr.8: 93718708-93718708 on Build GRCh38
rs977334661 Chr.8: 93718615-93718615 on Build GRCh38
rs749874325 Chr.8: 93718665-93718665 on Build GRCh38
rs979318646 Chr.8: 93718425-93718425 on Build GRCh38
rs765281776 Chr.8: 93718768-93718768 on Build GRCh38

View Full Product Details