Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53810129-53810656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTO
Entrez Gene ID
79068
Gene Name
fat mass and obesity associated
Gene Aliases
ALKBH9, BMIQ14, GDFD
UniGene
Hs.528833
Gene Chromosome Location
Chr.16: 53703963-54114467 on Build GRCh38
Transcripts
NM_001080432.2 XM_011523313.2 XM_011523314.2 XM_011523315.2 XM_011523316.2 XM_017023654.1 XM_017023655.1 XM_017023656.1 XM_017023657.1 XM_017023658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1378351 c.110A>C p.E37A Chr.16: 53810204-53810204 on Build GRCh38
6324273 c.91C>T p.L31L Chr.16: 53810185-53810185 on Build GRCh38
703447 c.84C>T p.L28L Chr.16: 53810178-53810178 on Build GRCh38
4972976 c.118C>G p.Q40E Chr.16: 53810212-53810212 on Build GRCh38
971509 c.123G>T p.Q41H Chr.16: 53810217-53810217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045847740 Chr.16: 53810484-53810484 on Build GRCh38
rs762149812 Chr.16: 53810154-53810154 on Build GRCh38
rs1048016586 Chr.16: 53810278-53810278 on Build GRCh38
rs201041270 Chr.16: 53810222-53810222 on Build GRCh38
rs542708952 Chr.16: 53810369-53810369 on Build GRCh38
rs886769563 Chr.16: 53810338-53810338 on Build GRCh38
rs555947573 Chr.16: 53810435-53810435 on Build GRCh38
rs768461316 Chr.16: 53810223-53810223 on Build GRCh38
rs897168379 Chr.16: 53810436-53810436 on Build GRCh38
rs760938787 Chr.16: 53810253-53810253 on Build GRCh38
rs922392817 Chr.16: 53810341-53810341 on Build GRCh38
rs966397356 Chr.16: 53810252-53810252 on Build GRCh38
rs995135852 Chr.16: 53810277-53810277 on Build GRCh38
rs956711560 Chr.16: 53810413-53810413 on Build GRCh38
rs573824091 Chr.16: 53810349-53810349 on Build GRCh38
rs113050836 Chr.16: 53810588-53810588 on Build GRCh38
rs979674979 Chr.16: 53810396-53810396 on Build GRCh38
rs756946176 Chr.16: 53810203-53810203 on Build GRCh38
rs772784163 Chr.16: 53810560-53810560 on Build GRCh38
rs193197607 Chr.16: 53810231-53810231 on Build GRCh38
rs373028121 Chr.16: 53810188-53810188 on Build GRCh38
rs756522457 Chr.16: 53810171-53810171 on Build GRCh38
rs943886331 Chr.16: 53810522-53810522 on Build GRCh38
rs1035432522 Chr.16: 53810356-53810356 on Build GRCh38
rs1048282439 Chr.16: 53810596-53810596 on Build GRCh38
rs932479362 Chr.16: 53810354-53810354 on Build GRCh38
rs200145312 Chr.16: 53810235-53810235 on Build GRCh38
rs750855307 Chr.16: 53810169-53810169 on Build GRCh38
rs565154088 Chr.16: 53810533-53810533 on Build GRCh38
rs112339041 Chr.16: 53810254-53810254 on Build GRCh38
rs7499606 Chr.16: 53810202-53810202 on Build GRCh38
rs780803760 Chr.16: 53810208-53810208 on Build GRCh38
rs755576124 Chr.16: 53810269-53810269 on Build GRCh38
rs775203421 Chr.16: 53810220-53810220 on Build GRCh38
rs754399299 Chr.16: 53810178-53810178 on Build GRCh38
rs141626455 Chr.16: 53810581-53810581 on Build GRCh38
rs1004393671 Chr.16: 53810350-53810350 on Build GRCh38
rs961174882 Chr.16: 53810373-53810373 on Build GRCh38
rs1001699708 Chr.16: 53810561-53810561 on Build GRCh38
rs563523575 Chr.16: 53810313-53810313 on Build GRCh38
rs969595862 Chr.16: 53810390-53810390 on Build GRCh38
rs886803420 Chr.16: 53810615-53810615 on Build GRCh38
rs1057303843 Chr.16: 53810619-53810619 on Build GRCh38
rs905982689 Chr.16: 53810512-53810512 on Build GRCh38
rs766854982 Chr.16: 53810261-53810261 on Build GRCh38
rs970217524 Chr.16: 53810172-53810172 on Build GRCh38
rs115090477 Chr.16: 53810462-53810462 on Build GRCh38
rs890975683 Chr.16: 53810190-53810190 on Build GRCh38
rs769612021 Chr.16: 53810210-53810210 on Build GRCh38
rs572790599 Chr.16: 53810168-53810168 on Build GRCh38
rs773590064 Chr.16: 53810251-53810251 on Build GRCh38
rs538745058 Chr.16: 53810411-53810411 on Build GRCh38
rs754140018 Chr.16: 53810267-53810267 on Build GRCh38
rs1023897264 Chr.16: 53810383-53810383 on Build GRCh38
rs149393601 Chr.16: 53810176-53810176 on Build GRCh38
rs550825039 Chr.16: 53810589-53810589 on Build GRCh38
rs553660653 Chr.16: 53810616-53810616 on Build GRCh38
rs1115005 Chr.16: 53810382-53810382 on Build GRCh38
rs762316478 Chr.16: 53810592-53810592 on Build GRCh38
rs774554017 Chr.16: 53810225-53810225 on Build GRCh38
rs116753298 Chr.16: 53810193-53810193 on Build GRCh38

View Full Product Details