Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92246719-92247281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLMN
Entrez Gene ID
11146
Gene Name
glomulin, FKBP associated protein
Gene Aliases
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM
UniGene
Hs.49105
Gene Chromosome Location
Chr.1: 92245786-92441936 on Build GRCh38
Transcripts
NM_001319683.1 NM_053274.2 NR_135089.1 XM_005270401.3 XM_006710309.2 XM_011540546.2 XM_017000137.1 XM_017000138.1 XM_017000139.1 XM_017000140.1 XM_017000141.1 XM_017000142.1 XM_017000143.1 XM_017000144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738452 c.1592A>G p.Q531R Chr.1: 92247138-92247138 on Build GRCh38
7075218 c.1628G>C p.G543A Chr.1: 92247102-92247102 on Build GRCh38
6671669 c.1662G>A p.Q554Q Chr.1: 92247068-92247068 on Build GRCh38
7251243 c.1586-3C>A p.? Chr.1: 92247147-92247147 on Build GRCh38
3493354 c.1652C>T p.P551L Chr.1: 92247078-92247078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563113276 Chr.1: 92246888-92246888 on Build GRCh38
rs112077260 Chr.1: 92247043-92247043 on Build GRCh38
rs949767297 Chr.1: 92246855-92246855 on Build GRCh38
rs758397101 Chr.1: 92247087-92247087 on Build GRCh38
rs753584301 Chr.1: 92247197-92247198 on Build GRCh38
rs962524609 Chr.1: 92246831-92246831 on Build GRCh38
rs773455783 Chr.1: 92247033-92247037 on Build GRCh38
rs35752451 Chr.1: 92247042-92247043 on Build GRCh38
rs998600964 Chr.1: 92246981-92246981 on Build GRCh38
rs951461003 Chr.1: 92246759-92246759 on Build GRCh38
rs750386801 Chr.1: 92247051-92247051 on Build GRCh38
rs560509557 Chr.1: 92246951-92246951 on Build GRCh38
rs902375041 Chr.1: 92246896-92246896 on Build GRCh38
rs902406104 Chr.1: 92247044-92247046 on Build GRCh38
rs112320006 Chr.1: 92247041-92247041 on Build GRCh38
rs1003356954 Chr.1: 92246897-92246897 on Build GRCh38
rs535875892 Chr.1: 92247221-92247221 on Build GRCh38
rs375995385 Chr.1: 92247023-92247023 on Build GRCh38
rs771987072 Chr.1: 92246856-92246856 on Build GRCh38
rs760218331 Chr.1: 92247152-92247152 on Build GRCh38
rs986900864 Chr.1: 92247014-92247014 on Build GRCh38
rs549417197 Chr.1: 92246966-92246966 on Build GRCh38
rs763871494 Chr.1: 92247017-92247017 on Build GRCh38
rs748327333 Chr.1: 92247077-92247077 on Build GRCh38
rs758508113 Chr.1: 92247170-92247170 on Build GRCh38
rs764094882 Chr.1: 92246988-92246988 on Build GRCh38
rs372027596 Chr.1: 92247027-92247027 on Build GRCh38
rs1054127279 Chr.1: 92246923-92246923 on Build GRCh38
rs991427988 Chr.1: 92246832-92246832 on Build GRCh38
rs770002756 Chr.1: 92247031-92247033 on Build GRCh38
rs371260645 Chr.1: 92246815-92246815 on Build GRCh38
rs1020874168 Chr.1: 92246895-92246895 on Build GRCh38
rs761930416 Chr.1: 92247025-92247027 on Build GRCh38
rs757602772 Chr.1: 92247192-92247192 on Build GRCh38
rs959415071 Chr.1: 92246915-92246915 on Build GRCh38
rs538898085 Chr.1: 92247115-92247115 on Build GRCh38
rs900051741 Chr.1: 92246972-92246972 on Build GRCh38
rs1038666830 Chr.1: 92246848-92246848 on Build GRCh38
rs777958203 Chr.1: 92247181-92247181 on Build GRCh38
rs1049445368 Chr.1: 92246780-92246780 on Build GRCh38
rs111287047 Chr.1: 92247042-92247042 on Build GRCh38
rs397737127 Chr.1: 92247044-92247045 on Build GRCh38
rs191480502 Chr.1: 92247091-92247091 on Build GRCh38
rs915808131 Chr.1: 92246850-92246850 on Build GRCh38
rs763265202 Chr.1: 92247040-92247041 on Build GRCh38
rs527699482 Chr.1: 92246956-92246956 on Build GRCh38
rs909755754 Chr.1: 92246826-92246826 on Build GRCh38
rs544813739 Chr.1: 92246857-92246857 on Build GRCh38
rs113775598 Chr.1: 92246945-92246945 on Build GRCh38
rs763505318 Chr.1: 92247179-92247180 on Build GRCh38
rs778068890 Chr.1: 92247089-92247089 on Build GRCh38
rs747255879 Chr.1: 92247185-92247185 on Build GRCh38
rs533659666 Chr.1: 92246916-92246916 on Build GRCh38
rs779825891 Chr.1: 92247065-92247065 on Build GRCh38
rs531732627 Chr.1: 92246975-92246975 on Build GRCh38
rs1013984553 Chr.1: 92247028-92247043 on Build GRCh38
rs746209178 Chr.1: 92247096-92247096 on Build GRCh38
rs756096722 Chr.1: 92247056-92247056 on Build GRCh38
rs1030172890 Chr.1: 92246743-92246743 on Build GRCh38
rs752038749 Chr.1: 92247150-92247151 on Build GRCh38
rs753662150 Chr.1: 92247165-92247165 on Build GRCh38
rs763449668 Chr.1: 92247120-92247120 on Build GRCh38
rs762334917 Chr.1: 92247015-92247015 on Build GRCh38
rs761409643 Chr.1: 92247130-92247130 on Build GRCh38
rs895223354 Chr.1: 92247225-92247225 on Build GRCh38
rs374721551 Chr.1: 92247142-92247142 on Build GRCh38
rs376960198 Chr.1: 92247095-92247095 on Build GRCh38
rs991293556 Chr.1: 92247092-92247092 on Build GRCh38
rs775854682 Chr.1: 92247106-92247106 on Build GRCh38
rs866275486 Chr.1: 92247117-92247117 on Build GRCh38
rs774026444 Chr.1: 92247126-92247126 on Build GRCh38
rs188290801 Chr.1: 92246849-92246849 on Build GRCh38
rs746190395 Chr.1: 92246940-92246940 on Build GRCh38
rs755857783 Chr.1: 92247163-92247163 on Build GRCh38
rs553799640 Chr.1: 92247156-92247156 on Build GRCh38
rs747869141 Chr.1: 92246854-92246854 on Build GRCh38
rs772773828 Chr.1: 92246998-92246998 on Build GRCh38
rs554717080 Chr.1: 92246838-92246838 on Build GRCh38
rs551064579 Chr.1: 92247023-92247036 on Build GRCh38
rs1003387970 Chr.1: 92247063-92247063 on Build GRCh38
rs368746622 Chr.1: 92247098-92247098 on Build GRCh38
rs755678026 Chr.1: 92246912-92246912 on Build GRCh38
rs934317812 Chr.1: 92247224-92247224 on Build GRCh38
rs200389652 Chr.1: 92247138-92247138 on Build GRCh38
rs567424928 Chr.1: 92246969-92246969 on Build GRCh38
rs372064317 Chr.1: 92247006-92247006 on Build GRCh38
rs781529909 Chr.1: 92247193-92247193 on Build GRCh38
rs141142924 Chr.1: 92247201-92247201 on Build GRCh38

View Full Product Details