Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52147386-52147942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB5
Entrez Gene ID
10681
Gene Name
G protein subunit beta 5
Gene Aliases
GB5
UniGene
Hs.155090
Gene Chromosome Location
Chr.15: 52120926-52191368 on Build GRCh38
Transcripts
NM_006578.3 NM_016194.3 XM_011521162.2 XM_011521163.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2217280 c.425C>T p.A142V Chr.15: 52147528-52147528 on Build GRCh38
3502135 c.473C>T p.S158L Chr.15: 52147480-52147480 on Build GRCh38
962795 c.454G>A p.A152T Chr.15: 52147499-52147499 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs553676677 Chr.15: 52147739-52147739 on Build GRCh38
rs766352486 Chr.15: 52147583-52147583 on Build GRCh38
rs756243496 Chr.15: 52147732-52147732 on Build GRCh38
rs1017220418 Chr.15: 52147459-52147460 on Build GRCh38
rs757984625 Chr.15: 52147476-52147476 on Build GRCh38
rs540650092 Chr.15: 52147699-52147699 on Build GRCh38
rs199692679 Chr.15: 52147551-52147551 on Build GRCh38
rs775843261 Chr.15: 52147530-52147530 on Build GRCh38
rs377518088 Chr.15: 52147450-52147450 on Build GRCh38
rs934515272 Chr.15: 52147623-52147623 on Build GRCh38
rs955979955 Chr.15: 52147911-52147911 on Build GRCh38
rs900186115 Chr.15: 52147761-52147761 on Build GRCh38
rs751616514 Chr.15: 52147582-52147583 on Build GRCh38
rs960833298 Chr.15: 52147846-52147846 on Build GRCh38
rs1052876347 Chr.15: 52147679-52147679 on Build GRCh38
rs370908779 Chr.15: 52147524-52147524 on Build GRCh38
rs1005671912 Chr.15: 52147465-52147465 on Build GRCh38
rs757452139 Chr.15: 52147583-52147586 on Build GRCh38
rs768134904 Chr.15: 52147486-52147486 on Build GRCh38
rs368969434 Chr.15: 52147529-52147529 on Build GRCh38
rs1012351651 Chr.15: 52147853-52147853 on Build GRCh38
rs762855246 Chr.15: 52147581-52147593 on Build GRCh38
rs770919050 Chr.15: 52147528-52147528 on Build GRCh38
rs910305762 Chr.15: 52147562-52147562 on Build GRCh38
rs757822736 Chr.15: 52147431-52147431 on Build GRCh38
rs146955118 Chr.15: 52147479-52147479 on Build GRCh38
rs1047674483 Chr.15: 52147592-52147592 on Build GRCh38
rs879238669 Chr.15: 52147595-52147595 on Build GRCh38
rs923137525 Chr.15: 52147859-52147859 on Build GRCh38
rs545110330 Chr.15: 52147877-52147877 on Build GRCh38
rs568961518 Chr.15: 52147558-52147558 on Build GRCh38
rs769366821 Chr.15: 52147551-52147555 on Build GRCh38
rs868020290 Chr.15: 52147513-52147513 on Build GRCh38
rs887660348 Chr.15: 52147601-52147601 on Build GRCh38
rs754276744 Chr.15: 52147419-52147419 on Build GRCh38
rs374968711 Chr.15: 52147439-52147439 on Build GRCh38
rs946708368 Chr.15: 52147591-52147591 on Build GRCh38
rs749100407 Chr.15: 52147527-52147527 on Build GRCh38
rs764293568 Chr.15: 52147416-52147416 on Build GRCh38
rs183997548 Chr.15: 52147577-52147577 on Build GRCh38
rs975820386 Chr.15: 52147833-52147833 on Build GRCh38
rs555345415 Chr.15: 52147738-52147738 on Build GRCh38
rs968474422 Chr.15: 52147869-52147869 on Build GRCh38
rs925893182 Chr.15: 52147897-52147897 on Build GRCh38
rs941937264 Chr.15: 52147914-52147914 on Build GRCh38
rs9972619 Chr.15: 52147811-52147811 on Build GRCh38
rs191832850 Chr.15: 52147802-52147802 on Build GRCh38
rs929124182 Chr.15: 52147867-52147867 on Build GRCh38
rs766785026 Chr.15: 52147572-52147572 on Build GRCh38
rs900137380 Chr.15: 52147472-52147472 on Build GRCh38
rs189136430 Chr.15: 52147679-52147679 on Build GRCh38
rs147993382 Chr.15: 52147509-52147509 on Build GRCh38
rs368749869 Chr.15: 52147861-52147861 on Build GRCh38
rs887706010 Chr.15: 52147900-52147900 on Build GRCh38
rs979441449 Chr.15: 52147884-52147884 on Build GRCh38
rs533204386 Chr.15: 52147829-52147829 on Build GRCh38
rs761569770 Chr.15: 52147437-52147451 on Build GRCh38
rs35709612 Chr.15: 52147743-52147743 on Build GRCh38
rs773096877 Chr.15: 52147556-52147557 on Build GRCh38
rs1016908753 Chr.15: 52147726-52147726 on Build GRCh38
rs977684566 Chr.15: 52147784-52147784 on Build GRCh38
rs1047559481 Chr.15: 52147876-52147876 on Build GRCh38
rs910201533 Chr.15: 52147812-52147812 on Build GRCh38
rs753893636 Chr.15: 52147583-52147587 on Build GRCh38
rs370348343 Chr.15: 52147436-52147436 on Build GRCh38
rs943109622 Chr.15: 52147563-52147563 on Build GRCh38
rs780797393 Chr.15: 52147492-52147492 on Build GRCh38
rs544030481 Chr.15: 52147779-52147779 on Build GRCh38
rs569996082 Chr.15: 52147859-52147859 on Build GRCh38
rs925872540 Chr.15: 52147617-52147617 on Build GRCh38
rs971530935 Chr.15: 52147782-52147782 on Build GRCh38
rs760659382 Chr.15: 52147583-52147584 on Build GRCh38
rs979104137 Chr.15: 52147595-52147596 on Build GRCh38
rs943140814 Chr.15: 52147825-52147825 on Build GRCh38
rs765783223 Chr.15: 52147559-52147559 on Build GRCh38
rs747999526 Chr.15: 52147493-52147493 on Build GRCh38
rs190432484 Chr.15: 52147482-52147482 on Build GRCh38
rs145489349 Chr.15: 52147552-52147552 on Build GRCh38
rs1008551552 Chr.15: 52147440-52147440 on Build GRCh38
rs750819330 Chr.15: 52147461-52147461 on Build GRCh38
rs957611290 Chr.15: 52147803-52147803 on Build GRCh38
rs750714965 Chr.15: 52147565-52147565 on Build GRCh38
rs757326706 Chr.15: 52147823-52147823 on Build GRCh38
rs372223105 Chr.15: 52147567-52147567 on Build GRCh38
rs754692858 Chr.15: 52147578-52147578 on Build GRCh38

View Full Product Details