Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30678705-30679250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSR
Entrez Gene ID
2936
Gene Name
glutathione-disulfide reductase
Gene Aliases
HEL-75, HEL-S-122m
UniGene
Hs.271510
Gene Chromosome Location
Chr.8: 30678061-30727969 on Build GRCh38
Transcripts
NM_000637.3 NM_001195102.1 NM_001195103.1 NM_001195104.1

SNP Detail

SNP ID Chromosome Location
rs1138054 Chr.8: 30679064-30679064 on Build GRCh38
rs758775349 Chr.8: 30678876-30678876 on Build GRCh38
rs2551734 Chr.8: 30678911-30678911 on Build GRCh38
rs572203293 Chr.8: 30678830-30678830 on Build GRCh38
rs961100745 Chr.8: 30678925-30678925 on Build GRCh38
rs915257232 Chr.8: 30679111-30679111 on Build GRCh38
rs747535348 Chr.8: 30678984-30678984 on Build GRCh38
rs35555269 Chr.8: 30679150-30679151 on Build GRCh38
rs1040997301 Chr.8: 30678805-30678805 on Build GRCh38
rs182562887 Chr.8: 30678910-30678910 on Build GRCh38
rs534116515 Chr.8: 30678978-30678978 on Build GRCh38
rs2551735 Chr.8: 30678908-30678908 on Build GRCh38
rs1049920224 Chr.8: 30678807-30678807 on Build GRCh38
rs935350272 Chr.8: 30678816-30678816 on Build GRCh38
rs1045142048 Chr.8: 30679214-30679214 on Build GRCh38
rs2551733 Chr.8: 30679077-30679077 on Build GRCh38
rs1013358894 Chr.8: 30679104-30679104 on Build GRCh38
rs923830521 Chr.8: 30678806-30678806 on Build GRCh38
rs2551736 Chr.8: 30678828-30678828 on Build GRCh38
rs944119784 Chr.8: 30678986-30678986 on Build GRCh38
rs995344564 Chr.8: 30679170-30679170 on Build GRCh38
rs1033434143 Chr.8: 30678902-30678902 on Build GRCh38
rs1024662150 Chr.8: 30678918-30678918 on Build GRCh38
rs948028340 Chr.8: 30679203-30679203 on Build GRCh38
rs962571092 Chr.8: 30678952-30678952 on Build GRCh38
rs71823444 Chr.8: 30679150-30679150 on Build GRCh38
rs913296522 Chr.8: 30678997-30678997 on Build GRCh38
rs985205540 Chr.8: 30678743-30678743 on Build GRCh38
rs146973669 Chr.8: 30678995-30678995 on Build GRCh38
rs911366325 Chr.8: 30678981-30678981 on Build GRCh38
rs1024759364 Chr.8: 30679192-30679196 on Build GRCh38
rs903772302 Chr.8: 30679014-30679014 on Build GRCh38
rs1000692938 Chr.8: 30678875-30678875 on Build GRCh38
rs972803607 Chr.8: 30678938-30678938 on Build GRCh38
rs568218175 Chr.8: 30678924-30678924 on Build GRCh38
rs1053690295 Chr.8: 30679107-30679107 on Build GRCh38
rs946104503 Chr.8: 30679008-30679008 on Build GRCh38
rs1010633614 Chr.8: 30678853-30678853 on Build GRCh38
rs531396090 Chr.8: 30678912-30678912 on Build GRCh38
rs1054897479 Chr.8: 30679057-30679057 on Build GRCh38
rs976954120 Chr.8: 30679019-30679019 on Build GRCh38
rs370089677 Chr.8: 30678870-30678870 on Build GRCh38
rs756558112 Chr.8: 30679020-30679020 on Build GRCh38
rs910945348 Chr.8: 30679007-30679007 on Build GRCh38
rs1000704509 Chr.8: 30679030-30679030 on Build GRCh38
rs552233138 Chr.8: 30678887-30678887 on Build GRCh38
rs8191041 Chr.8: 30679011-30679011 on Build GRCh38
rs570052585 Chr.8: 30678916-30678916 on Build GRCh38
rs576245359 Chr.8: 30679118-30679118 on Build GRCh38
rs532433375 Chr.8: 30678823-30678823 on Build GRCh38
rs796525475 Chr.8: 30678864-30678864 on Build GRCh38
rs2551737 Chr.8: 30678744-30678744 on Build GRCh38
rs10216724 Chr.8: 30678920-30678920 on Build GRCh38
rs931534610 Chr.8: 30678798-30678798 on Build GRCh38
rs776626325 Chr.8: 30679225-30679225 on Build GRCh38
rs943841694 Chr.8: 30678779-30678779 on Build GRCh38
rs553455623 Chr.8: 30678980-30678980 on Build GRCh38
rs11552962 Chr.8: 30678963-30678963 on Build GRCh38
rs397893015 Chr.8: 30679168-30679169 on Build GRCh38
rs747732351 Chr.8: 30678834-30678834 on Build GRCh38
rs911084307 Chr.8: 30678748-30678748 on Build GRCh38
rs376725408 Chr.8: 30678826-30678826 on Build GRCh38

View Full Product Details