Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100049905-100050460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD14A
Entrez Gene ID
64645
Gene Name
major facilitator superfamily domain containing 14A
Gene Aliases
HIAT1
UniGene
Hs.124156
Gene Chromosome Location
Chr.1: 100038078-100083377 on Build GRCh38
Transcripts
NM_033055.2 XM_005271118.1 XM_011541957.1 XM_017002084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6679794 c.166G>A p.A56T Chr.1: 100049990-100049990 on Build GRCh38
4019783 c.113A>G p.Y38C Chr.1: 100049937-100049937 on Build GRCh38
6679792 c.126C>T p.I42I Chr.1: 100049950-100049950 on Build GRCh38
892916 c.101C>A p.S34Y Chr.1: 100049925-100049925 on Build GRCh38
6679789 c.127G>A p.V43I Chr.1: 100049951-100049951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977641495 Chr.1: 100049948-100049948 on Build GRCh38
rs535096988 Chr.1: 100050086-100050086 on Build GRCh38
rs373404943 Chr.1: 100049974-100049974 on Build GRCh38
rs748084416 Chr.1: 100049983-100049983 on Build GRCh38
rs947817336 Chr.1: 100050436-100050436 on Build GRCh38
rs1049955981 Chr.1: 100050426-100050426 on Build GRCh38
rs955041602 Chr.1: 100050175-100050175 on Build GRCh38
rs189585993 Chr.1: 100050428-100050428 on Build GRCh38
rs559784115 Chr.1: 100050289-100050289 on Build GRCh38
rs1021648218 Chr.1: 100050292-100050292 on Build GRCh38
rs570280150 Chr.1: 100049951-100049951 on Build GRCh38
rs376451133 Chr.1: 100050056-100050056 on Build GRCh38
rs969867944 Chr.1: 100050197-100050197 on Build GRCh38
rs867177262 Chr.1: 100050252-100050252 on Build GRCh38
rs530929530 Chr.1: 100050399-100050399 on Build GRCh38
rs1028510909 Chr.1: 100050175-100050175 on Build GRCh38
rs924844750 Chr.1: 100049949-100049949 on Build GRCh38
rs776633185 Chr.1: 100050221-100050221 on Build GRCh38
rs765112348 Chr.1: 100050341-100050341 on Build GRCh38
rs763766183 Chr.1: 100050185-100050186 on Build GRCh38
rs772784750 Chr.1: 100050045-100050047 on Build GRCh38
rs754847168 Chr.1: 100049965-100049965 on Build GRCh38
rs939361636 Chr.1: 100050077-100050077 on Build GRCh38
rs947636721 Chr.1: 100049944-100049944 on Build GRCh38
rs575431435 Chr.1: 100050106-100050106 on Build GRCh38
rs1008516090 Chr.1: 100050188-100050188 on Build GRCh38
rs1019434671 Chr.1: 100050286-100050286 on Build GRCh38
rs17121792 Chr.1: 100049941-100049941 on Build GRCh38
rs537904803 Chr.1: 100050109-100050109 on Build GRCh38
rs41286540 Chr.1: 100049981-100049981 on Build GRCh38
rs898671046 Chr.1: 100050122-100050122 on Build GRCh38
rs557621934 Chr.1: 100050130-100050130 on Build GRCh38
rs114029430 Chr.1: 100050060-100050060 on Build GRCh38
rs115052076 Chr.1: 100050287-100050287 on Build GRCh38
rs7532378 Chr.1: 100050094-100050094 on Build GRCh38
rs561720768 Chr.1: 100050393-100050393 on Build GRCh38
rs762713771 Chr.1: 100050418-100050418 on Build GRCh38
rs1008297429 Chr.1: 100050181-100050181 on Build GRCh38
rs748296147 Chr.1: 100050073-100050073 on Build GRCh38
rs746089322 Chr.1: 100049998-100049998 on Build GRCh38
rs1056496745 Chr.1: 100050082-100050083 on Build GRCh38
rs773053573 Chr.1: 100049997-100049997 on Build GRCh38
rs184051823 Chr.1: 100050329-100050329 on Build GRCh38
rs748868845 Chr.1: 100049964-100049964 on Build GRCh38
rs946796070 Chr.1: 100050046-100050048 on Build GRCh38
rs924898197 Chr.1: 100050360-100050360 on Build GRCh38
rs866296749 Chr.1: 100050196-100050196 on Build GRCh38
rs968736058 Chr.1: 100050339-100050339 on Build GRCh38
rs771796811 Chr.1: 100049994-100049994 on Build GRCh38
rs770098812 Chr.1: 100050012-100050012 on Build GRCh38
rs755852549 Chr.1: 100049950-100049950 on Build GRCh38
rs762748225 Chr.1: 100050045-100050046 on Build GRCh38
rs775413240 Chr.1: 100050033-100050033 on Build GRCh38
rs1027956482 Chr.1: 100049935-100049935 on Build GRCh38
rs1031360771 Chr.1: 100050187-100050187 on Build GRCh38
rs1025543167 Chr.1: 100050168-100050168 on Build GRCh38
rs546556817 Chr.1: 100050043-100050043 on Build GRCh38
rs867662808 Chr.1: 100050313-100050313 on Build GRCh38
rs762610116 Chr.1: 100050097-100050098 on Build GRCh38
rs887010610 Chr.1: 100050170-100050170 on Build GRCh38
rs975455910 Chr.1: 100050290-100050290 on Build GRCh38
rs581097 Chr.1: 100050364-100050364 on Build GRCh38
rs577344909 Chr.1: 100050190-100050190 on Build GRCh38

View Full Product Details