Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43630309-43630801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF2BP
Entrez Gene ID
11077
Gene Name
heat shock transcription factor 2 binding protein
Gene Aliases
-
UniGene
Hs.406157
Gene Chromosome Location
Chr.21: 43453987-43659499 on Build GRCh38
Transcripts
NM_007031.1 XM_011529443.2 XM_011529445.2 XM_011529446.2 XM_011529447.2 XM_017028267.1 XM_017028268.1 XM_017028269.1 XM_017028270.1 XM_017028271.1 XM_017028272.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4101865 c.469A>G p.T157A Chr.21: 43630427-43630427 on Build GRCh38
4164855 c.473G>A p.G158D Chr.21: 43630423-43630423 on Build GRCh38
1327382 c.501G>A p.S167S Chr.21: 43630395-43630395 on Build GRCh38
5532438 c.482T>C p.M161T Chr.21: 43630414-43630414 on Build GRCh38
1162256 c.507C>T p.D169D Chr.21: 43630389-43630389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556363312 Chr.21: 43630697-43630697 on Build GRCh38
rs148591527 Chr.21: 43630379-43630379 on Build GRCh38
rs761974681 Chr.21: 43630472-43630473 on Build GRCh38
rs375041296 Chr.21: 43630343-43630343 on Build GRCh38
rs377477420 Chr.21: 43630492-43630492 on Build GRCh38
rs909931201 Chr.21: 43630677-43630677 on Build GRCh38
rs372205488 Chr.21: 43630635-43630635 on Build GRCh38
rs917276810 Chr.21: 43630600-43630600 on Build GRCh38
rs779174127 Chr.21: 43630495-43630495 on Build GRCh38
rs750365241 Chr.21: 43630388-43630388 on Build GRCh38
rs200214253 Chr.21: 43630424-43630424 on Build GRCh38
rs751221863 Chr.21: 43630455-43630455 on Build GRCh38
rs566497332 Chr.21: 43630663-43630663 on Build GRCh38
rs147021279 Chr.21: 43630328-43630328 on Build GRCh38
rs373629090 Chr.21: 43630470-43630470 on Build GRCh38
rs373359025 Chr.21: 43630410-43630410 on Build GRCh38
rs554222819 Chr.21: 43630555-43630555 on Build GRCh38
rs143866318 Chr.21: 43630553-43630553 on Build GRCh38
rs779086075 Chr.21: 43630414-43630414 on Build GRCh38
rs921450782 Chr.21: 43630366-43630366 on Build GRCh38
rs558859956 Chr.21: 43630667-43630667 on Build GRCh38
rs762714999 Chr.21: 43630451-43630451 on Build GRCh38
rs751569523 Chr.21: 43630336-43630336 on Build GRCh38
rs80173670 Chr.21: 43630757-43630757 on Build GRCh38
rs77849024 Chr.21: 43630665-43630665 on Build GRCh38
rs753968882 Chr.21: 43630489-43630489 on Build GRCh38
rs190341172 Chr.21: 43630480-43630480 on Build GRCh38
rs376045952 Chr.21: 43630728-43630728 on Build GRCh38
rs768819769 Chr.21: 43630413-43630413 on Build GRCh38
rs779849697 Chr.21: 43630510-43630510 on Build GRCh38
rs766471357 Chr.21: 43630488-43630488 on Build GRCh38
rs140147527 Chr.21: 43630364-43630364 on Build GRCh38
rs769190824 Chr.21: 43630430-43630430 on Build GRCh38
rs1015697318 Chr.21: 43630668-43630668 on Build GRCh38
rs748388677 Chr.21: 43630497-43630497 on Build GRCh38
rs1051405941 Chr.21: 43630448-43630448 on Build GRCh38
rs761901910 Chr.21: 43630344-43630344 on Build GRCh38
rs751240449 Chr.21: 43630520-43630520 on Build GRCh38
rs775098572 Chr.21: 43630434-43630434 on Build GRCh38
rs934641587 Chr.21: 43630431-43630431 on Build GRCh38
rs544407573 Chr.21: 43630695-43630695 on Build GRCh38
rs900048216 Chr.21: 43630428-43630428 on Build GRCh38
rs138271603 Chr.21: 43630341-43630341 on Build GRCh38
rs995854645 Chr.21: 43630429-43630429 on Build GRCh38
rs202091286 Chr.21: 43630474-43630474 on Build GRCh38
rs1034694924 Chr.21: 43630549-43630549 on Build GRCh38
rs761185639 Chr.21: 43630427-43630427 on Build GRCh38
rs992689166 Chr.21: 43630566-43630566 on Build GRCh38
rs757051161 Chr.21: 43630378-43630378 on Build GRCh38
rs866318407 Chr.21: 43630613-43630613 on Build GRCh38
rs531978498 Chr.21: 43630371-43630371 on Build GRCh38
rs763884466 Chr.21: 43630452-43630452 on Build GRCh38
rs200655253 Chr.21: 43630396-43630396 on Build GRCh38
rs11908893 Chr.21: 43630332-43630332 on Build GRCh38
rs756041744 Chr.21: 43630484-43630484 on Build GRCh38
rs577019972 Chr.21: 43630681-43630681 on Build GRCh38
rs780045050 Chr.21: 43630395-43630395 on Build GRCh38
rs549915111 Chr.21: 43630389-43630389 on Build GRCh38
rs1001868275 Chr.21: 43630530-43630530 on Build GRCh38
rs571045633 Chr.21: 43630365-43630365 on Build GRCh38
rs1051333192 Chr.21: 43630736-43630736 on Build GRCh38
rs748502183 Chr.21: 43630415-43630415 on Build GRCh38
rs747641120 Chr.21: 43630678-43630678 on Build GRCh38
rs982450105 Chr.21: 43630337-43630337 on Build GRCh38

View Full Product Details