Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148451089-148451618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR4
Entrez Gene ID
3360
Gene Name
5-hydroxytryptamine receptor 4
Gene Aliases
5-HT4, 5-HT4R
UniGene
Hs.483773
Gene Chromosome Location
Chr.5: 148451032-148654527 on Build GRCh38
Transcripts
NM_001040169.2 NM_199453.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6170079 c.1116G>C p.E372D Chr.5: 148451233-148451233 on Build GRCh38
6170078 c.1114G>T p.E372* Chr.5: 148451235-148451235 on Build GRCh38
5018320 c.1139delC p.P380fs*>8 Chr.5: 148451210-148451210 on Build GRCh38
6170080 c.1148T>A p.L383Q Chr.5: 148451201-148451201 on Build GRCh38
388104 c.1113C>T p.L371L Chr.5: 148451236-148451236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944008228 Chr.5: 148451526-148451526 on Build GRCh38
rs780491280 Chr.5: 148451211-148451211 on Build GRCh38
rs1008464358 Chr.5: 148451544-148451544 on Build GRCh38
rs773888647 Chr.5: 148451212-148451212 on Build GRCh38
rs201244212 Chr.5: 148451170-148451170 on Build GRCh38
rs181874273 Chr.5: 148451371-148451371 on Build GRCh38
rs200329789 Chr.5: 148451151-148451151 on Build GRCh38
rs991809722 Chr.5: 148451127-148451127 on Build GRCh38
rs112975240 Chr.5: 148451417-148451417 on Build GRCh38
rs201201172 Chr.5: 148451265-148451265 on Build GRCh38
rs569040545 Chr.5: 148451575-148451575 on Build GRCh38
rs774685276 Chr.5: 148451293-148451293 on Build GRCh38
rs765886319 Chr.5: 148451136-148451136 on Build GRCh38
rs757011799 Chr.5: 148451165-148451165 on Build GRCh38
rs778672638 Chr.5: 148451552-148451552 on Build GRCh38
rs369831099 Chr.5: 148451248-148451248 on Build GRCh38
rs775718821 Chr.5: 148451308-148451308 on Build GRCh38
rs146547482 Chr.5: 148451236-148451236 on Build GRCh38
rs776539429 Chr.5: 148451532-148451532 on Build GRCh38
rs912473853 Chr.5: 148451518-148451518 on Build GRCh38
rs550636092 Chr.5: 148451572-148451572 on Build GRCh38
rs1044789536 Chr.5: 148451482-148451482 on Build GRCh38
rs768029951 Chr.5: 148451261-148451261 on Build GRCh38
rs932500933 Chr.5: 148451507-148451507 on Build GRCh38
rs768411943 Chr.5: 148451192-148451192 on Build GRCh38
rs1029256691 Chr.5: 148451403-148451403 on Build GRCh38
rs753696321 Chr.5: 148451159-148451159 on Build GRCh38
rs771416974 Chr.5: 148451301-148451301 on Build GRCh38
rs202214083 Chr.5: 148451173-148451173 on Build GRCh38
rs747707642 Chr.5: 148451235-148451235 on Build GRCh38
rs762318794 Chr.5: 148451204-148451204 on Build GRCh38
rs796522910 Chr.5: 148451324-148451325 on Build GRCh38
rs749606815 Chr.5: 148451276-148451276 on Build GRCh38
rs762264016 Chr.5: 148451135-148451135 on Build GRCh38
rs908238907 Chr.5: 148451399-148451399 on Build GRCh38
rs1038080938 Chr.5: 148451160-148451160 on Build GRCh38
rs376377532 Chr.5: 148451206-148451206 on Build GRCh38
rs199967111 Chr.5: 148451118-148451118 on Build GRCh38
rs115413216 Chr.5: 148451325-148451325 on Build GRCh38
rs781261418 Chr.5: 148451155-148451155 on Build GRCh38
rs202071733 Chr.5: 148451150-148451150 on Build GRCh38
rs548468722 Chr.5: 148451450-148451450 on Build GRCh38
rs898639926 Chr.5: 148451296-148451296 on Build GRCh38
rs189513783 Chr.5: 148451327-148451327 on Build GRCh38
rs767533263 Chr.5: 148451300-148451300 on Build GRCh38
rs199619780 Chr.5: 148451266-148451266 on Build GRCh38
rs201133383 Chr.5: 148451134-148451134 on Build GRCh38
rs779842145 Chr.5: 148451249-148451249 on Build GRCh38
rs945339803 Chr.5: 148451594-148451594 on Build GRCh38
rs763453342 Chr.5: 148451220-148451220 on Build GRCh38
rs1053437679 Chr.5: 148451570-148451570 on Build GRCh38
rs771539821 Chr.5: 148451283-148451283 on Build GRCh38
rs200342469 Chr.5: 148451176-148451176 on Build GRCh38
rs964502484 Chr.5: 148451355-148451355 on Build GRCh38
rs759706410 Chr.5: 148451295-148451295 on Build GRCh38
rs372616383 Chr.5: 148451253-148451253 on Build GRCh38
rs770289684 Chr.5: 148451271-148451271 on Build GRCh38
rs538634680 Chr.5: 148451515-148451515 on Build GRCh38
rs201869859 Chr.5: 148451117-148451117 on Build GRCh38
rs199793887 Chr.5: 148451228-148451228 on Build GRCh38
rs200707776 Chr.5: 148451273-148451273 on Build GRCh38
rs992263124 Chr.5: 148451452-148451452 on Build GRCh38
rs750944902 Chr.5: 148451143-148451143 on Build GRCh38
rs766384809 Chr.5: 148451224-148451224 on Build GRCh38
rs776392350 Chr.5: 148451197-148451197 on Build GRCh38
rs979321232 Chr.5: 148451414-148451414 on Build GRCh38
rs560757048 Chr.5: 148451469-148451469 on Build GRCh38
rs931428039 Chr.5: 148451508-148451508 on Build GRCh38
rs975012790 Chr.5: 148451164-148451164 on Build GRCh38
rs760817429 Chr.5: 148451314-148451314 on Build GRCh38
rs887107572 Chr.5: 148451326-148451326 on Build GRCh38
rs746654331 Chr.5: 148451191-148451191 on Build GRCh38
rs1019600267 Chr.5: 148451328-148451328 on Build GRCh38
rs967821009 Chr.5: 148451404-148451404 on Build GRCh38
rs968229904 Chr.5: 148451309-148451309 on Build GRCh38
rs779548812 Chr.5: 148451184-148451184 on Build GRCh38
rs201026517 Chr.5: 148451227-148451227 on Build GRCh38
rs189051315 Chr.5: 148451486-148451486 on Build GRCh38
rs917571660 Chr.5: 148451462-148451462 on Build GRCh38
rs935239642 Chr.5: 148451429-148451429 on Build GRCh38
rs185016026 Chr.5: 148451478-148451478 on Build GRCh38
rs540745040 Chr.5: 148451225-148451225 on Build GRCh38
rs986888446 Chr.5: 148451131-148451131 on Build GRCh38
rs763150311 Chr.5: 148451148-148451148 on Build GRCh38
rs35228439 Chr.5: 148451208-148451208 on Build GRCh38
rs181721050 Chr.5: 148451512-148451512 on Build GRCh38
rs1020052788 Chr.5: 148451562-148451562 on Build GRCh38

View Full Product Details