Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89403406-89403908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIT1
Entrez Gene ID
3434
Gene Name
interferon induced protein with tetratricopeptide repeats 1
Gene Aliases
C56, G10P1, IFI-56, IFI-56K, IFI56, IFIT-1, IFNAI1, ISG56, P56, RNM561
UniGene
Hs.20315
Gene Chromosome Location
Chr.10: 89392546-89406487 on Build GRCh38
Transcripts
NM_001270927.1 NM_001270928.1 NM_001270929.1 NM_001270930.1 NM_001548.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4957821 c.1251A>G p.K417K Chr.10: 89403526-89403526 on Build GRCh38
1968737 c.1327C>T p.L443F Chr.10: 89403602-89403602 on Build GRCh38
6344161 c.1330G>T p.G444W Chr.10: 89403605-89403605 on Build GRCh38
6455907 c.1156C>T p.R386W Chr.10: 89403431-89403431 on Build GRCh38
1968739 c.1384C>T p.R462W Chr.10: 89403659-89403659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567346992 Chr.10: 89403448-89403448 on Build GRCh38
rs758970620 Chr.10: 89403461-89403461 on Build GRCh38
rs200504886 Chr.10: 89403520-89403520 on Build GRCh38
rs749646750 Chr.10: 89403663-89403663 on Build GRCh38
rs923633798 Chr.10: 89403694-89403694 on Build GRCh38
rs544662751 Chr.10: 89403818-89403818 on Build GRCh38
rs149279973 Chr.10: 89403633-89403633 on Build GRCh38
rs751246202 Chr.10: 89403656-89403656 on Build GRCh38
rs766802096 Chr.10: 89403497-89403497 on Build GRCh38
rs779391588 Chr.10: 89403657-89403657 on Build GRCh38
rs377321153 Chr.10: 89403630-89403630 on Build GRCh38
rs187430618 Chr.10: 89403753-89403753 on Build GRCh38
rs930690399 Chr.10: 89403676-89403676 on Build GRCh38
rs776325161 Chr.10: 89403661-89403661 on Build GRCh38
rs556284768 Chr.10: 89403880-89403880 on Build GRCh38
rs201610844 Chr.10: 89403567-89403567 on Build GRCh38
rs149781102 Chr.10: 89403879-89403880 on Build GRCh38
rs768096763 Chr.10: 89403708-89403708 on Build GRCh38
rs909132140 Chr.10: 89403827-89403827 on Build GRCh38
rs753225912 Chr.10: 89403737-89403741 on Build GRCh38
rs1005534310 Chr.10: 89403431-89403431 on Build GRCh38
rs555473177 Chr.10: 89403724-89403724 on Build GRCh38
rs941977264 Chr.10: 89403856-89403856 on Build GRCh38
rs777657919 Chr.10: 89403521-89403521 on Build GRCh38
rs781471256 Chr.10: 89403512-89403512 on Build GRCh38
rs774785394 Chr.10: 89403606-89403606 on Build GRCh38
rs750102354 Chr.10: 89403493-89403493 on Build GRCh38
rs754156662 Chr.10: 89403748-89403748 on Build GRCh38
rs745845470 Chr.10: 89403649-89403649 on Build GRCh38
rs1031885469 Chr.10: 89403622-89403622 on Build GRCh38
rs900785662 Chr.10: 89403821-89403821 on Build GRCh38
rs139524521 Chr.10: 89403453-89403453 on Build GRCh38
rs762138107 Chr.10: 89403465-89403465 on Build GRCh38
rs994668292 Chr.10: 89403476-89403476 on Build GRCh38
rs201342263 Chr.10: 89403669-89403669 on Build GRCh38
rs758894171 Chr.10: 89403762-89403774 on Build GRCh38
rs750808912 Chr.10: 89403479-89403479 on Build GRCh38
rs558170816 Chr.10: 89403471-89403471 on Build GRCh38
rs757055837 Chr.10: 89403751-89403751 on Build GRCh38
rs1047816719 Chr.10: 89403688-89403688 on Build GRCh38
rs183139112 Chr.10: 89403460-89403460 on Build GRCh38
rs944911951 Chr.10: 89403734-89403734 on Build GRCh38
rs370975704 Chr.10: 89403515-89403515 on Build GRCh38
rs759911243 Chr.10: 89403700-89403700 on Build GRCh38
rs375850117 Chr.10: 89403611-89403611 on Build GRCh38
rs920676927 Chr.10: 89403660-89403660 on Build GRCh38
rs993128904 Chr.10: 89403632-89403632 on Build GRCh38
rs773499776 Chr.10: 89403575-89403575 on Build GRCh38
rs144483233 Chr.10: 89403659-89403659 on Build GRCh38
rs765632811 Chr.10: 89403701-89403702 on Build GRCh38
rs372737209 Chr.10: 89403610-89403610 on Build GRCh38
rs79377094 Chr.10: 89403432-89403432 on Build GRCh38
rs571195246 Chr.10: 89403566-89403566 on Build GRCh38
rs756360767 Chr.10: 89403613-89403613 on Build GRCh38
rs771744609 Chr.10: 89403464-89403464 on Build GRCh38
rs774899198 Chr.10: 89403690-89403690 on Build GRCh38
rs923080931 Chr.10: 89403782-89403785 on Build GRCh38
rs147620337 Chr.10: 89403534-89403534 on Build GRCh38
rs770923139 Chr.10: 89403527-89403527 on Build GRCh38
rs751601507 Chr.10: 89403502-89403502 on Build GRCh38
rs766898563 Chr.10: 89403603-89403603 on Build GRCh38
rs755069064 Chr.10: 89403511-89403511 on Build GRCh38
rs763411464 Chr.10: 89403493-89403493 on Build GRCh38
rs764395347 Chr.10: 89403727-89403727 on Build GRCh38
rs755297444 Chr.10: 89403528-89403528 on Build GRCh38
rs775640877 Chr.10: 89403716-89403716 on Build GRCh38
rs763322304 Chr.10: 89403584-89403584 on Build GRCh38
rs996507857 Chr.10: 89403871-89403871 on Build GRCh38
rs745382349 Chr.10: 89403541-89403541 on Build GRCh38
rs775189682 Chr.10: 89403561-89403561 on Build GRCh38
rs202103656 Chr.10: 89403640-89403640 on Build GRCh38
rs771525327 Chr.10: 89403543-89403543 on Build GRCh38

View Full Product Details