Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1733371-1733893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFITM10
Entrez Gene ID
402778
Gene Name
interferon induced transmembrane protein 10
Gene Aliases
DSPA3
UniGene
Hs.121575
Gene Chromosome Location
Chr.11: 1732410-1750594 on Build GRCh38
Transcripts
NM_001170820.3

SNP Detail

SNP ID Chromosome Location
rs3740626 Chr.11: 1733420-1733420 on Build GRCh38
rs558552352 Chr.11: 1733686-1733686 on Build GRCh38
rs145165445 Chr.11: 1733812-1733812 on Build GRCh38
rs770247404 Chr.11: 1733700-1733700 on Build GRCh38
rs948116801 Chr.11: 1733617-1733617 on Build GRCh38
rs935429950 Chr.11: 1733660-1733660 on Build GRCh38
rs951003578 Chr.11: 1733781-1733781 on Build GRCh38
rs998689259 Chr.11: 1733730-1733730 on Build GRCh38
rs150061800 Chr.11: 1733757-1733757 on Build GRCh38
rs919111947 Chr.11: 1733862-1733863 on Build GRCh38
rs956099671 Chr.11: 1733718-1733718 on Build GRCh38
rs1010843619 Chr.11: 1733457-1733457 on Build GRCh38
rs1000306608 Chr.11: 1733667-1733669 on Build GRCh38
rs1013135167 Chr.11: 1733610-1733610 on Build GRCh38
rs543250377 Chr.11: 1733507-1733507 on Build GRCh38
rs1046507294 Chr.11: 1733402-1733402 on Build GRCh38
rs897567572 Chr.11: 1733562-1733562 on Build GRCh38
rs982356194 Chr.11: 1733836-1733836 on Build GRCh38
rs1037575661 Chr.11: 1733553-1733553 on Build GRCh38
rs970050088 Chr.11: 1733833-1733833 on Build GRCh38
rs537014193 Chr.11: 1733637-1733637 on Build GRCh38
rs147943375 Chr.11: 1733426-1733426 on Build GRCh38
rs190973353 Chr.11: 1733500-1733500 on Build GRCh38
rs906621742 Chr.11: 1733404-1733404 on Build GRCh38
rs904818332 Chr.11: 1733635-1733635 on Build GRCh38
rs908059379 Chr.11: 1733598-1733598 on Build GRCh38
rs922611445 Chr.11: 1733505-1733505 on Build GRCh38
rs982495100 Chr.11: 1733556-1733556 on Build GRCh38
rs1013605789 Chr.11: 1733417-1733417 on Build GRCh38
rs1018592063 Chr.11: 1733458-1733458 on Build GRCh38
rs895485843 Chr.11: 1733607-1733607 on Build GRCh38
rs956152136 Chr.11: 1733476-1733476 on Build GRCh38
rs185769778 Chr.11: 1733440-1733440 on Build GRCh38
rs1016679960 Chr.11: 1733805-1733805 on Build GRCh38
rs562940250 Chr.11: 1733542-1733542 on Build GRCh38
rs375885022 Chr.11: 1733806-1733806 on Build GRCh38
rs531747291 Chr.11: 1733563-1733563 on Build GRCh38
rs773303191 Chr.11: 1733758-1733758 on Build GRCh38
rs865840286 Chr.11: 1733419-1733419 on Build GRCh38
rs545311312 Chr.11: 1733807-1733807 on Build GRCh38
rs963321431 Chr.11: 1733808-1733808 on Build GRCh38
rs906508795 Chr.11: 1733649-1733649 on Build GRCh38
rs576250059 Chr.11: 1733395-1733395 on Build GRCh38
rs910345983 Chr.11: 1733538-1733543 on Build GRCh38
rs973708529 Chr.11: 1733602-1733602 on Build GRCh38
rs929072297 Chr.11: 1733574-1733574 on Build GRCh38
rs893665215 Chr.11: 1733733-1733733 on Build GRCh38
rs1011146067 Chr.11: 1733742-1733742 on Build GRCh38
rs571530914 Chr.11: 1733394-1733394 on Build GRCh38
rs916712964 Chr.11: 1733828-1733828 on Build GRCh38
rs1029453776 Chr.11: 1733527-1733527 on Build GRCh38
rs982552086 Chr.11: 1733783-1733783 on Build GRCh38
rs916809913 Chr.11: 1733613-1733613 on Build GRCh38
rs942157162 Chr.11: 1733600-1733600 on Build GRCh38
rs1029340709 Chr.11: 1733774-1733774 on Build GRCh38
rs140143172 Chr.11: 1733746-1733746 on Build GRCh38

View Full Product Details