Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27446099-27446501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT172
Entrez Gene ID
26160
Gene Name
intraflagellar transport 172
Gene Aliases
BBS20, NPHP17, RP71, SLB, SRTD10, osm-1, wim
UniGene
Hs.127401
Gene Chromosome Location
Chr.2: 27444373-27489819 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Gene Symbol
KRTCAP3
Entrez Gene ID
200634
Gene Name
keratinocyte associated protein 3
Gene Aliases
KCP3
UniGene
Hs.59509
Gene Chromosome Location
Chr.2: 27442366-27446481 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4826416 c.4675A>G p.R1559G Chr.2: 27446340-27446340 on Build GRCh38
281864 c.4721A>G p.D1574G Chr.2: 27446294-27446294 on Build GRCh38
6261122 c.4701C>A p.H1567Q Chr.2: 27446314-27446314 on Build GRCh38
6684031 c.4672G>A p.A1558T Chr.2: 27446343-27446343 on Build GRCh38
1019697 c.4701C>G p.H1567Q Chr.2: 27446314-27446314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778725103 Chr.2: 27446244-27446244 on Build GRCh38
rs536833971 Chr.2: 27446469-27446469 on Build GRCh38
rs114253314 Chr.2: 27446150-27446150 on Build GRCh38
rs141098495 Chr.2: 27446349-27446349 on Build GRCh38
rs768149033 Chr.2: 27446260-27446260 on Build GRCh38
rs375261962 Chr.2: 27446251-27446251 on Build GRCh38
rs11333597 Chr.2: 27446475-27446475 on Build GRCh38
rs776771308 Chr.2: 27446401-27446401 on Build GRCh38
rs370933808 Chr.2: 27446326-27446326 on Build GRCh38
rs777980611 Chr.2: 27446338-27446338 on Build GRCh38
rs756728624 Chr.2: 27446334-27446334 on Build GRCh38
rs786205855 Chr.2: 27446314-27446314 on Build GRCh38
rs1012346808 Chr.2: 27446447-27446447 on Build GRCh38
rs756273133 Chr.2: 27446223-27446223 on Build GRCh38
rs566292247 Chr.2: 27446408-27446408 on Build GRCh38
rs369307448 Chr.2: 27446229-27446229 on Build GRCh38
rs61742074 Chr.2: 27446270-27446270 on Build GRCh38
rs888578414 Chr.2: 27446415-27446415 on Build GRCh38
rs139979160 Chr.2: 27446133-27446135 on Build GRCh38
rs771110267 Chr.2: 27446235-27446235 on Build GRCh38
rs372614272 Chr.2: 27446387-27446387 on Build GRCh38
rs139062436 Chr.2: 27446398-27446398 on Build GRCh38
rs761504464 Chr.2: 27446282-27446282 on Build GRCh38
rs757464563 Chr.2: 27446342-27446342 on Build GRCh38
rs994445963 Chr.2: 27446155-27446155 on Build GRCh38
rs376189412 Chr.2: 27446137-27446139 on Build GRCh38
rs878941908 Chr.2: 27446234-27446234 on Build GRCh38
rs915032282 Chr.2: 27446318-27446318 on Build GRCh38
rs369487724 Chr.2: 27446252-27446252 on Build GRCh38
rs558185038 Chr.2: 27446213-27446213 on Build GRCh38
rs745618994 Chr.2: 27446364-27446364 on Build GRCh38
rs749498090 Chr.2: 27446231-27446231 on Build GRCh38
rs936897069 Chr.2: 27446444-27446444 on Build GRCh38
rs747641741 Chr.2: 27446400-27446400 on Build GRCh38
rs376974765 Chr.2: 27446389-27446389 on Build GRCh38
rs918818981 Chr.2: 27446369-27446369 on Build GRCh38
rs1006823501 Chr.2: 27446196-27446196 on Build GRCh38
rs772502337 Chr.2: 27446238-27446239 on Build GRCh38
rs769417539 Chr.2: 27446405-27446405 on Build GRCh38
rs781771585 Chr.2: 27446209-27446209 on Build GRCh38
rs779891948 Chr.2: 27446373-27446373 on Build GRCh38
rs759979976 Chr.2: 27446312-27446312 on Build GRCh38
rs953134859 Chr.2: 27446323-27446323 on Build GRCh38
rs925489125 Chr.2: 27446442-27446442 on Build GRCh38
rs1030462015 Chr.2: 27446272-27446272 on Build GRCh38
rs764818890 Chr.2: 27446283-27446283 on Build GRCh38
rs1026821400 Chr.2: 27446167-27446167 on Build GRCh38
rs573058783 Chr.2: 27446154-27446154 on Build GRCh38
rs571624967 Chr.2: 27446299-27446299 on Build GRCh38
rs776563892 Chr.2: 27446247-27446247 on Build GRCh38
rs897338264 Chr.2: 27446124-27446124 on Build GRCh38
rs971103505 Chr.2: 27446204-27446204 on Build GRCh38
rs753891390 Chr.2: 27446340-27446340 on Build GRCh38
rs141196620 Chr.2: 27446173-27446173 on Build GRCh38
rs771843422 Chr.2: 27446367-27446367 on Build GRCh38
rs775669156 Chr.2: 27446249-27446249 on Build GRCh38

View Full Product Details