Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 43613478-43614000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT52
Entrez Gene ID
51098
Gene Name
intraflagellar transport 52
Gene Aliases
C20orf9, CGI-53, NGD2, NGD5
UniGene
Hs.444332
Gene Chromosome Location
Chr.20: 43590613-43647296 on Build GRCh38
Transcripts
NM_001303458.2 NM_001303459.2 NM_001323578.1 NM_001323579.1 NM_001323580.1 NM_001323581.1 NM_016004.4 XM_017027863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3363261 c.573C>A p.F191L Chr.20: 43613937-43613937 on Build GRCh38
577730 c.612G>T p.K204N Chr.20: 43613976-43613976 on Build GRCh38
478178 c.546G>A p.A182A Chr.20: 43613910-43613910 on Build GRCh38
724551 c.565G>C p.V189L Chr.20: 43613929-43613929 on Build GRCh38
5028695 c.602A>G p.Y201C Chr.20: 43613966-43613966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773616340 Chr.20: 43613875-43613875 on Build GRCh38
rs758241733 Chr.20: 43613949-43613949 on Build GRCh38
rs772230218 Chr.20: 43613824-43613824 on Build GRCh38
rs762365050 Chr.20: 43613847-43613847 on Build GRCh38
rs775945843 Chr.20: 43613777-43613777 on Build GRCh38
rs113764648 Chr.20: 43613636-43613636 on Build GRCh38
rs774611753 Chr.20: 43613510-43613510 on Build GRCh38
rs752780937 Chr.20: 43613754-43613754 on Build GRCh38
rs867855085 Chr.20: 43613551-43613551 on Build GRCh38
rs768979989 Chr.20: 43613940-43613940 on Build GRCh38
rs1043129959 Chr.20: 43613534-43613534 on Build GRCh38
rs1031673802 Chr.20: 43613935-43613935 on Build GRCh38
rs943257065 Chr.20: 43613725-43613725 on Build GRCh38
rs896056399 Chr.20: 43613591-43613591 on Build GRCh38
rs761369330 Chr.20: 43613808-43613808 on Build GRCh38
rs772617116 Chr.20: 43613734-43613734 on Build GRCh38
rs750454266 Chr.20: 43613920-43613921 on Build GRCh38
rs768707464 Chr.20: 43613970-43613970 on Build GRCh38
rs139093073 Chr.20: 43613893-43613893 on Build GRCh38
rs963582526 Chr.20: 43613656-43613656 on Build GRCh38
rs757304461 Chr.20: 43613914-43613914 on Build GRCh38
rs1039250302 Chr.20: 43613511-43613511 on Build GRCh38
rs918478392 Chr.20: 43613733-43613733 on Build GRCh38
rs6017124 Chr.20: 43613778-43613778 on Build GRCh38
rs886037869 Chr.20: 43613959-43613959 on Build GRCh38
rs778825696 Chr.20: 43613945-43613945 on Build GRCh38
rs766004382 Chr.20: 43613910-43613910 on Build GRCh38
rs895836168 Chr.20: 43613931-43613931 on Build GRCh38
rs1023533205 Chr.20: 43613642-43613642 on Build GRCh38
rs762543292 Chr.20: 43613890-43613890 on Build GRCh38
rs1024156225 Chr.20: 43613598-43613598 on Build GRCh38
rs746963347 Chr.20: 43613762-43613762 on Build GRCh38
rs1031033053 Chr.20: 43613570-43613570 on Build GRCh38
rs189463599 Chr.20: 43613679-43613679 on Build GRCh38
rs999727777 Chr.20: 43613557-43613557 on Build GRCh38
rs146213304 Chr.20: 43613884-43613884 on Build GRCh38
rs780070990 Chr.20: 43613964-43613964 on Build GRCh38
rs775799322 Chr.20: 43613825-43613825 on Build GRCh38
rs776758734 Chr.20: 43613974-43613974 on Build GRCh38
rs909700152 Chr.20: 43613708-43613708 on Build GRCh38
rs368598294 Chr.20: 43613854-43613854 on Build GRCh38
rs372133278 Chr.20: 43613946-43613946 on Build GRCh38
rs747016258 Chr.20: 43613834-43613834 on Build GRCh38
rs372532772 Chr.20: 43613803-43613803 on Build GRCh38
rs768726052 Chr.20: 43613835-43613835 on Build GRCh38
rs143476502 Chr.20: 43613966-43613966 on Build GRCh38
rs777064142 Chr.20: 43613840-43613840 on Build GRCh38
rs150518875 Chr.20: 43613909-43613910 on Build GRCh38
rs1035675140 Chr.20: 43613569-43613569 on Build GRCh38
rs987116642 Chr.20: 43613711-43613711 on Build GRCh38
rs893657814 Chr.20: 43613516-43613516 on Build GRCh38
rs76895994 Chr.20: 43613716-43613716 on Build GRCh38
rs148203804 Chr.20: 43613519-43613519 on Build GRCh38
rs950487807 Chr.20: 43613689-43613689 on Build GRCh38
rs755525066 Chr.20: 43613909-43613909 on Build GRCh38
rs1022404491 Chr.20: 43613520-43613520 on Build GRCh38
rs1026061772 Chr.20: 43613688-43613688 on Build GRCh38
rs781302097 Chr.20: 43613827-43613828 on Build GRCh38
rs960998010 Chr.20: 43613584-43613584 on Build GRCh38
rs992133313 Chr.20: 43613585-43613585 on Build GRCh38
rs903308975 Chr.20: 43613545-43613545 on Build GRCh38
rs746068227 Chr.20: 43613821-43613821 on Build GRCh38
rs915023176 Chr.20: 43613785-43613785 on Build GRCh38
rs933656548 Chr.20: 43613769-43613769 on Build GRCh38
rs60813278 Chr.20: 43613812-43613812 on Build GRCh38
rs946562227 Chr.20: 43613793-43613793 on Build GRCh38

View Full Product Details