Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136430720-136431160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5E
Entrez Gene ID
56623
Gene Name
inositol polyphosphate-5-phosphatase E
Gene Aliases
CORS1, CPD4, JBTS1, MORMS, PPI5PIV
UniGene
Hs.120998
Gene Chromosome Location
Chr.9: 136428615-136439861 on Build GRCh38
Transcripts
NM_001318502.1 NM_019892.5 XM_017014926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832973 c.1598C>T p.S533L Chr.9: 136431069-136431069 on Build GRCh38
3905682 c.1652C>T p.T551M Chr.9: 136431015-136431015 on Build GRCh38
3699547 c.1653G>A p.T551T Chr.9: 136431014-136431014 on Build GRCh38
7208009 c.1614C>T p.I538I Chr.9: 136431053-136431053 on Build GRCh38
3748984 c.1552T>G p.S518A Chr.9: 136431115-136431115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577420256 Chr.9: 136430875-136430875 on Build GRCh38
rs762137331 Chr.9: 136431107-136431107 on Build GRCh38
rs927827274 Chr.9: 136430842-136430842 on Build GRCh38
rs1054422459 Chr.9: 136430950-136430950 on Build GRCh38
rs964987505 Chr.9: 136430918-136430918 on Build GRCh38
rs541897913 Chr.9: 136430936-136430936 on Build GRCh38
rs757592615 Chr.9: 136431069-136431069 on Build GRCh38
rs989722252 Chr.9: 136430920-136430920 on Build GRCh38
rs375126841 Chr.9: 136431052-136431052 on Build GRCh38
rs771113008 Chr.9: 136431089-136431090 on Build GRCh38
rs75568636 Chr.9: 136430860-136430860 on Build GRCh38
rs934763510 Chr.9: 136430943-136430943 on Build GRCh38
rs916409747 Chr.9: 136431030-136431030 on Build GRCh38
rs748250664 Chr.9: 136430765-136430765 on Build GRCh38
rs984435188 Chr.9: 136430896-136430896 on Build GRCh38
rs769912825 Chr.9: 136431089-136431089 on Build GRCh38
rs966661242 Chr.9: 136430930-136430930 on Build GRCh38
rs941589520 Chr.9: 136430917-136430917 on Build GRCh38
rs10870184 Chr.9: 136430739-136430739 on Build GRCh38
rs753398503 Chr.9: 136431038-136431038 on Build GRCh38
rs369050561 Chr.9: 136430981-136430981 on Build GRCh38
rs376485593 Chr.9: 136431026-136431026 on Build GRCh38
rs773033007 Chr.9: 136431106-136431106 on Build GRCh38
rs765986823 Chr.9: 136431031-136431031 on Build GRCh38
rs539405556 Chr.9: 136430841-136430841 on Build GRCh38
rs201644680 Chr.9: 136431116-136431116 on Build GRCh38
rs954556116 Chr.9: 136430786-136430786 on Build GRCh38
rs950409881 Chr.9: 136431083-136431083 on Build GRCh38
rs370618502 Chr.9: 136431042-136431042 on Build GRCh38
rs755521797 Chr.9: 136430956-136430956 on Build GRCh38
rs771866500 Chr.9: 136431102-136431102 on Build GRCh38
rs761578787 Chr.9: 136431003-136431003 on Build GRCh38
rs1029789856 Chr.9: 136430882-136430882 on Build GRCh38
rs373331546 Chr.9: 136430985-136430985 on Build GRCh38
rs752095117 Chr.9: 136431138-136431138 on Build GRCh38
rs75342839 Chr.9: 136431015-136431015 on Build GRCh38
rs774331779 Chr.9: 136431002-136431002 on Build GRCh38
rs768522686 Chr.9: 136430997-136430997 on Build GRCh38
rs746867724 Chr.9: 136431090-136431090 on Build GRCh38
rs781540345 Chr.9: 136431076-136431076 on Build GRCh38
rs77248046 Chr.9: 136431044-136431044 on Build GRCh38
rs1033440303 Chr.9: 136430783-136430783 on Build GRCh38
rs1009454706 Chr.9: 136430911-136430911 on Build GRCh38
rs376935441 Chr.9: 136430980-136430980 on Build GRCh38
rs989505032 Chr.9: 136430802-136430802 on Build GRCh38
rs531628363 Chr.9: 136431077-136431077 on Build GRCh38
rs1055930088 Chr.9: 136430739-136430745 on Build GRCh38
rs896826221 Chr.9: 136430876-136430876 on Build GRCh38
rs1045787734 Chr.9: 136430766-136430766 on Build GRCh38
rs945477322 Chr.9: 136430832-136430832 on Build GRCh38
rs563087580 Chr.9: 136430925-136430925 on Build GRCh38
rs568225897 Chr.9: 136431132-136431132 on Build GRCh38
rs1005414067 Chr.9: 136430862-136430862 on Build GRCh38
rs750136842 Chr.9: 136431021-136431021 on Build GRCh38
rs572055477 Chr.9: 136430803-136430803 on Build GRCh38
rs760443315 Chr.9: 136431115-136431115 on Build GRCh38
rs369317991 Chr.9: 136430858-136430858 on Build GRCh38
rs964724130 Chr.9: 136430782-136430782 on Build GRCh38
rs1017766523 Chr.9: 136430912-136430912 on Build GRCh38
rs879343800 Chr.9: 136430926-136430926 on Build GRCh38
rs574881928 Chr.9: 136430935-136430935 on Build GRCh38
rs772484080 Chr.9: 136430975-136430975 on Build GRCh38
rs371689939 Chr.9: 136430893-136430893 on Build GRCh38
rs748139386 Chr.9: 136431092-136431092 on Build GRCh38
rs553832167 Chr.9: 136430771-136430771 on Build GRCh38
rs747333701 Chr.9: 136431067-136431067 on Build GRCh38
rs1055069946 Chr.9: 136430855-136430855 on Build GRCh38
rs779456720 Chr.9: 136430968-136430968 on Build GRCh38
rs560146547 Chr.9: 136430942-136430942 on Build GRCh38
rs778220171 Chr.9: 136431053-136431053 on Build GRCh38
rs939152581 Chr.9: 136430852-136430852 on Build GRCh38
rs776879039 Chr.9: 136430986-136430986 on Build GRCh38
rs181576122 Chr.9: 136431131-136431131 on Build GRCh38
rs150464071 Chr.9: 136431041-136431041 on Build GRCh38
rs776265755 Chr.9: 136431130-136431130 on Build GRCh38
rs73566940 Chr.9: 136430902-136430902 on Build GRCh38
rs201386578 Chr.9: 136431137-136431137 on Build GRCh38
rs568105714 Chr.9: 136430738-136430738 on Build GRCh38
rs748524349 Chr.9: 136430970-136430970 on Build GRCh38
rs987147444 Chr.9: 136430927-136430927 on Build GRCh38
rs993935996 Chr.9: 136430877-136430877 on Build GRCh38
rs747219455 Chr.9: 136431082-136431082 on Build GRCh38
rs778019120 Chr.9: 136431015-136431016 on Build GRCh38

View Full Product Details