Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210022050-210022597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1L
Entrez Gene ID
151050
Gene Name
KAT8 regulatory NSL complex subunit 1 like
Gene Aliases
C2orf67, MSL1v2
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210021421-210171370 on Build GRCh38
Transcripts
NM_001307976.1 NM_152519.3 XM_005246328.4 XM_005246329.4 XM_005246330.4 XM_005246332.4 XM_011510706.2 XM_011510707.2 XM_011510710.2 XM_017003430.1 XM_017003431.1 NM_001278282.1 NM_001278283.1 NM_001278285.1 NM_001278286.1 NM_001278288.1 NM_001278289.1 NM_001318926.1 NM_001318927.1 NM_001318928.1 NM_001318929.1 NM_001318930.1 NM_001318931.1 NM_006916.2 NM_199229.2 XM_006712677.3

Gene Symbol
RPE
Entrez Gene ID
6120
Gene Name
ribulose-5-phosphate-3-epimerase
Gene Aliases
RPE2-1
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210002565-210029156 on Build GRCh38
Transcripts
NM_001307976.1 NM_152519.3 XM_005246328.4 XM_005246329.4 XM_005246330.4 XM_005246332.4 XM_011510706.2 XM_011510707.2 XM_011510710.2 XM_017003430.1 XM_017003431.1 NM_001278282.1 NM_001278283.1 NM_001278285.1 NM_001278286.1 NM_001278288.1 NM_001278289.1 NM_001318926.1 NM_001318927.1 NM_001318928.1 NM_001318929.1 NM_001318930.1 NM_001318931.1 NM_006916.2 NM_199229.2 XM_006712677.3

SNP Detail

SNP ID Chromosome Location
rs193179420 Chr.2: 210022351-210022351 on Build GRCh38
rs978309656 Chr.2: 210022419-210022419 on Build GRCh38
rs564705314 Chr.2: 210022549-210022549 on Build GRCh38
rs918031041 Chr.2: 210022475-210022476 on Build GRCh38
rs149558434 Chr.2: 210022479-210022480 on Build GRCh38
rs754916752 Chr.2: 210022480-210022481 on Build GRCh38
rs1048972703 Chr.2: 210022322-210022323 on Build GRCh38
rs935302494 Chr.2: 210022264-210022264 on Build GRCh38
rs901577687 Chr.2: 210022346-210022346 on Build GRCh38
rs542116319 Chr.2: 210022473-210022473 on Build GRCh38
rs371677759 Chr.2: 210022262-210022262 on Build GRCh38
rs564230299 Chr.2: 210022304-210022305 on Build GRCh38
rs763834634 Chr.2: 210022100-210022100 on Build GRCh38
rs576909116 Chr.2: 210022357-210022357 on Build GRCh38
rs989379589 Chr.2: 210022441-210022441 on Build GRCh38
rs572695713 Chr.2: 210022480-210022480 on Build GRCh38
rs556254168 Chr.2: 210022391-210022391 on Build GRCh38
rs995132288 Chr.2: 210022332-210022332 on Build GRCh38
rs776415892 Chr.2: 210022495-210022495 on Build GRCh38
rs73063916 Chr.2: 210022470-210022470 on Build GRCh38
rs546254398 Chr.2: 210022547-210022547 on Build GRCh38
rs1031176832 Chr.2: 210022425-210022425 on Build GRCh38
rs770540539 Chr.2: 210022171-210022172 on Build GRCh38
rs756486110 Chr.2: 210022338-210022338 on Build GRCh38
rs1044225553 Chr.2: 210022305-210022310 on Build GRCh38
rs906637542 Chr.2: 210022278-210022278 on Build GRCh38
rs923837578 Chr.2: 210022427-210022427 on Build GRCh38
rs1018666729 Chr.2: 210022104-210022104 on Build GRCh38
rs762551343 Chr.2: 210022093-210022093 on Build GRCh38
rs765835099 Chr.2: 210022307-210022308 on Build GRCh38
rs953672964 Chr.2: 210022354-210022354 on Build GRCh38
rs188640836 Chr.2: 210022322-210022322 on Build GRCh38
rs965128299 Chr.2: 210022404-210022404 on Build GRCh38
rs968741578 Chr.2: 210022413-210022413 on Build GRCh38
rs928023322 Chr.2: 210022502-210022502 on Build GRCh38
rs940779915 Chr.2: 210022538-210022538 on Build GRCh38
rs751018617 Chr.2: 210022109-210022109 on Build GRCh38
rs534515470 Chr.2: 210022298-210022298 on Build GRCh38
rs974906931 Chr.2: 210022196-210022196 on Build GRCh38
rs3180813 Chr.2: 210022550-210022550 on Build GRCh38
rs560807847 Chr.2: 210022336-210022336 on Build GRCh38
rs569663566 Chr.2: 210022150-210022150 on Build GRCh38
rs977963701 Chr.2: 210022110-210022110 on Build GRCh38
rs969482100 Chr.2: 210022479-210022484 on Build GRCh38
rs1029685042 Chr.2: 210022215-210022216 on Build GRCh38
rs899408970 Chr.2: 210022325-210022325 on Build GRCh38
rs958188451 Chr.2: 210022429-210022429 on Build GRCh38
rs542888283 Chr.2: 210022291-210022291 on Build GRCh38
rs923840298 Chr.2: 210022130-210022130 on Build GRCh38
rs917977007 Chr.2: 210022300-210022300 on Build GRCh38
rs371200259 Chr.2: 210022478-210022479 on Build GRCh38
rs895182822 Chr.2: 210022214-210022214 on Build GRCh38
rs936626058 Chr.2: 210022157-210022157 on Build GRCh38
rs1004527988 Chr.2: 210022340-210022340 on Build GRCh38
rs561578589 Chr.2: 210022380-210022381 on Build GRCh38
rs759487101 Chr.2: 210022305-210022307 on Build GRCh38
rs377280997 Chr.2: 210022364-210022364 on Build GRCh38
rs1006529891 Chr.2: 210022361-210022361 on Build GRCh38
rs905800024 Chr.2: 210022310-210022311 on Build GRCh38
rs13018349 Chr.2: 210022261-210022261 on Build GRCh38
rs562407490 Chr.2: 210022479-210022479 on Build GRCh38
rs968308499 Chr.2: 210022256-210022256 on Build GRCh38
rs878946991 Chr.2: 210022478-210022478 on Build GRCh38
rs1054194279 Chr.2: 210022160-210022160 on Build GRCh38
rs541634269 Chr.2: 210022546-210022546 on Build GRCh38

View Full Product Details