Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63329045-63329557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDSR
Entrez Gene ID
2531
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
DHSR, FVT1, SDR35C1
UniGene
Hs.74050
Gene Chromosome Location
Chr.18: 63327736-63367273 on Build GRCh38
Transcripts
NM_002035.2 XM_005266677.2 XM_017025690.1

SNP Detail

SNP ID Chromosome Location
rs977293287 Chr.18: 63329276-63329276 on Build GRCh38
rs546838153 Chr.18: 63329377-63329377 on Build GRCh38
rs943730746 Chr.18: 63329202-63329202 on Build GRCh38
rs183499262 Chr.18: 63329143-63329143 on Build GRCh38
rs997531280 Chr.18: 63329068-63329068 on Build GRCh38
rs141601714 Chr.18: 63329443-63329443 on Build GRCh38
rs748312759 Chr.18: 63329494-63329494 on Build GRCh38
rs568329226 Chr.18: 63329092-63329092 on Build GRCh38
rs973010592 Chr.18: 63329093-63329093 on Build GRCh38
rs910866075 Chr.18: 63329196-63329196 on Build GRCh38
rs538938667 Chr.18: 63329444-63329444 on Build GRCh38
rs188186407 Chr.18: 63329504-63329504 on Build GRCh38
rs115273133 Chr.18: 63329105-63329105 on Build GRCh38
rs566538740 Chr.18: 63329447-63329447 on Build GRCh38
rs961551640 Chr.18: 63329482-63329482 on Build GRCh38
rs1035785587 Chr.18: 63329159-63329159 on Build GRCh38
rs531042012 Chr.18: 63329219-63329219 on Build GRCh38
rs567019865 Chr.18: 63329215-63329215 on Build GRCh38
rs551046215 Chr.18: 63329288-63329288 on Build GRCh38
rs567760636 Chr.18: 63329327-63329327 on Build GRCh38
rs1049090211 Chr.18: 63329371-63329371 on Build GRCh38
rs770114574 Chr.18: 63329400-63329400 on Build GRCh38
rs958414279 Chr.18: 63329204-63329204 on Build GRCh38
rs972685335 Chr.18: 63329502-63329502 on Build GRCh38
rs536888438 Chr.18: 63329386-63329386 on Build GRCh38
rs770016867 Chr.18: 63329499-63329499 on Build GRCh38
rs10460159 Chr.18: 63329127-63329127 on Build GRCh38
rs892077045 Chr.18: 63329293-63329293 on Build GRCh38
rs759805245 Chr.18: 63329339-63329339 on Build GRCh38
rs575092075 Chr.18: 63329476-63329476 on Build GRCh38
rs924682823 Chr.18: 63329286-63329286 on Build GRCh38
rs991785937 Chr.18: 63329209-63329209 on Build GRCh38
rs541608120 Chr.18: 63329274-63329275 on Build GRCh38
rs1034449557 Chr.18: 63329407-63329407 on Build GRCh38
rs1041287130 Chr.18: 63329242-63329242 on Build GRCh38
rs368666331 Chr.18: 63329463-63329463 on Build GRCh38
rs1004348111 Chr.18: 63329107-63329107 on Build GRCh38
rs1010400272 Chr.18: 63329295-63329295 on Build GRCh38
rs904697413 Chr.18: 63329343-63329343 on Build GRCh38
rs547199866 Chr.18: 63329415-63329415 on Build GRCh38
rs1014589299 Chr.18: 63329109-63329109 on Build GRCh38

View Full Product Details