Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80880766-80881168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEMIP
Entrez Gene ID
57214
Gene Name
cell migration inducing hyaluronan binding protein
Gene Aliases
CCSP1, HYBID, KIAA1199, TMEM2L
UniGene
Hs.459088
Gene Chromosome Location
Chr.15: 80779343-80951662 on Build GRCh38
Transcripts
NM_001293298.1 NM_001293304.1 NM_018689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6030711 c.474G>A p.K158K Chr.15: 80880993-80880993 on Build GRCh38
3887576 c.464G>A p.G155E Chr.15: 80880983-80880983 on Build GRCh38
3504459 c.416G>A p.G139D Chr.15: 80880935-80880935 on Build GRCh38
3504460 c.479C>T p.S160F Chr.15: 80880998-80880998 on Build GRCh38
5952805 c.502C>G p.L168V Chr.15: 80881021-80881021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778874487 Chr.15: 80880990-80880990 on Build GRCh38
rs1023448038 Chr.15: 80881007-80881007 on Build GRCh38
rs375136109 Chr.15: 80881102-80881102 on Build GRCh38
rs749110354 Chr.15: 80880884-80880884 on Build GRCh38
rs139497635 Chr.15: 80880921-80880921 on Build GRCh38
rs779935139 Chr.15: 80880974-80880974 on Build GRCh38
rs144446375 Chr.15: 80881079-80881079 on Build GRCh38
rs976656495 Chr.15: 80881088-80881088 on Build GRCh38
rs776205085 Chr.15: 80881031-80881031 on Build GRCh38
rs780025059 Chr.15: 80880879-80880879 on Build GRCh38
rs72740115 Chr.15: 80880993-80880993 on Build GRCh38
rs146635298 Chr.15: 80881101-80881101 on Build GRCh38
rs745528766 Chr.15: 80881141-80881141 on Build GRCh38
rs754784643 Chr.15: 80881087-80881087 on Build GRCh38
rs778020593 Chr.15: 80880967-80880967 on Build GRCh38
rs560273892 Chr.15: 80880863-80880863 on Build GRCh38
rs771524778 Chr.15: 80881011-80881011 on Build GRCh38
rs778110456 Chr.15: 80880865-80880865 on Build GRCh38
rs149698686 Chr.15: 80880966-80880966 on Build GRCh38
rs768399715 Chr.15: 80880988-80880988 on Build GRCh38
rs142610156 Chr.15: 80881136-80881136 on Build GRCh38
rs377682740 Chr.15: 80880875-80880875 on Build GRCh38
rs750646524 Chr.15: 80881082-80881082 on Build GRCh38
rs1032028832 Chr.15: 80880904-80880904 on Build GRCh38
rs370472918 Chr.15: 80881035-80881035 on Build GRCh38
rs772742080 Chr.15: 80881013-80881013 on Build GRCh38
rs535071040 Chr.15: 80880797-80880798 on Build GRCh38
rs368428285 Chr.15: 80881135-80881135 on Build GRCh38
rs932367898 Chr.15: 80880793-80880793 on Build GRCh38
rs751790405 Chr.15: 80880968-80880968 on Build GRCh38
rs777209135 Chr.15: 80881121-80881121 on Build GRCh38
rs774209975 Chr.15: 80880900-80880900 on Build GRCh38
rs753314096 Chr.15: 80881053-80881053 on Build GRCh38
rs182850909 Chr.15: 80881142-80881142 on Build GRCh38
rs537795110 Chr.15: 80881059-80881059 on Build GRCh38
rs139680841 Chr.15: 80881044-80881044 on Build GRCh38
rs375373858 Chr.15: 80880970-80880970 on Build GRCh38
rs972241449 Chr.15: 80881089-80881089 on Build GRCh38
rs747282585 Chr.15: 80880867-80880867 on Build GRCh38
rs374461087 Chr.15: 80881054-80881054 on Build GRCh38
rs753198988 Chr.15: 80881053-80881053 on Build GRCh38
rs768648936 Chr.15: 80880889-80880889 on Build GRCh38
rs901075306 Chr.15: 80880962-80880962 on Build GRCh38
rs368854657 Chr.15: 80881078-80881078 on Build GRCh38
rs551524324 Chr.15: 80881066-80881066 on Build GRCh38
rs147026015 Chr.15: 80880798-80880798 on Build GRCh38
rs778598227 Chr.15: 80881090-80881090 on Build GRCh38
rs564424758 Chr.15: 80881131-80881131 on Build GRCh38
rs367627306 Chr.15: 80880975-80880975 on Build GRCh38
rs529223212 Chr.15: 80880909-80880909 on Build GRCh38
rs1042748863 Chr.15: 80880833-80880833 on Build GRCh38
rs748274137 Chr.15: 80880852-80880852 on Build GRCh38
rs761205116 Chr.15: 80881052-80881053 on Build GRCh38
rs963900211 Chr.15: 80881074-80881074 on Build GRCh38
rs546514004 Chr.15: 80880830-80880830 on Build GRCh38
rs370607399 Chr.15: 80880911-80880911 on Build GRCh38
rs948471054 Chr.15: 80880823-80880823 on Build GRCh38
rs765762495 Chr.15: 80880947-80880947 on Build GRCh38
rs568880342 Chr.15: 80881024-80881024 on Build GRCh38
rs754208381 Chr.15: 80880936-80880936 on Build GRCh38
rs894264472 Chr.15: 80880799-80880799 on Build GRCh38
rs766691380 Chr.15: 80880928-80880928 on Build GRCh38
rs544904377 Chr.15: 80881132-80881132 on Build GRCh38
rs893577545 Chr.15: 80880808-80880808 on Build GRCh38
rs903408476 Chr.15: 80880954-80880954 on Build GRCh38
rs753211103 Chr.15: 80880956-80880956 on Build GRCh38
rs754629555 Chr.15: 80880855-80880855 on Build GRCh38
rs759903288 Chr.15: 80880946-80880946 on Build GRCh38
rs919531612 Chr.15: 80881098-80881098 on Build GRCh38
rs960301233 Chr.15: 80880980-80880980 on Build GRCh38
rs770584637 Chr.15: 80881026-80881026 on Build GRCh38

View Full Product Details