Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132693617-132694114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF3A
Entrez Gene ID
11127
Gene Name
kinesin family member 3A
Gene Aliases
FLA10, KLP-20
UniGene
Hs.43670
Gene Chromosome Location
Chr.5: 132688808-132737633 on Build GRCh38
Transcripts
NM_001300791.1 NM_001300792.1 NM_007054.6 XM_006714526.3 XM_017008995.1 XM_017008996.1

SNP Detail

SNP ID Chromosome Location
rs1005248400 Chr.5: 132693803-132693803 on Build GRCh38
rs940605305 Chr.5: 132693937-132693937 on Build GRCh38
rs1139661 Chr.5: 132693755-132693755 on Build GRCh38
rs185408688 Chr.5: 132693959-132693959 on Build GRCh38
rs1002062070 Chr.5: 132694085-132694085 on Build GRCh38
rs1014264230 Chr.5: 132693810-132693810 on Build GRCh38
rs1139664 Chr.5: 132693747-132693747 on Build GRCh38
rs981338350 Chr.5: 132693709-132693709 on Build GRCh38
rs139334750 Chr.5: 132693960-132693960 on Build GRCh38
rs917402439 Chr.5: 132693707-132693707 on Build GRCh38
rs562002055 Chr.5: 132693992-132693992 on Build GRCh38
rs17539066 Chr.5: 132693674-132693674 on Build GRCh38
rs1139663 Chr.5: 132693748-132693748 on Build GRCh38
rs570610485 Chr.5: 132693733-132693733 on Build GRCh38
rs891355975 Chr.5: 132694092-132694092 on Build GRCh38
rs753479660 Chr.5: 132693981-132693981 on Build GRCh38
rs980301758 Chr.5: 132693837-132693837 on Build GRCh38
rs905946924 Chr.5: 132694077-132694077 on Build GRCh38
rs567913810 Chr.5: 132693690-132693690 on Build GRCh38
rs576178714 Chr.5: 132693986-132693986 on Build GRCh38
rs1018461420 Chr.5: 132693813-132693814 on Build GRCh38
rs190975173 Chr.5: 132693925-132693925 on Build GRCh38
rs1139660 Chr.5: 132693757-132693757 on Build GRCh38
rs868549858 Chr.5: 132693975-132693975 on Build GRCh38
rs866588430 Chr.5: 132693849-132693849 on Build GRCh38
rs914374550 Chr.5: 132693968-132693969 on Build GRCh38
rs556161740 Chr.5: 132693739-132693739 on Build GRCh38
rs756710166 Chr.5: 132693692-132693692 on Build GRCh38
rs755081982 Chr.5: 132693839-132693839 on Build GRCh38
rs1052777740 Chr.5: 132693735-132693735 on Build GRCh38
rs928497338 Chr.5: 132693932-132693932 on Build GRCh38
rs1062247 Chr.5: 132693704-132693704 on Build GRCh38
rs961387405 Chr.5: 132693807-132693807 on Build GRCh38
rs150074408 Chr.5: 132694054-132694054 on Build GRCh38
rs539442602 Chr.5: 132693738-132693738 on Build GRCh38
rs946843970 Chr.5: 132693980-132693980 on Build GRCh38
rs1017991722 Chr.5: 132693642-132693642 on Build GRCh38
rs554448371 Chr.5: 132693930-132693930 on Build GRCh38
rs751403052 Chr.5: 132693695-132693695 on Build GRCh38
rs771266570 Chr.5: 132693910-132693910 on Build GRCh38
rs887630814 Chr.5: 132693936-132693936 on Build GRCh38
rs1025338472 Chr.5: 132693885-132693885 on Build GRCh38
rs180748536 Chr.5: 132693935-132693935 on Build GRCh38
rs921689005 Chr.5: 132693918-132693918 on Build GRCh38
rs187328280 Chr.5: 132693804-132693804 on Build GRCh38
rs376103073 Chr.5: 132693950-132693950 on Build GRCh38
rs4705963 Chr.5: 132693799-132693799 on Build GRCh38
rs1043784569 Chr.5: 132693984-132693984 on Build GRCh38
rs935302382 Chr.5: 132694018-132694018 on Build GRCh38
rs951307315 Chr.5: 132693705-132693705 on Build GRCh38
rs1024308556 Chr.5: 132694066-132694066 on Build GRCh38
rs555364112 Chr.5: 132693826-132693826 on Build GRCh38
rs574966440 Chr.5: 132693867-132693867 on Build GRCh38
rs1040162978 Chr.5: 132693756-132693756 on Build GRCh38
rs989489523 Chr.5: 132693862-132693862 on Build GRCh38
rs564422478 Chr.5: 132694091-132694091 on Build GRCh38
rs748318132 Chr.5: 132694088-132694088 on Build GRCh38
rs1016046276 Chr.5: 132693649-132693649 on Build GRCh38
rs540631188 Chr.5: 132693729-132693729 on Build GRCh38
rs897138332 Chr.5: 132694014-132694014 on Build GRCh38
rs1024361046 Chr.5: 132693825-132693825 on Build GRCh38
rs992896922 Chr.5: 132693702-132693702 on Build GRCh38
rs944427466 Chr.5: 132693753-132693753 on Build GRCh38
rs970213304 Chr.5: 132693720-132693720 on Build GRCh38
rs749572666 Chr.5: 132693840-132693840 on Build GRCh38
rs17510339 Chr.5: 132693722-132693722 on Build GRCh38
rs541914288 Chr.5: 132693987-132693987 on Build GRCh38
rs879136998 Chr.5: 132693752-132693752 on Build GRCh38
rs944375565 Chr.5: 132693909-132693909 on Build GRCh38
rs1049418203 Chr.5: 132693786-132693786 on Build GRCh38
rs996014741 Chr.5: 132693765-132693765 on Build GRCh38
rs918521508 Chr.5: 132693699-132693699 on Build GRCh38
rs912875113 Chr.5: 132693903-132693903 on Build GRCh38
rs190048812 Chr.5: 132694026-132694026 on Build GRCh38

View Full Product Details