Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9652498-9652992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3CD
Entrez Gene ID
5293
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta
Gene Aliases
APDS, IMD14, P110DELTA, PI3K, p110D
UniGene
Hs.518451
Gene Chromosome Location
Chr.1: 9629889-9729114 on Build GRCh38
Transcripts
NM_005026.3 XM_006710689.2 XM_017001477.1 XM_017001478.1 XM_017001479.1 XM_017001480.1 NR_027045.1

Gene Symbol
PIK3CD-AS1
Entrez Gene ID
644997
Gene Name
PIK3CD antisense RNA 1
Gene Aliases
C1orf200
UniGene
Hs.568595
Gene Chromosome Location
Chr.1: 9652610-9654586 on Build GRCh38
Transcripts
NM_005026.3 XM_006710689.2 XM_017001477.1 XM_017001478.1 XM_017001479.1 XM_017001480.1 NR_027045.1

SNP Detail

SNP ID Chromosome Location
rs966556653 Chr.1: 9652835-9652835 on Build GRCh38
rs574356088 Chr.1: 9652683-9652683 on Build GRCh38
rs1038707037 Chr.1: 9652603-9652603 on Build GRCh38
rs1054394383 Chr.1: 9652860-9652860 on Build GRCh38
rs1015528936 Chr.1: 9652760-9652760 on Build GRCh38
rs921644147 Chr.1: 9652897-9652897 on Build GRCh38
rs1055227310 Chr.1: 9652576-9652577 on Build GRCh38
rs900140360 Chr.1: 9652620-9652620 on Build GRCh38
rs573336333 Chr.1: 9652577-9652577 on Build GRCh38
rs1032422114 Chr.1: 9652673-9652673 on Build GRCh38
rs1039173125 Chr.1: 9652892-9652892 on Build GRCh38
rs545658444 Chr.1: 9652776-9652776 on Build GRCh38
rs888702412 Chr.1: 9652936-9652936 on Build GRCh38
rs961162114 Chr.1: 9652767-9652767 on Build GRCh38
rs548684735 Chr.1: 9652848-9652848 on Build GRCh38
rs544943861 Chr.1: 9652686-9652686 on Build GRCh38
rs539499256 Chr.1: 9652538-9652538 on Build GRCh38
rs538447077 Chr.1: 9652949-9652949 on Build GRCh38
rs986615107 Chr.1: 9652531-9652531 on Build GRCh38
rs560953199 Chr.1: 9652854-9652854 on Build GRCh38
rs1033704865 Chr.1: 9652801-9652801 on Build GRCh38
rs944421920 Chr.1: 9652594-9652594 on Build GRCh38
rs919673600 Chr.1: 9652568-9652568 on Build GRCh38
rs557808848 Chr.1: 9652666-9652666 on Build GRCh38
rs1038089865 Chr.1: 9652558-9652558 on Build GRCh38
rs551569849 Chr.1: 9652735-9652735 on Build GRCh38
rs1037215573 Chr.1: 9652943-9652943 on Build GRCh38
rs955304812 Chr.1: 9652527-9652527 on Build GRCh38
rs944474486 Chr.1: 9652891-9652891 on Build GRCh38
rs911694545 Chr.1: 9652871-9652871 on Build GRCh38
rs917553001 Chr.1: 9652812-9652812 on Build GRCh38
rs915136213 Chr.1: 9652821-9652821 on Build GRCh38
rs939849077 Chr.1: 9652549-9652549 on Build GRCh38
rs921186099 Chr.1: 9652521-9652521 on Build GRCh38
rs906675420 Chr.1: 9652946-9652946 on Build GRCh38
rs533921040 Chr.1: 9652604-9652604 on Build GRCh38
rs930295211 Chr.1: 9652921-9652921 on Build GRCh38
rs111554447 Chr.1: 9652804-9652804 on Build GRCh38
rs549665264 Chr.1: 9652905-9652905 on Build GRCh38
rs774231281 Chr.1: 9652756-9652756 on Build GRCh38
rs902652930 Chr.1: 9652632-9652632 on Build GRCh38
rs115728442 Chr.1: 9652939-9652939 on Build GRCh38
rs1004007837 Chr.1: 9652798-9652798 on Build GRCh38
rs1042154888 Chr.1: 9652576-9652576 on Build GRCh38
rs557976346 Chr.1: 9652550-9652550 on Build GRCh38
rs534352064 Chr.1: 9652534-9652534 on Build GRCh38
rs559988388 Chr.1: 9652808-9652808 on Build GRCh38
rs527408632 Chr.1: 9652844-9652844 on Build GRCh38
rs954712976 Chr.1: 9652744-9652744 on Build GRCh38
rs531471364 Chr.1: 9652895-9652895 on Build GRCh38
rs970809771 Chr.1: 9652788-9652788 on Build GRCh38
rs1001441756 Chr.1: 9652655-9652655 on Build GRCh38
rs888649801 Chr.1: 9652727-9652727 on Build GRCh38
rs563025928 Chr.1: 9652718-9652718 on Build GRCh38
rs1048677452 Chr.1: 9652922-9652922 on Build GRCh38
rs779209999 Chr.1: 9652530-9652530 on Build GRCh38
rs923663745 Chr.1: 9652519-9652519 on Build GRCh38
rs896723174 Chr.1: 9652595-9652595 on Build GRCh38
rs989441948 Chr.1: 9652815-9652815 on Build GRCh38
rs997129339 Chr.1: 9652621-9652621 on Build GRCh38
rs945199580 Chr.1: 9652571-9652571 on Build GRCh38

View Full Product Details