Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 661916-662419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2C
Entrez Gene ID
22982
Gene Name
disco interacting protein 2 homolog C
Gene Aliases
KIAA0934
UniGene
Hs.432397
Gene Chromosome Location
Chr.10: 274190-689710 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_011519432.1 NR_027152.2

Gene Symbol
PRR26
Entrez Gene ID
414235
Gene Name
proline rich 26
Gene Aliases
C10orf108
UniGene
Hs.587103
Gene Chromosome Location
Chr.10: 649948-665169 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_011519432.1 NR_027152.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263891 c.379G>T p.G127C Chr.10: 662072-662072 on Build GRCh38
2159795 c.372C>T p.H124H Chr.10: 662065-662065 on Build GRCh38
6207212 c.333-4G>A p.? Chr.10: 662022-662022 on Build GRCh38
4165527 c.368C>T p.P123L Chr.10: 662061-662061 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563267842 Chr.10: 662024-662025 on Build GRCh38
rs889102788 Chr.10: 661999-662028 on Build GRCh38
rs964524263 Chr.10: 662076-662076 on Build GRCh38
rs917542904 Chr.10: 661996-661996 on Build GRCh38
rs188277910 Chr.10: 662176-662176 on Build GRCh38
rs774590065 Chr.10: 662065-662065 on Build GRCh38
rs960187918 Chr.10: 662381-662381 on Build GRCh38
rs867478409 Chr.10: 662295-662295 on Build GRCh38
rs929951700 Chr.10: 662276-662276 on Build GRCh38
rs781782215 Chr.10: 662087-662087 on Build GRCh38
rs943195411 Chr.10: 662201-662201 on Build GRCh38
rs573207990 Chr.10: 662246-662246 on Build GRCh38
rs980774237 Chr.10: 662213-662214 on Build GRCh38
rs143366207 Chr.10: 662174-662174 on Build GRCh38
rs1026227864 Chr.10: 662308-662308 on Build GRCh38
rs555976038 Chr.10: 662166-662166 on Build GRCh38
rs970094024 Chr.10: 662347-662347 on Build GRCh38
rs768944844 Chr.10: 662036-662036 on Build GRCh38
rs1006188729 Chr.10: 662323-662323 on Build GRCh38
rs17136375 Chr.10: 662353-662353 on Build GRCh38
rs775987299 Chr.10: 661984-661984 on Build GRCh38
rs1048095460 Chr.10: 662035-662035 on Build GRCh38
rs530584625 Chr.10: 662131-662131 on Build GRCh38
rs1038862082 Chr.10: 662227-662227 on Build GRCh38
rs71489277 Chr.10: 661985-661985 on Build GRCh38
rs891135301 Chr.10: 662305-662305 on Build GRCh38
rs913589529 Chr.10: 662161-662161 on Build GRCh38
rs994206993 Chr.10: 662030-662030 on Build GRCh38
rs1016775041 Chr.10: 662346-662346 on Build GRCh38
rs544802218 Chr.10: 662387-662387 on Build GRCh38
rs41289273 Chr.10: 661971-661971 on Build GRCh38
rs3853289 Chr.10: 662252-662252 on Build GRCh38
rs758585238 Chr.10: 662026-662026 on Build GRCh38
rs766144540 Chr.10: 662021-662021 on Build GRCh38
rs763287791 Chr.10: 662022-662022 on Build GRCh38
rs538633508 Chr.10: 662274-662274 on Build GRCh38
rs928910343 Chr.10: 662086-662086 on Build GRCh38
rs985320725 Chr.10: 662043-662043 on Build GRCh38
rs1051125351 Chr.10: 662284-662284 on Build GRCh38
rs1053363427 Chr.10: 662157-662157 on Build GRCh38
rs941512340 Chr.10: 662239-662239 on Build GRCh38
rs558955254 Chr.10: 662332-662332 on Build GRCh38
rs1035851420 Chr.10: 662364-662364 on Build GRCh38
rs777924708 Chr.10: 662062-662062 on Build GRCh38
rs866346311 Chr.10: 662297-662297 on Build GRCh38
rs903722120 Chr.10: 661953-661953 on Build GRCh38
rs117768263 Chr.10: 662008-662008 on Build GRCh38
rs535145173 Chr.10: 662017-662017 on Build GRCh38
rs746347721 Chr.10: 662145-662145 on Build GRCh38
rs890405361 Chr.10: 662044-662044 on Build GRCh38
rs530250012 Chr.10: 661937-661937 on Build GRCh38
rs927411851 Chr.10: 662234-662234 on Build GRCh38
rs567001789 Chr.10: 662169-662169 on Build GRCh38
rs138652954 Chr.10: 662077-662077 on Build GRCh38
rs538572841 Chr.10: 662315-662315 on Build GRCh38
rs909261277 Chr.10: 662045-662045 on Build GRCh38
rs767635731 Chr.10: 662175-662175 on Build GRCh38
rs551345401 Chr.10: 662073-662073 on Build GRCh38

View Full Product Details