Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 664465-665005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2C
Entrez Gene ID
22982
Gene Name
disco interacting protein 2 homolog C
Gene Aliases
KIAA0934
UniGene
Hs.432397
Gene Chromosome Location
Chr.10: 274190-689710 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_011519432.1 NR_027152.2

Gene Symbol
PRR26
Entrez Gene ID
414235
Gene Name
proline rich 26
Gene Aliases
C10orf108
UniGene
Hs.587103
Gene Chromosome Location
Chr.10: 649948-665169 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_011519432.1 NR_027152.2

SNP Detail

SNP ID Chromosome Location
rs771817975 Chr.10: 664821-664821 on Build GRCh38
rs1015395913 Chr.10: 664523-664523 on Build GRCh38
rs978367953 Chr.10: 664782-664782 on Build GRCh38
rs570755528 Chr.10: 664903-664903 on Build GRCh38
rs940725160 Chr.10: 664850-664850 on Build GRCh38
rs891017052 Chr.10: 664820-664820 on Build GRCh38
rs897154530 Chr.10: 664848-664848 on Build GRCh38
rs182603273 Chr.10: 664555-664555 on Build GRCh38
rs888391582 Chr.10: 664508-664508 on Build GRCh38
rs187069436 Chr.10: 664575-664575 on Build GRCh38
rs575778552 Chr.10: 664713-664713 on Build GRCh38
rs949762842 Chr.10: 664767-664767 on Build GRCh38
rs1054199673 Chr.10: 664796-664796 on Build GRCh38
rs549956245 Chr.10: 664661-664661 on Build GRCh38
rs191481775 Chr.10: 664626-664626 on Build GRCh38
rs796279932 Chr.10: 664762-664763 on Build GRCh38
rs924255823 Chr.10: 664793-664793 on Build GRCh38
rs566773586 Chr.10: 664644-664644 on Build GRCh38
rs886450056 Chr.10: 664665-664665 on Build GRCh38
rs540689335 Chr.10: 664641-664641 on Build GRCh38
rs1036941955 Chr.10: 664890-664890 on Build GRCh38
rs577225894 Chr.10: 664549-664549 on Build GRCh38
rs1026986805 Chr.10: 664865-664865 on Build GRCh38
rs965596149 Chr.10: 664535-664535 on Build GRCh38
rs953702077 Chr.10: 664586-664586 on Build GRCh38
rs939541003 Chr.10: 664636-664636 on Build GRCh38
rs576255880 Chr.10: 664949-664949 on Build GRCh38
rs543447100 Chr.10: 664697-664697 on Build GRCh38
rs907830648 Chr.10: 664845-664845 on Build GRCh38
rs1050116333 Chr.10: 664818-664818 on Build GRCh38
rs1035715363 Chr.10: 664790-664790 on Build GRCh38
rs561633070 Chr.10: 664623-664623 on Build GRCh38
rs538877064 Chr.10: 664682-664682 on Build GRCh38
rs934496537 Chr.10: 664505-664505 on Build GRCh38
rs1028012366 Chr.10: 664719-664719 on Build GRCh38
rs866254502 Chr.10: 664946-664946 on Build GRCh38
rs1021928879 Chr.10: 664749-664749 on Build GRCh38
rs989131003 Chr.10: 664589-664589 on Build GRCh38
rs189057886 Chr.10: 664984-664984 on Build GRCh38
rs12267081 Chr.10: 664728-664728 on Build GRCh38
rs191899165 Chr.10: 664526-664526 on Build GRCh38
rs566061406 Chr.10: 664779-664779 on Build GRCh38
rs1015624587 Chr.10: 664824-664824 on Build GRCh38
rs966783220 Chr.10: 664764-664764 on Build GRCh38
rs556505987 Chr.10: 664930-664930 on Build GRCh38
rs893016341 Chr.10: 664805-664805 on Build GRCh38
rs998008131 Chr.10: 664716-664716 on Build GRCh38
rs761508345 Chr.10: 664605-664605 on Build GRCh38
rs114101009 Chr.10: 664748-664748 on Build GRCh38
rs200851151 Chr.10: 664920-664921 on Build GRCh38
rs995587157 Chr.10: 664859-664859 on Build GRCh38
rs932434315 Chr.10: 664809-664809 on Build GRCh38
rs978739400 Chr.10: 664920-664920 on Build GRCh38
rs1008194653 Chr.10: 664628-664628 on Build GRCh38
rs753138344 Chr.10: 664968-664968 on Build GRCh38
rs930683582 Chr.10: 664819-664819 on Build GRCh38
rs115092195 Chr.10: 664823-664823 on Build GRCh38
rs541907342 Chr.10: 664614-664614 on Build GRCh38
rs372096472 Chr.10: 664591-664591 on Build GRCh38
rs151226431 Chr.10: 664646-664646 on Build GRCh38
rs751196403 Chr.10: 664590-664590 on Build GRCh38
rs539784476 Chr.10: 664907-664907 on Build GRCh38
rs1025663745 Chr.10: 664905-664905 on Build GRCh38
rs955690361 Chr.10: 664942-664942 on Build GRCh38
rs184134698 Chr.10: 664794-664794 on Build GRCh38
rs1054316109 Chr.10: 664522-664522 on Build GRCh38
rs974115302 Chr.10: 664649-664649 on Build GRCh38
rs959538866 Chr.10: 664902-664902 on Build GRCh38
rs901001815 Chr.10: 664715-664715 on Build GRCh38
rs981104914 Chr.10: 664780-664780 on Build GRCh38
rs924337318 Chr.10: 664933-664933 on Build GRCh38
rs763879708 Chr.10: 664556-664556 on Build GRCh38

View Full Product Details