Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26939139-26939634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26
Entrez Gene ID
22852
Gene Name
ankyrin repeat domain 26
Gene Aliases
THC2, bA145E8.1
UniGene
Hs.361041
Gene Chromosome Location
Chr.10: 26938195-27100498 on Build GRCh38
Transcripts
XM_017015929.1 NR_026795.1

Gene Symbol
LINC00202-1
Entrez Gene ID
387644
Gene Name
long intergenic non-protein coding RNA 202-1
Gene Aliases
C10orf51, LINC00202, NCRNA00202, bB27G4.1
UniGene
Hs.529965
Gene Chromosome Location
Chr.10: 26931206-26942001 on Build GRCh38
Transcripts
XM_017015929.1 NR_026795.1

SNP Detail

SNP ID Chromosome Location
rs951385296 Chr.10: 26939586-26939586 on Build GRCh38
rs184377836 Chr.10: 26939463-26939463 on Build GRCh38
rs998734625 Chr.10: 26939475-26939475 on Build GRCh38
rs878954059 Chr.10: 26939567-26939567 on Build GRCh38
rs182265583 Chr.10: 26939283-26939283 on Build GRCh38
rs535493013 Chr.10: 26939434-26939434 on Build GRCh38
rs796661752 Chr.10: 26939224-26939224 on Build GRCh38
rs146983761 Chr.10: 26939353-26939353 on Build GRCh38
rs531575723 Chr.10: 26939523-26939523 on Build GRCh38
rs982971410 Chr.10: 26939588-26939588 on Build GRCh38
rs560710293 Chr.10: 26939477-26939477 on Build GRCh38
rs575111893 Chr.10: 26939456-26939456 on Build GRCh38
rs2488585 Chr.10: 26939259-26939259 on Build GRCh38
rs562688070 Chr.10: 26939518-26939518 on Build GRCh38
rs1034949029 Chr.10: 26939573-26939573 on Build GRCh38
rs12573020 Chr.10: 26939492-26939492 on Build GRCh38
rs570574777 Chr.10: 26939348-26939348 on Build GRCh38
rs957405739 Chr.10: 26939496-26939496 on Build GRCh38
rs1050743007 Chr.10: 26939393-26939393 on Build GRCh38
rs910147914 Chr.10: 26939592-26939592 on Build GRCh38
rs547532314 Chr.10: 26939333-26939333 on Build GRCh38
rs755699957 Chr.10: 26939454-26939454 on Build GRCh38
rs969440395 Chr.10: 26939538-26939538 on Build GRCh38
rs576099461 Chr.10: 26939419-26939419 on Build GRCh38
rs764767136 Chr.10: 26939336-26939336 on Build GRCh38
rs754718062 Chr.10: 26939458-26939458 on Build GRCh38
rs796198831 Chr.10: 26939229-26939229 on Build GRCh38
rs11015399 Chr.10: 26939510-26939510 on Build GRCh38
rs749888162 Chr.10: 26939453-26939453 on Build GRCh38
rs539470016 Chr.10: 26939350-26939350 on Build GRCh38
rs562501085 Chr.10: 26939220-26939220 on Build GRCh38
rs982193611 Chr.10: 26939563-26939563 on Build GRCh38
rs141395485 Chr.10: 26939327-26939327 on Build GRCh38
rs555283483 Chr.10: 26939455-26939455 on Build GRCh38
rs369257729 Chr.10: 26939317-26939317 on Build GRCh38
rs552412631 Chr.10: 26939217-26939217 on Build GRCh38
rs1012541194 Chr.10: 26939202-26939202 on Build GRCh38

View Full Product Details