Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26941383-26941874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26
Entrez Gene ID
22852
Gene Name
ankyrin repeat domain 26
Gene Aliases
THC2, bA145E8.1
UniGene
Hs.361041
Gene Chromosome Location
Chr.10: 26938195-27100498 on Build GRCh38
Transcripts
XM_017015929.1 NR_026795.1

Gene Symbol
LINC00202-1
Entrez Gene ID
387644
Gene Name
long intergenic non-protein coding RNA 202-1
Gene Aliases
C10orf51, LINC00202, NCRNA00202, bB27G4.1
UniGene
Hs.529965
Gene Chromosome Location
Chr.10: 26931206-26942001 on Build GRCh38
Transcripts
XM_017015929.1 NR_026795.1

SNP Detail

SNP ID Chromosome Location
rs190818562 Chr.10: 26941504-26941504 on Build GRCh38
rs985514746 Chr.10: 26941540-26941540 on Build GRCh38
rs573857432 Chr.10: 26941513-26941513 on Build GRCh38
rs113846435 Chr.10: 26941599-26941599 on Build GRCh38
rs75143508 Chr.10: 26941598-26941598 on Build GRCh38
rs551447452 Chr.10: 26941518-26941518 on Build GRCh38
rs77778595 Chr.10: 26941597-26941597 on Build GRCh38
rs983062887 Chr.10: 26941461-26941461 on Build GRCh38
rs787722 Chr.10: 26941600-26941600 on Build GRCh38
rs987285275 Chr.10: 26941715-26941715 on Build GRCh38
rs535137247 Chr.10: 26941413-26941413 on Build GRCh38
rs891829158 Chr.10: 26941567-26941567 on Build GRCh38
rs796343038 Chr.10: 26941850-26941850 on Build GRCh38
rs879461886 Chr.10: 26941597-26941598 on Build GRCh38
rs761774533 Chr.10: 26941446-26941446 on Build GRCh38
rs562744688 Chr.10: 26941450-26941450 on Build GRCh38
rs183246796 Chr.10: 26941404-26941404 on Build GRCh38
rs944128060 Chr.10: 26941556-26941556 on Build GRCh38
rs187726557 Chr.10: 26941807-26941807 on Build GRCh38
rs75551836 Chr.10: 26941810-26941810 on Build GRCh38
rs927529940 Chr.10: 26941825-26941825 on Build GRCh38
rs1057219900 Chr.10: 26941840-26941840 on Build GRCh38
rs531755312 Chr.10: 26941516-26941516 on Build GRCh38
rs192841679 Chr.10: 26941670-26941670 on Build GRCh38
rs71504724 Chr.10: 26941597-26941598 on Build GRCh38
rs906034459 Chr.10: 26941431-26941431 on Build GRCh38
rs931823768 Chr.10: 26941532-26941532 on Build GRCh38
rs774671187 Chr.10: 26941445-26941445 on Build GRCh38
rs912640048 Chr.10: 26941542-26941542 on Build GRCh38
rs1001216276 Chr.10: 26941439-26941439 on Build GRCh38
rs182644831 Chr.10: 26941779-26941779 on Build GRCh38
rs766027693 Chr.10: 26941747-26941747 on Build GRCh38
rs34163327 Chr.10: 26941692-26941692 on Build GRCh38
rs947422437 Chr.10: 26941800-26941800 on Build GRCh38
rs963081693 Chr.10: 26941672-26941672 on Build GRCh38
rs186410967 Chr.10: 26941501-26941501 on Build GRCh38
rs563616292 Chr.10: 26941512-26941512 on Build GRCh38
rs947587585 Chr.10: 26941585-26941585 on Build GRCh38
rs544437322 Chr.10: 26941759-26941759 on Build GRCh38
rs78463442 Chr.10: 26941814-26941814 on Build GRCh38
rs1053528220 Chr.10: 26941560-26941560 on Build GRCh38
rs386742196 Chr.10: 26941597-26941600 on Build GRCh38
rs1004998412 Chr.10: 26941843-26941843 on Build GRCh38
rs557840018 Chr.10: 26941429-26941429 on Build GRCh38
rs973350012 Chr.10: 26941509-26941509 on Build GRCh38
rs921724145 Chr.10: 26941511-26941511 on Build GRCh38
rs965445454 Chr.10: 26941751-26941751 on Build GRCh38
rs565423549 Chr.10: 26941569-26941569 on Build GRCh38
rs895564916 Chr.10: 26941636-26941636 on Build GRCh38
rs11015402 Chr.10: 26941746-26941746 on Build GRCh38
rs188108475 Chr.10: 26941647-26941647 on Build GRCh38
rs548003156 Chr.10: 26941618-26941618 on Build GRCh38
rs1022202478 Chr.10: 26941418-26941418 on Build GRCh38
rs995141564 Chr.10: 26941687-26941687 on Build GRCh38
rs144503362 Chr.10: 26941828-26941828 on Build GRCh38
rs116159826 Chr.10: 26941696-26941696 on Build GRCh38
rs895994200 Chr.10: 26941842-26941842 on Build GRCh38

View Full Product Details