Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88991888-88992397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAS
Entrez Gene ID
355
Gene Name
Fas cell surface death receptor
Gene Aliases
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
UniGene
Hs.244139
Gene Chromosome Location
Chr.10: 88968429-89017061 on Build GRCh38
Transcripts
NM_000043.5 NM_001320619.1 NM_152871.3 NM_152872.3 NR_028033.3 NR_028034.3 NR_028035.3 NR_028036.3 NR_135313.1 NR_135314.1 NR_135315.1 XM_006717819.3 XM_011539764.2 XM_011539765.2 XM_011539766.2 XM_011539767.2 NR_028371.1

Gene Symbol
FAS-AS1
Entrez Gene ID
100302740
Gene Name
FAS antisense RNA 1
Gene Aliases
FAS-AS, FASAS, SAF
UniGene
Hs.244139
Gene Chromosome Location
Chr.10: 88991422-88992975 on Build GRCh38
Transcripts
NM_000043.5 NM_001320619.1 NM_152871.3 NM_152872.3 NR_028033.3 NR_028034.3 NR_028035.3 NR_028036.3 NR_135313.1 NR_135314.1 NR_135315.1 XM_006717819.3 XM_011539764.2 XM_011539765.2 XM_011539766.2 XM_011539767.2 NR_028371.1

SNP Detail

SNP ID Chromosome Location
rs745824317 Chr.10: 88992325-88992325 on Build GRCh38
rs10509561 Chr.10: 88992155-88992155 on Build GRCh38
rs769161006 Chr.10: 88991943-88991943 on Build GRCh38
rs764066452 Chr.10: 88992232-88992232 on Build GRCh38
rs758150166 Chr.10: 88992354-88992354 on Build GRCh38
rs955953832 Chr.10: 88992109-88992109 on Build GRCh38
rs886896461 Chr.10: 88992299-88992299 on Build GRCh38
rs147036839 Chr.10: 88992035-88992035 on Build GRCh38
rs930010490 Chr.10: 88992215-88992216 on Build GRCh38
rs114955372 Chr.10: 88991930-88991930 on Build GRCh38
rs774671346 Chr.10: 88991986-88991986 on Build GRCh38
rs942589098 Chr.10: 88992179-88992179 on Build GRCh38
rs6586164 Chr.10: 88992274-88992274 on Build GRCh38
rs1044328024 Chr.10: 88991996-88991997 on Build GRCh38
rs531442100 Chr.10: 88992305-88992305 on Build GRCh38
rs1054266942 Chr.10: 88992283-88992283 on Build GRCh38
rs3824730 Chr.10: 88992371-88992371 on Build GRCh38
rs971322189 Chr.10: 88992030-88992030 on Build GRCh38
rs1014552233 Chr.10: 88991987-88991987 on Build GRCh38
rs905817488 Chr.10: 88992031-88992031 on Build GRCh38
rs567092206 Chr.10: 88992369-88992369 on Build GRCh38
rs527537883 Chr.10: 88992224-88992224 on Build GRCh38
rs955076011 Chr.10: 88992153-88992153 on Build GRCh38
rs963584227 Chr.10: 88991978-88991978 on Build GRCh38
rs1016575293 Chr.10: 88992319-88992319 on Build GRCh38
rs948031461 Chr.10: 88992239-88992239 on Build GRCh38
rs1010006732 Chr.10: 88992112-88992112 on Build GRCh38
rs1032874546 Chr.10: 88992075-88992075 on Build GRCh38
rs545464475 Chr.10: 88992210-88992210 on Build GRCh38
rs1012480834 Chr.10: 88992355-88992355 on Build GRCh38
rs775006722 Chr.10: 88991985-88991985 on Build GRCh38
rs760677836 Chr.10: 88992204-88992204 on Build GRCh38
rs778781591 Chr.10: 88992302-88992302 on Build GRCh38
rs1023775816 Chr.10: 88992023-88992023 on Build GRCh38
rs911473636 Chr.10: 88992171-88992171 on Build GRCh38
rs560175595 Chr.10: 88992222-88992222 on Build GRCh38
rs918779216 Chr.10: 88992181-88992181 on Build GRCh38
rs897215852 Chr.10: 88991944-88991944 on Build GRCh38
rs974181839 Chr.10: 88992177-88992177 on Build GRCh38
rs777513306 Chr.10: 88992357-88992357 on Build GRCh38
rs6586163 Chr.10: 88992261-88992261 on Build GRCh38
rs1045114321 Chr.10: 88992257-88992257 on Build GRCh38
rs148155729 Chr.10: 88992125-88992125 on Build GRCh38
rs567837001 Chr.10: 88992176-88992176 on Build GRCh38
rs989480101 Chr.10: 88992170-88992170 on Build GRCh38
rs766937196 Chr.10: 88991961-88991961 on Build GRCh38
rs1005254690 Chr.10: 88991948-88991948 on Build GRCh38
rs951064245 Chr.10: 88991940-88991940 on Build GRCh38
rs896487193 Chr.10: 88991983-88991985 on Build GRCh38

View Full Product Details