Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88992904-88993417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAS
Entrez Gene ID
355
Gene Name
Fas cell surface death receptor
Gene Aliases
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
UniGene
Hs.244139
Gene Chromosome Location
Chr.10: 88968429-89017061 on Build GRCh38
Transcripts
NM_000043.5 NM_001320619.1 NM_152871.3 NM_152872.3 NR_028033.3 NR_028034.3 NR_028035.3 NR_028036.3 NR_135313.1 NR_135314.1 NR_135315.1 XM_006717819.3 XM_011539764.2 XM_011539765.2 XM_011539766.2 XM_011539767.2 NR_028371.1

Gene Symbol
FAS-AS1
Entrez Gene ID
100302740
Gene Name
FAS antisense RNA 1
Gene Aliases
FAS-AS, FASAS, SAF
UniGene
Hs.244139
Gene Chromosome Location
Chr.10: 88991422-88992975 on Build GRCh38
Transcripts
NM_000043.5 NM_001320619.1 NM_152871.3 NM_152872.3 NR_028033.3 NR_028034.3 NR_028035.3 NR_028036.3 NR_135313.1 NR_135314.1 NR_135315.1 XM_006717819.3 XM_011539764.2 XM_011539765.2 XM_011539766.2 XM_011539767.2 NR_028371.1

SNP Detail

SNP ID Chromosome Location
rs948412080 Chr.10: 88993220-88993220 on Build GRCh38
rs190609320 Chr.10: 88993271-88993271 on Build GRCh38
rs55803311 Chr.10: 88993378-88993378 on Build GRCh38
rs982732126 Chr.10: 88993009-88993009 on Build GRCh38
rs987128760 Chr.10: 88993265-88993265 on Build GRCh38
rs922629309 Chr.10: 88992984-88992984 on Build GRCh38
rs1025108020 Chr.10: 88993138-88993138 on Build GRCh38
rs912512211 Chr.10: 88993267-88993267 on Build GRCh38
rs1014271492 Chr.10: 88993115-88993115 on Build GRCh38
rs76193522 Chr.10: 88993318-88993318 on Build GRCh38
rs186035382 Chr.10: 88993200-88993200 on Build GRCh38
rs1039419442 Chr.10: 88993007-88993007 on Build GRCh38
rs373884946 Chr.10: 88993228-88993228 on Build GRCh38
rs1052656529 Chr.10: 88993263-88993263 on Build GRCh38
rs980971259 Chr.10: 88993144-88993144 on Build GRCh38
rs775991564 Chr.10: 88993232-88993232 on Build GRCh38
rs1030046923 Chr.10: 88993337-88993337 on Build GRCh38
rs554086663 Chr.10: 88993234-88993234 on Build GRCh38
rs769322523 Chr.10: 88993325-88993325 on Build GRCh38
rs397953719 Chr.10: 88993317-88993318 on Build GRCh38
rs955433716 Chr.10: 88992999-88992999 on Build GRCh38
rs774847081 Chr.10: 88993342-88993342 on Build GRCh38
rs150665502 Chr.10: 88993374-88993374 on Build GRCh38
rs969553329 Chr.10: 88993142-88993142 on Build GRCh38
rs941198450 Chr.10: 88993122-88993122 on Build GRCh38
rs906878065 Chr.10: 88993235-88993235 on Build GRCh38
rs10887877 Chr.10: 88993281-88993281 on Build GRCh38
rs7076197 Chr.10: 88993377-88993377 on Build GRCh38
rs1025760123 Chr.10: 88993071-88993071 on Build GRCh38
rs973942602 Chr.10: 88993386-88993386 on Build GRCh38
rs1032426466 Chr.10: 88993190-88993190 on Build GRCh38
rs890959022 Chr.10: 88992964-88992964 on Build GRCh38
rs955419136 Chr.10: 88993344-88993344 on Build GRCh38
rs12359362 Chr.10: 88993388-88993388 on Build GRCh38
rs887337527 Chr.10: 88993096-88993096 on Build GRCh38
rs898288285 Chr.10: 88993044-88993044 on Build GRCh38
rs958134358 Chr.10: 88993191-88993191 on Build GRCh38
rs1045772781 Chr.10: 88993233-88993233 on Build GRCh38
rs942417089 Chr.10: 88993005-88993005 on Build GRCh38
rs59583029 Chr.10: 88993305-88993306 on Build GRCh38
rs1004176305 Chr.10: 88993236-88993236 on Build GRCh38
rs908350345 Chr.10: 88993054-88993054 on Build GRCh38
rs11596616 Chr.10: 88993322-88993322 on Build GRCh38
rs1038112718 Chr.10: 88993149-88993149 on Build GRCh38
rs964435058 Chr.10: 88993323-88993323 on Build GRCh38

View Full Product Details