Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46753025-46753533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP5
Entrez Gene ID
9793
Gene Name
cytoskeleton associated protein 5
Gene Aliases
CHTOG, MSPS, TOG, TOGp, ch-TOG
UniGene
Hs.201253
Gene Chromosome Location
Chr.11: 46743534-46846308 on Build GRCh38
Transcripts
NM_001008938.3 NM_014756.3 NR_049846.1

Gene Symbol
MIR5582
Entrez Gene ID
100847020
Gene Name
microRNA 5582
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 46753125-46753192 on Build GRCh38
Transcripts
NM_001008938.3 NM_014756.3 NR_049846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5372524 c.5052C>T p.I1684I Chr.11: 46753315-46753315 on Build GRCh38
3809287 c.4878G>C p.Q1626H Chr.11: 46753489-46753489 on Build GRCh38
352371 c.5055G>C p.L1685L Chr.11: 46753312-46753312 on Build GRCh38
927299 c.4957T>G p.S1653A Chr.11: 46753410-46753410 on Build GRCh38
5372525 c.5052C>T p.I1684I Chr.11: 46753315-46753315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144359837 Chr.11: 46753137-46753138 on Build GRCh38
rs770753241 Chr.11: 46753302-46753302 on Build GRCh38
rs575063426 Chr.11: 46753107-46753107 on Build GRCh38
rs564974043 Chr.11: 46753236-46753236 on Build GRCh38
rs776879803 Chr.11: 46753354-46753354 on Build GRCh38
rs554132563 Chr.11: 46753178-46753178 on Build GRCh38
rs905831862 Chr.11: 46753319-46753319 on Build GRCh38
rs759450365 Chr.11: 46753293-46753293 on Build GRCh38
rs775766467 Chr.11: 46753222-46753222 on Build GRCh38
rs758806157 Chr.11: 46753172-46753172 on Build GRCh38
rs889478190 Chr.11: 46753119-46753122 on Build GRCh38
rs267602897 Chr.11: 46753463-46753463 on Build GRCh38
rs370599536 Chr.11: 46753269-46753269 on Build GRCh38
rs72895811 Chr.11: 46753122-46753122 on Build GRCh38
rs544204787 Chr.11: 46753081-46753081 on Build GRCh38
rs773964900 Chr.11: 46753287-46753287 on Build GRCh38
rs767529537 Chr.11: 46753418-46753418 on Build GRCh38
rs769801478 Chr.11: 46753508-46753508 on Build GRCh38
rs752282517 Chr.11: 46753272-46753272 on Build GRCh38
rs1048103961 Chr.11: 46753148-46753148 on Build GRCh38
rs144138791 Chr.11: 46753462-46753462 on Build GRCh38
rs532729696 Chr.11: 46753190-46753190 on Build GRCh38
rs781383858 Chr.11: 46753499-46753499 on Build GRCh38
rs763294001 Chr.11: 46753242-46753242 on Build GRCh38
rs779737708 Chr.11: 46753185-46753185 on Build GRCh38
rs780701108 Chr.11: 46753271-46753271 on Build GRCh38
rs777829660 Chr.11: 46753288-46753288 on Build GRCh38
rs754509311 Chr.11: 46753196-46753196 on Build GRCh38
rs774597459 Chr.11: 46753381-46753381 on Build GRCh38
rs768792551 Chr.11: 46753346-46753346 on Build GRCh38
rs374316828 Chr.11: 46753371-46753371 on Build GRCh38
rs767042226 Chr.11: 46753406-46753406 on Build GRCh38
rs560566891 Chr.11: 46753156-46753156 on Build GRCh38
rs768724616 Chr.11: 46753231-46753231 on Build GRCh38
rs111327819 Chr.11: 46753399-46753399 on Build GRCh38
rs750981919 Chr.11: 46753239-46753239 on Build GRCh38
rs754307028 Chr.11: 46753084-46753084 on Build GRCh38
rs372306286 Chr.11: 46753403-46753403 on Build GRCh38
rs778731229 Chr.11: 46753482-46753482 on Build GRCh38
rs763763033 Chr.11: 46753467-46753467 on Build GRCh38
rs1020782691 Chr.11: 46753468-46753468 on Build GRCh38
rs748209214 Chr.11: 46753369-46753369 on Build GRCh38
rs755725496 Chr.11: 46753285-46753285 on Build GRCh38
rs748038419 Chr.11: 46753203-46753203 on Build GRCh38
rs376318569 Chr.11: 46753472-46753472 on Build GRCh38
rs373354355 Chr.11: 46753159-46753159 on Build GRCh38
rs112678804 Chr.11: 46753268-46753268 on Build GRCh38
rs758262399 Chr.11: 46753495-46753495 on Build GRCh38
rs769924107 Chr.11: 46753370-46753370 on Build GRCh38
rs760870929 Chr.11: 46753224-46753224 on Build GRCh38
rs745620401 Chr.11: 46753483-46753483 on Build GRCh38
rs932316637 Chr.11: 46753127-46753127 on Build GRCh38
rs777667726 Chr.11: 46753210-46753210 on Build GRCh38
rs142490555 Chr.11: 46753106-46753106 on Build GRCh38
rs187486848 Chr.11: 46753202-46753202 on Build GRCh38
rs747346321 Chr.11: 46753275-46753276 on Build GRCh38
rs765538626 Chr.11: 46753116-46753116 on Build GRCh38
rs368214910 Chr.11: 46753377-46753377 on Build GRCh38
rs763458372 Chr.11: 46753374-46753374 on Build GRCh38
rs757166590 Chr.11: 46753473-46753473 on Build GRCh38
rs748440168 Chr.11: 46753505-46753505 on Build GRCh38
rs747207592 Chr.11: 46753303-46753303 on Build GRCh38
rs752446179 Chr.11: 46753407-46753407 on Build GRCh38
rs769871471 Chr.11: 46753205-46753205 on Build GRCh38
rs770679157 Chr.11: 46753212-46753212 on Build GRCh38

View Full Product Details