Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54118524-54119024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ12825
Entrez Gene ID
440101
Gene Name
uncharacterized LOC440101
Gene Aliases
-
UniGene
Hs.350378
Gene Chromosome Location
Chr.12: 54058254-54122234 on Build GRCh38
Transcripts
NR_026655.1

SNP Detail

SNP ID Chromosome Location
rs889110440 Chr.12: 54118734-54118734 on Build GRCh38
rs200775735 Chr.12: 54118770-54118771 on Build GRCh38
rs917591542 Chr.12: 54118872-54118872 on Build GRCh38
rs575746215 Chr.12: 54118718-54118718 on Build GRCh38
rs2370805 Chr.12: 54118992-54118992 on Build GRCh38
rs992272965 Chr.12: 54118843-54118843 on Build GRCh38
rs952230484 Chr.12: 54118796-54118796 on Build GRCh38
rs1033873908 Chr.12: 54118836-54118836 on Build GRCh38
rs993721205 Chr.12: 54118771-54118771 on Build GRCh38
rs141031039 Chr.12: 54118968-54118968 on Build GRCh38
rs192246354 Chr.12: 54118943-54118943 on Build GRCh38
rs1008008722 Chr.12: 54118764-54118764 on Build GRCh38
rs398055589 Chr.12: 54118773-54118774 on Build GRCh38
rs1054971890 Chr.12: 54118730-54118730 on Build GRCh38
rs559385219 Chr.12: 54118963-54118963 on Build GRCh38
rs1026410043 Chr.12: 54118772-54118772 on Build GRCh38
rs548462652 Chr.12: 54119001-54119001 on Build GRCh38
rs10876538 Chr.12: 54118599-54118599 on Build GRCh38
rs543174178 Chr.12: 54118794-54118794 on Build GRCh38
rs116928091 Chr.12: 54118842-54118842 on Build GRCh38
rs564957099 Chr.12: 54118805-54118805 on Build GRCh38
rs945241228 Chr.12: 54118922-54118922 on Build GRCh38
rs36075486 Chr.12: 54118771-54118772 on Build GRCh38
rs984536697 Chr.12: 54118832-54118832 on Build GRCh38
rs576393299 Chr.12: 54118956-54118956 on Build GRCh38
rs148148900 Chr.12: 54118618-54118618 on Build GRCh38
rs1019192767 Chr.12: 54118765-54118765 on Build GRCh38
rs563550157 Chr.12: 54118659-54118659 on Build GRCh38
rs200638094 Chr.12: 54118663-54118663 on Build GRCh38

View Full Product Details