Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54120066-54120574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ12825
Entrez Gene ID
440101
Gene Name
uncharacterized LOC440101
Gene Aliases
-
UniGene
Hs.350378
Gene Chromosome Location
Chr.12: 54058254-54122234 on Build GRCh38
Transcripts
NR_026655.1

SNP Detail

SNP ID Chromosome Location
rs1046990330 Chr.12: 54120549-54120549 on Build GRCh38
rs201102133 Chr.12: 54120383-54120383 on Build GRCh38
rs11170820 Chr.12: 54120131-54120131 on Build GRCh38
rs201504803 Chr.12: 54120439-54120439 on Build GRCh38
rs200945623 Chr.12: 54120434-54120434 on Build GRCh38
rs982377194 Chr.12: 54120385-54120385 on Build GRCh38
rs564309831 Chr.12: 54120531-54120531 on Build GRCh38
rs376014827 Chr.12: 54120301-54120301 on Build GRCh38
rs541876691 Chr.12: 54120315-54120315 on Build GRCh38
rs1018607251 Chr.12: 54120255-54120255 on Build GRCh38
rs760333125 Chr.12: 54120382-54120382 on Build GRCh38
rs935517484 Chr.12: 54120435-54120435 on Build GRCh38
rs189598070 Chr.12: 54120475-54120475 on Build GRCh38
rs900860727 Chr.12: 54120518-54120518 on Build GRCh38
rs199990556 Chr.12: 54120452-54120452 on Build GRCh38
rs373448761 Chr.12: 54120354-54120354 on Build GRCh38
rs75934050 Chr.12: 54120507-54120507 on Build GRCh38
rs376226814 Chr.12: 54120383-54120383 on Build GRCh38
rs991472427 Chr.12: 54120123-54120123 on Build GRCh38
rs371448392 Chr.12: 54120477-54120477 on Build GRCh38
rs935047583 Chr.12: 54120216-54120216 on Build GRCh38
rs368548493 Chr.12: 54120320-54120320 on Build GRCh38
rs1016383448 Chr.12: 54120339-54120339 on Build GRCh38
rs111388317 Chr.12: 54120207-54120207 on Build GRCh38
rs200768860 Chr.12: 54120478-54120478 on Build GRCh38
rs369232007 Chr.12: 54120118-54120118 on Build GRCh38
rs942448817 Chr.12: 54120476-54120476 on Build GRCh38
rs924085016 Chr.12: 54120416-54120416 on Build GRCh38
rs998067198 Chr.12: 54120281-54120281 on Build GRCh38
rs192819254 Chr.12: 54120234-54120234 on Build GRCh38
rs901059671 Chr.12: 54120266-54120266 on Build GRCh38
rs11170821 Chr.12: 54120216-54120216 on Build GRCh38
rs377157952 Chr.12: 54120107-54120107 on Build GRCh38
rs976685678 Chr.12: 54120137-54120148 on Build GRCh38
rs528243836 Chr.12: 54120547-54120547 on Build GRCh38
rs1039338276 Chr.12: 54120500-54120500 on Build GRCh38
rs902697920 Chr.12: 54120167-54120167 on Build GRCh38
rs963705497 Chr.12: 54120362-54120362 on Build GRCh38
rs182360604 Chr.12: 54120230-54120230 on Build GRCh38
rs563137127 Chr.12: 54120338-54120338 on Build GRCh38
rs184713459 Chr.12: 54120448-54120448 on Build GRCh38
rs372352403 Chr.12: 54120090-54120090 on Build GRCh38
rs747631539 Chr.12: 54120377-54120377 on Build GRCh38

View Full Product Details