Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50807728-50808234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEU7
Entrez Gene ID
220107
Gene Name
deleted in lymphocytic leukemia, 7
Gene Aliases
-
UniGene
Hs.673860
Gene Chromosome Location
Chr.13: 50711026-50843939 on Build GRCh38
Transcripts
NM_198989.3 NR_046551.1

Gene Symbol
DLEU7-AS1
Entrez Gene ID
100874074
Gene Name
DLEU7 antisense RNA 1
Gene Aliases
-
UniGene
Hs.734030
Gene Chromosome Location
Chr.13: 50807855-50849905 on Build GRCh38
Transcripts
NM_198989.3 NR_046551.1

SNP Detail

SNP ID Chromosome Location
rs539446198 Chr.13: 50807770-50807770 on Build GRCh38
rs646791 Chr.13: 50807840-50807840 on Build GRCh38
rs34131808 Chr.13: 50807954-50807955 on Build GRCh38
rs746125712 Chr.13: 50807772-50807772 on Build GRCh38
rs573849139 Chr.13: 50807894-50807894 on Build GRCh38
rs891459605 Chr.13: 50807863-50807863 on Build GRCh38
rs75516392 Chr.13: 50808053-50808053 on Build GRCh38
rs1044253013 Chr.13: 50807854-50807854 on Build GRCh38
rs993261899 Chr.13: 50808126-50808128 on Build GRCh38
rs182409980 Chr.13: 50807786-50807786 on Build GRCh38
rs141760059 Chr.13: 50807798-50807798 on Build GRCh38
rs533802787 Chr.13: 50807826-50807826 on Build GRCh38
rs904049466 Chr.13: 50807855-50807855 on Build GRCh38
rs74078447 Chr.13: 50807906-50807906 on Build GRCh38
rs41292762 Chr.13: 50807799-50807799 on Build GRCh38
rs972624459 Chr.13: 50807982-50807982 on Build GRCh38
rs1051182566 Chr.13: 50807827-50807827 on Build GRCh38
rs879270980 Chr.13: 50807962-50807962 on Build GRCh38
rs775929903 Chr.13: 50807918-50807918 on Build GRCh38
rs1014302607 Chr.13: 50807898-50807898 on Build GRCh38
rs9535491 Chr.13: 50807970-50807970 on Build GRCh38
rs999759871 Chr.13: 50807861-50807861 on Build GRCh38
rs968890951 Chr.13: 50808051-50808051 on Build GRCh38
rs1023976877 Chr.13: 50807904-50807904 on Build GRCh38
rs927369345 Chr.13: 50808059-50808059 on Build GRCh38
rs955292583 Chr.13: 50808139-50808139 on Build GRCh38
rs911236597 Chr.13: 50808206-50808206 on Build GRCh38
rs958809719 Chr.13: 50808072-50808072 on Build GRCh38
rs970188691 Chr.13: 50807905-50807905 on Build GRCh38
rs368785519 Chr.13: 50808159-50808159 on Build GRCh38
rs1009016558 Chr.13: 50807880-50807880 on Build GRCh38
rs962575279 Chr.13: 50807784-50807784 on Build GRCh38
rs934034966 Chr.13: 50807819-50807819 on Build GRCh38
rs987181132 Chr.13: 50808153-50808153 on Build GRCh38
rs1020517789 Chr.13: 50808037-50808037 on Build GRCh38
rs940659580 Chr.13: 50808169-50808169 on Build GRCh38
rs1052726584 Chr.13: 50807862-50807862 on Build GRCh38
rs911255617 Chr.13: 50807839-50807839 on Build GRCh38

View Full Product Details