Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 97926280-97926788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01550
Entrez Gene ID
388011
Gene Name
long intergenic non-protein coding RNA 1550
Gene Aliases
C14orf64
UniGene
Hs.651477
Gene Chromosome Location
Chr.14: 97925610-97978124 on Build GRCh38
Transcripts
NR_015430.1

SNP Detail

SNP ID Chromosome Location
rs536369921 Chr.14: 97926481-97926481 on Build GRCh38
rs563354479 Chr.14: 97926668-97926668 on Build GRCh38
rs1043182565 Chr.14: 97926599-97926599 on Build GRCh38
rs905616301 Chr.14: 97926532-97926532 on Build GRCh38
rs144987043 Chr.14: 97926672-97926672 on Build GRCh38
rs112012263 Chr.14: 97926492-97926492 on Build GRCh38
rs757211650 Chr.14: 97926538-97926538 on Build GRCh38
rs778952960 Chr.14: 97926584-97926584 on Build GRCh38
rs899930513 Chr.14: 97926341-97926341 on Build GRCh38
rs1045404627 Chr.14: 97926531-97926531 on Build GRCh38
rs908141191 Chr.14: 97926525-97926525 on Build GRCh38
rs920694779 Chr.14: 97926685-97926685 on Build GRCh38
rs974791074 Chr.14: 97926680-97926680 on Build GRCh38
rs899960514 Chr.14: 97926548-97926548 on Build GRCh38
rs991333190 Chr.14: 97926463-97926463 on Build GRCh38
rs987514921 Chr.14: 97926449-97926449 on Build GRCh38
rs1048421386 Chr.14: 97926520-97926520 on Build GRCh38
rs34439432 Chr.14: 97926638-97926639 on Build GRCh38
rs1035647273 Chr.14: 97926446-97926446 on Build GRCh38
rs1047978452 Chr.14: 97926337-97926337 on Build GRCh38
rs1054138290 Chr.14: 97926540-97926540 on Build GRCh38
rs149090417 Chr.14: 97926708-97926708 on Build GRCh38
rs544959481 Chr.14: 97926643-97926643 on Build GRCh38
rs143163632 Chr.14: 97926735-97926735 on Build GRCh38
rs1023227102 Chr.14: 97926678-97926678 on Build GRCh38
rs1010091894 Chr.14: 97926617-97926617 on Build GRCh38
rs192356875 Chr.14: 97926424-97926424 on Build GRCh38
rs1039870078 Chr.14: 97926541-97926541 on Build GRCh38
rs957201324 Chr.14: 97926420-97926420 on Build GRCh38
rs367804270 Chr.14: 97926701-97926701 on Build GRCh38
rs115828368 Chr.14: 97926526-97926526 on Build GRCh38
rs1039837763 Chr.14: 97926328-97926328 on Build GRCh38
rs967092391 Chr.14: 97926433-97926433 on Build GRCh38
rs963275527 Chr.14: 97926696-97926696 on Build GRCh38
rs1008895783 Chr.14: 97926640-97926640 on Build GRCh38
rs1020604647 Chr.14: 97926618-97926618 on Build GRCh38
rs950367833 Chr.14: 97926737-97926737 on Build GRCh38
rs753745040 Chr.14: 97926518-97926518 on Build GRCh38
rs528303389 Chr.14: 97926330-97926330 on Build GRCh38
rs561259463 Chr.14: 97926715-97926715 on Build GRCh38
rs538592129 Chr.14: 97926377-97926377 on Build GRCh38
rs183065217 Chr.14: 97926482-97926482 on Build GRCh38
rs915721644 Chr.14: 97926464-97926464 on Build GRCh38
rs568233373 Chr.14: 97926395-97926395 on Build GRCh38
rs80340345 Chr.14: 97926729-97926729 on Build GRCh38
rs571608093 Chr.14: 97926362-97926362 on Build GRCh38
rs763813224 Chr.14: 97926498-97926498 on Build GRCh38
rs926925511 Chr.14: 97926753-97926753 on Build GRCh38
rs2925642 Chr.14: 97926500-97926500 on Build GRCh38
rs1032845573 Chr.14: 97926396-97926396 on Build GRCh38
rs1020215875 Chr.14: 97926427-97926427 on Build GRCh38
rs557007640 Chr.14: 97926382-97926382 on Build GRCh38
rs959932671 Chr.14: 97926461-97926461 on Build GRCh38
rs541305464 Chr.14: 97926511-97926511 on Build GRCh38
rs1023989226 Chr.14: 97926372-97926372 on Build GRCh38
rs918992355 Chr.14: 97926505-97926505 on Build GRCh38
rs565137019 Chr.14: 97926741-97926741 on Build GRCh38
rs147900274 Chr.14: 97926343-97926343 on Build GRCh38
rs959962511 Chr.14: 97926646-97926646 on Build GRCh38
rs944267747 Chr.14: 97926534-97926534 on Build GRCh38
rs149926053 Chr.14: 97926631-97926631 on Build GRCh38
rs944298446 Chr.14: 97926755-97926755 on Build GRCh38
rs752425643 Chr.14: 97926375-97926375 on Build GRCh38

View Full Product Details