Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 97977838-97978337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01550
Entrez Gene ID
388011
Gene Name
long intergenic non-protein coding RNA 1550
Gene Aliases
C14orf64
UniGene
Hs.651477
Gene Chromosome Location
Chr.14: 97925610-97978124 on Build GRCh38
Transcripts
NR_015430.1

SNP Detail

SNP ID Chromosome Location
rs780188868 Chr.14: 97978107-97978107 on Build GRCh38
rs986438885 Chr.14: 97977933-97977933 on Build GRCh38
rs924094183 Chr.14: 97978221-97978221 on Build GRCh38
rs758226906 Chr.14: 97977878-97977878 on Build GRCh38
rs1025361360 Chr.14: 97977919-97977919 on Build GRCh38
rs767946860 Chr.14: 97978086-97978086 on Build GRCh38
rs981318558 Chr.14: 97978023-97978023 on Build GRCh38
rs781556589 Chr.14: 97977986-97977986 on Build GRCh38
rs948032118 Chr.14: 97978065-97978065 on Build GRCh38
rs763659790 Chr.14: 97978028-97978028 on Build GRCh38
rs1056415118 Chr.14: 97978041-97978041 on Build GRCh38
rs964046439 Chr.14: 97978132-97978132 on Build GRCh38
rs755167350 Chr.14: 97977965-97977965 on Build GRCh38
rs112907746 Chr.14: 97978101-97978101 on Build GRCh38
rs571033346 Chr.14: 97978109-97978109 on Build GRCh38
rs542739179 Chr.14: 97978139-97978139 on Build GRCh38
rs1022934526 Chr.14: 97978161-97978161 on Build GRCh38
rs759333999 Chr.14: 97977951-97977951 on Build GRCh38
rs779122081 Chr.14: 97978141-97978141 on Build GRCh38
rs994375533 Chr.14: 97978080-97978080 on Build GRCh38
rs560028132 Chr.14: 97978177-97978177 on Build GRCh38
rs8016584 Chr.14: 97978032-97978032 on Build GRCh38
rs968257648 Chr.14: 97978171-97978171 on Build GRCh38
rs915335727 Chr.14: 97978076-97978076 on Build GRCh38
rs915450218 Chr.14: 97977958-97977958 on Build GRCh38
rs1042485248 Chr.14: 97977993-97977993 on Build GRCh38
rs971312951 Chr.14: 97978019-97978019 on Build GRCh38
rs12890382 Chr.14: 97977948-97977948 on Build GRCh38
rs944961698 Chr.14: 97978136-97978136 on Build GRCh38
rs916620703 Chr.14: 97978291-97978291 on Build GRCh38
rs539991161 Chr.14: 97978261-97978261 on Build GRCh38
rs931676173 Chr.14: 97978164-97978164 on Build GRCh38
rs959332056 Chr.14: 97978043-97978043 on Build GRCh38
rs573806350 Chr.14: 97977980-97977980 on Build GRCh38
rs115940378 Chr.14: 97977907-97977907 on Build GRCh38
rs1012560655 Chr.14: 97977888-97977888 on Build GRCh38
rs992197093 Chr.14: 97978287-97978287 on Build GRCh38
rs377431663 Chr.14: 97978118-97978118 on Build GRCh38
rs1042540818 Chr.14: 97977868-97977868 on Build GRCh38
rs762414580 Chr.14: 97977994-97977994 on Build GRCh38
rs939467658 Chr.14: 97978251-97978251 on Build GRCh38
rs750819039 Chr.14: 97978098-97978098 on Build GRCh38
rs923999295 Chr.14: 97978011-97978011 on Build GRCh38
rs990823643 Chr.14: 97978058-97978058 on Build GRCh38
rs376538680 Chr.14: 97978044-97978044 on Build GRCh38
rs900188222 Chr.14: 97978140-97978140 on Build GRCh38
rs8015127 Chr.14: 97977916-97977916 on Build GRCh38
rs902802824 Chr.14: 97977877-97977877 on Build GRCh38
rs1038693016 Chr.14: 97978067-97978067 on Build GRCh38
rs955035488 Chr.14: 97977929-97977929 on Build GRCh38
rs895138389 Chr.14: 97978051-97978051 on Build GRCh38
rs749494199 Chr.14: 97978126-97978126 on Build GRCh38
rs551308563 Chr.14: 97977895-97977895 on Build GRCh38
rs764825065 Chr.14: 97977952-97977952 on Build GRCh38

View Full Product Details