Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89850209-89850713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3S2
Entrez Gene ID
10239
Gene Name
adaptor related protein complex 3 sigma 2 subunit
Gene Aliases
AP3S3, sigma3b
UniGene
Hs.632161
Gene Chromosome Location
Chr.15: 89830599-89894385 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1 NR_049817.1

Gene Symbol
C15orf38-AP3S2
Entrez Gene ID
100526783
Gene Name
C15orf38-AP3S2 readthrough
Gene Aliases
-
UniGene
Hs.744890
Gene Chromosome Location
Chr.15: 89830599-89912990 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1 NR_049817.1

Gene Symbol
MIR5094
Entrez Gene ID
100847059
Gene Name
microRNA 5094
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.15: 89850637-89850721 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1 NR_049817.1

SNP Detail

SNP ID Chromosome Location
rs578262569 Chr.15: 89850252-89850252 on Build GRCh38
rs965807453 Chr.15: 89850442-89850442 on Build GRCh38
rs924279020 Chr.15: 89850330-89850330 on Build GRCh38
rs185355265 Chr.15: 89850408-89850408 on Build GRCh38
rs977016952 Chr.15: 89850326-89850326 on Build GRCh38
rs747140307 Chr.15: 89850437-89850437 on Build GRCh38
rs948891748 Chr.15: 89850353-89850353 on Build GRCh38
rs4932252 Chr.15: 89850576-89850576 on Build GRCh38
rs922303981 Chr.15: 89850348-89850348 on Build GRCh38
rs116839953 Chr.15: 89850239-89850239 on Build GRCh38
rs543199576 Chr.15: 89850483-89850483 on Build GRCh38
rs928783842 Chr.15: 89850521-89850521 on Build GRCh38
rs114848378 Chr.15: 89850361-89850361 on Build GRCh38
rs898484309 Chr.15: 89850381-89850381 on Build GRCh38
rs1044480313 Chr.15: 89850463-89850463 on Build GRCh38
rs765015166 Chr.15: 89850329-89850329 on Build GRCh38
rs985143437 Chr.15: 89850626-89850626 on Build GRCh38
rs1007429296 Chr.15: 89850655-89850655 on Build GRCh38
rs916086956 Chr.15: 89850349-89850349 on Build GRCh38
rs939285191 Chr.15: 89850407-89850407 on Build GRCh38
rs948905185 Chr.15: 89850496-89850496 on Build GRCh38
rs766872159 Chr.15: 89850310-89850313 on Build GRCh38
rs758307596 Chr.15: 89850336-89850336 on Build GRCh38
rs1005698043 Chr.15: 89850246-89850246 on Build GRCh38
rs990826084 Chr.15: 89850485-89850485 on Build GRCh38
rs373760381 Chr.15: 89850297-89850297 on Build GRCh38
rs796682616 Chr.15: 89850368-89850368 on Build GRCh38
rs528957071 Chr.15: 89850580-89850580 on Build GRCh38
rs575961792 Chr.15: 89850443-89850443 on Build GRCh38
rs995188550 Chr.15: 89850391-89850391 on Build GRCh38
rs371768995 Chr.15: 89850665-89850665 on Build GRCh38
rs1016152777 Chr.15: 89850323-89850323 on Build GRCh38
rs193245460 Chr.15: 89850244-89850244 on Build GRCh38
rs1021941300 Chr.15: 89850643-89850643 on Build GRCh38
rs940004668 Chr.15: 89850372-89850372 on Build GRCh38
rs953222175 Chr.15: 89850279-89850279 on Build GRCh38
rs1018769183 Chr.15: 89850661-89850661 on Build GRCh38
rs965691325 Chr.15: 89850323-89850323 on Build GRCh38
rs770348696 Chr.15: 89850604-89850604 on Build GRCh38
rs940119436 Chr.15: 89850535-89850535 on Build GRCh38
rs916136958 Chr.15: 89850495-89850495 on Build GRCh38
rs907249990 Chr.15: 89850364-89850364 on Build GRCh38
rs780034212 Chr.15: 89850392-89850392 on Build GRCh38
rs754962846 Chr.15: 89850472-89850472 on Build GRCh38
rs138561338 Chr.15: 89850462-89850462 on Build GRCh38
rs1056307856 Chr.15: 89850411-89850411 on Build GRCh38
rs1007359430 Chr.15: 89850419-89850419 on Build GRCh38
rs986304881 Chr.15: 89850289-89850289 on Build GRCh38
rs546656511 Chr.15: 89850635-89850635 on Build GRCh38
rs1037072092 Chr.15: 89850376-89850376 on Build GRCh38
rs781696202 Chr.15: 89850543-89850544 on Build GRCh38
rs557624786 Chr.15: 89850335-89850335 on Build GRCh38
rs888999214 Chr.15: 89850639-89850639 on Build GRCh38
rs1027931754 Chr.15: 89850249-89850249 on Build GRCh38
rs898510248 Chr.15: 89850590-89850590 on Build GRCh38
rs781489831 Chr.15: 89850501-89850501 on Build GRCh38
rs1031326678 Chr.15: 89850478-89850478 on Build GRCh38

View Full Product Details