Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44182740-44183248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB16-AS1
Entrez Gene ID
339201
Gene Name
ASB16 antisense RNA 1
Gene Aliases
C17orf65
UniGene
Hs.656564
Gene Chromosome Location
Chr.17: 44175973-44186717 on Build GRCh38
Transcripts
NR_049729.1 NR_049730.1

SNP Detail

SNP ID Chromosome Location
rs567452877 Chr.17: 44182840-44182840 on Build GRCh38
rs970997398 Chr.17: 44183087-44183087 on Build GRCh38
rs541757163 Chr.17: 44182766-44182766 on Build GRCh38
rs190534438 Chr.17: 44182970-44182970 on Build GRCh38
rs530397492 Chr.17: 44182833-44182833 on Build GRCh38
rs926415355 Chr.17: 44183112-44183112 on Build GRCh38
rs186163404 Chr.17: 44182893-44182893 on Build GRCh38
rs914922976 Chr.17: 44183207-44183207 on Build GRCh38
rs981819208 Chr.17: 44183097-44183097 on Build GRCh38
rs943586738 Chr.17: 44182831-44182831 on Build GRCh38
rs1017525039 Chr.17: 44183063-44183063 on Build GRCh38
rs926914384 Chr.17: 44183225-44183225 on Build GRCh38
rs773560894 Chr.17: 44182775-44182775 on Build GRCh38
rs1000077907 Chr.17: 44182996-44182996 on Build GRCh38
rs964944732 Chr.17: 44183069-44183069 on Build GRCh38
rs537404989 Chr.17: 44183020-44183020 on Build GRCh38
rs949744097 Chr.17: 44183173-44183173 on Build GRCh38
rs538829433 Chr.17: 44182773-44182774 on Build GRCh38
rs910803820 Chr.17: 44182816-44182816 on Build GRCh38
rs558919215 Chr.17: 44183075-44183075 on Build GRCh38
rs150099571 Chr.17: 44182850-44182850 on Build GRCh38
rs562924997 Chr.17: 44182773-44182773 on Build GRCh38
rs920731595 Chr.17: 44182852-44182852 on Build GRCh38
rs138522260 Chr.17: 44182940-44182940 on Build GRCh38
rs949135163 Chr.17: 44182906-44182910 on Build GRCh38
rs916982307 Chr.17: 44183143-44183143 on Build GRCh38
rs528196527 Chr.17: 44182855-44182855 on Build GRCh38
rs760421434 Chr.17: 44183079-44183080 on Build GRCh38
rs1021451645 Chr.17: 44183025-44183025 on Build GRCh38
rs919872732 Chr.17: 44182851-44182851 on Build GRCh38
rs996888261 Chr.17: 44183021-44183021 on Build GRCh38
rs1044090378 Chr.17: 44183186-44183186 on Build GRCh38
rs536395990 Chr.17: 44182875-44182875 on Build GRCh38
rs1054305635 Chr.17: 44183010-44183010 on Build GRCh38
rs954862864 Chr.17: 44182783-44182783 on Build GRCh38
rs950934778 Chr.17: 44183107-44183107 on Build GRCh38
rs920797020 Chr.17: 44183092-44183092 on Build GRCh38
rs570381761 Chr.17: 44182995-44182995 on Build GRCh38
rs1044372991 Chr.17: 44182950-44182950 on Build GRCh38
rs56338108 Chr.17: 44183032-44183032 on Build GRCh38
rs1046595507 Chr.17: 44182911-44182915 on Build GRCh38
rs553116084 Chr.17: 44182934-44182934 on Build GRCh38
rs983253684 Chr.17: 44183111-44183111 on Build GRCh38
rs769344164 Chr.17: 44182880-44182880 on Build GRCh38
rs987718313 Chr.17: 44182784-44182784 on Build GRCh38
rs891598348 Chr.17: 44183019-44183019 on Build GRCh38
rs949690825 Chr.17: 44182944-44182944 on Build GRCh38
rs903370611 Chr.17: 44182976-44182976 on Build GRCh38
rs904880617 Chr.17: 44182772-44182773 on Build GRCh38
rs1019736974 Chr.17: 44183014-44183014 on Build GRCh38
rs1008857598 Chr.17: 44183056-44183056 on Build GRCh38
rs937765532 Chr.17: 44183125-44183125 on Build GRCh38
rs926279565 Chr.17: 44182923-44182923 on Build GRCh38

View Full Product Details