Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81181274-81181764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AATK-AS1
Entrez Gene ID
388428
Gene Name
AATK antisense RNA 1
Gene Aliases
-
UniGene
Hs.591202
Gene Chromosome Location
Chr.17: 81165507-81183164 on Build GRCh38
Transcripts
NR_027255.1

SNP Detail

SNP ID Chromosome Location
rs930622416 Chr.17: 81181359-81181359 on Build GRCh38
rs891030821 Chr.17: 81181581-81181581 on Build GRCh38
rs376234109 Chr.17: 81181354-81181354 on Build GRCh38
rs889809079 Chr.17: 81181461-81181461 on Build GRCh38
rs931847039 Chr.17: 81181744-81181744 on Build GRCh38
rs556479179 Chr.17: 81181315-81181315 on Build GRCh38
rs185241415 Chr.17: 81181454-81181454 on Build GRCh38
rs963878155 Chr.17: 81181595-81181595 on Build GRCh38
rs572471178 Chr.17: 81181372-81181372 on Build GRCh38
rs906180 Chr.17: 81181316-81181316 on Build GRCh38
rs79963994 Chr.17: 81181637-81181637 on Build GRCh38
rs977988339 Chr.17: 81181720-81181720 on Build GRCh38
rs114895555 Chr.17: 81181641-81181641 on Build GRCh38
rs770802874 Chr.17: 81181636-81181636 on Build GRCh38
rs955208942 Chr.17: 81181444-81181444 on Build GRCh38
rs910998171 Chr.17: 81181448-81181448 on Build GRCh38
rs895269498 Chr.17: 81181528-81181528 on Build GRCh38
rs749002570 Chr.17: 81181351-81181351 on Build GRCh38
rs544420636 Chr.17: 81181443-81181443 on Build GRCh38
rs1049511286 Chr.17: 81181491-81181491 on Build GRCh38
rs554102640 Chr.17: 81181361-81181361 on Build GRCh38
rs950261179 Chr.17: 81181347-81181347 on Build GRCh38
rs769331170 Chr.17: 81181329-81181338 on Build GRCh38
rs917379668 Chr.17: 81181468-81181468 on Build GRCh38
rs369396132 Chr.17: 81181584-81181584 on Build GRCh38
rs558333378 Chr.17: 81181715-81181715 on Build GRCh38
rs562646165 Chr.17: 81181447-81181447 on Build GRCh38
rs977045088 Chr.17: 81181614-81181614 on Build GRCh38
rs898552251 Chr.17: 81181709-81181709 on Build GRCh38
rs948206403 Chr.17: 81181687-81181687 on Build GRCh38
rs376388581 Chr.17: 81181295-81181295 on Build GRCh38
rs747681559 Chr.17: 81181575-81181575 on Build GRCh38
rs923335697 Chr.17: 81181732-81181732 on Build GRCh38
rs548858501 Chr.17: 81181356-81181356 on Build GRCh38
rs113860147 Chr.17: 81181594-81181594 on Build GRCh38
rs35208109 Chr.17: 81181715-81181716 on Build GRCh38
rs924216986 Chr.17: 81181623-81181623 on Build GRCh38
rs369262430 Chr.17: 81181590-81181590 on Build GRCh38
rs917515455 Chr.17: 81181341-81181341 on Build GRCh38
rs930676664 Chr.17: 81181490-81181490 on Build GRCh38
rs568699045 Chr.17: 81181365-81181365 on Build GRCh38
rs964132422 Chr.17: 81181451-81181451 on Build GRCh38
rs756937632 Chr.17: 81181647-81181647 on Build GRCh38
rs112414438 Chr.17: 81181305-81181305 on Build GRCh38
rs971619706 Chr.17: 81181677-81181677 on Build GRCh38
rs550302309 Chr.17: 81181589-81181589 on Build GRCh38
rs1028299935 Chr.17: 81181452-81181452 on Build GRCh38
rs770600033 Chr.17: 81181352-81181352 on Build GRCh38
rs1018750434 Chr.17: 81181420-81181420 on Build GRCh38
rs1024676606 Chr.17: 81181675-81181675 on Build GRCh38
rs962653465 Chr.17: 81181369-81181369 on Build GRCh38
rs868373366 Chr.17: 81181544-81181544 on Build GRCh38
rs74459181 Chr.17: 81181712-81181712 on Build GRCh38
rs757549282 Chr.17: 81181364-81181364 on Build GRCh38
rs924167969 Chr.17: 81181471-81181471 on Build GRCh38
rs994145156 Chr.17: 81181558-81181558 on Build GRCh38
rs183203435 Chr.17: 81181423-81181423 on Build GRCh38
rs1022239594 Chr.17: 81181706-81181706 on Build GRCh38
rs1021308933 Chr.17: 81181610-81181610 on Build GRCh38
rs1029323777 Chr.17: 81181441-81181441 on Build GRCh38
rs1030763703 Chr.17: 81181450-81181450 on Build GRCh38
rs546348057 Chr.17: 81181378-81181378 on Build GRCh38
rs934858421 Chr.17: 81181310-81181310 on Build GRCh38
rs372074596 Chr.17: 81181478-81181478 on Build GRCh38
rs76435140 Chr.17: 81181585-81181585 on Build GRCh38
rs769641470 Chr.17: 81181597-81181597 on Build GRCh38
rs980650029 Chr.17: 81181680-81181680 on Build GRCh38
rs1038421642 Chr.17: 81181536-81181536 on Build GRCh38
rs1040506396 Chr.17: 81181717-81181717 on Build GRCh38
rs746322988 Chr.17: 81181379-81181379 on Build GRCh38
rs377106235 Chr.17: 81181440-81181440 on Build GRCh38
rs376294044 Chr.17: 81181534-81181534 on Build GRCh38
rs551886119 Chr.17: 81181527-81181527 on Build GRCh38
rs547889989 Chr.17: 81181456-81181456 on Build GRCh38
rs927588436 Chr.17: 81181682-81181682 on Build GRCh38
rs939600704 Chr.17: 81181626-81181626 on Build GRCh38
rs762857187 Chr.17: 81181611-81181611 on Build GRCh38
rs772602446 Chr.17: 81181417-81181417 on Build GRCh38
rs777525513 Chr.17: 81181331-81181331 on Build GRCh38
rs1017343494 Chr.17: 81181368-81181368 on Build GRCh38
rs536118308 Chr.17: 81181605-81181605 on Build GRCh38
rs113556535 Chr.17: 81181470-81181470 on Build GRCh38
rs767462863 Chr.17: 81181746-81181746 on Build GRCh38
rs527269293 Chr.17: 81181494-81181494 on Build GRCh38
rs1011221424 Chr.17: 81181495-81181495 on Build GRCh38
rs945678708 Chr.17: 81181469-81181469 on Build GRCh38
rs374102639 Chr.17: 81181359-81181359 on Build GRCh38

View Full Product Details